Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151742864-151743368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA2
Entrez Gene ID
1260
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
CNCA, CNCA1, CNG2, OCNC1, OCNCALPHA, OCNCa
UniGene
Hs.447360
Gene Chromosome Location
Chr.X: 151734746-151745564 on Build GRCh38
Transcripts
NM_005140.1

SNP Detail

SNP ID Chromosome Location
rs375317479 Chr.X: 151743345-151743345 on Build GRCh38
rs868773716 Chr.X: 151742997-151742997 on Build GRCh38
rs372211404 Chr.X: 151742994-151742994 on Build GRCh38
rs867359982 Chr.X: 151742988-151742988 on Build GRCh38
rs867608783 Chr.X: 151742984-151742984 on Build GRCh38
rs373303653 Chr.X: 151743139-151743139 on Build GRCh38
rs865818644 Chr.X: 151742938-151742938 on Build GRCh38
rs140376001 Chr.X: 151743343-151743343 on Build GRCh38
rs754448035 Chr.X: 151743267-151743267 on Build GRCh38
rs370205402 Chr.X: 151743330-151743330 on Build GRCh38
rs770767034 Chr.X: 151743302-151743302 on Build GRCh38
rs940027538 Chr.X: 151742954-151742954 on Build GRCh38
rs56329955 Chr.X: 151742971-151742971 on Build GRCh38
rs151204211 Chr.X: 151743324-151743324 on Build GRCh38
rs142147022 Chr.X: 151742937-151742937 on Build GRCh38
rs373219141 Chr.X: 151743059-151743059 on Build GRCh38
rs867745603 Chr.X: 151742940-151742940 on Build GRCh38
rs867254478 Chr.X: 151742944-151742944 on Build GRCh38
rs867845623 Chr.X: 151743166-151743166 on Build GRCh38
rs764432677 Chr.X: 151743105-151743105 on Build GRCh38
rs879605804 Chr.X: 151743040-151743041 on Build GRCh38
rs771999167 Chr.X: 151743323-151743323 on Build GRCh38
rs112354647 Chr.X: 151742948-151742948 on Build GRCh38
rs748747201 Chr.X: 151743266-151743266 on Build GRCh38
rs867003932 Chr.X: 151742949-151742949 on Build GRCh38
rs766958484 Chr.X: 151743242-151743242 on Build GRCh38
rs747811111 Chr.X: 151743156-151743156 on Build GRCh38
rs777699828 Chr.X: 151743007-151743007 on Build GRCh38
rs376742825 Chr.X: 151743071-151743071 on Build GRCh38
rs1041570194 Chr.X: 151743293-151743293 on Build GRCh38
rs1025799922 Chr.X: 151743064-151743064 on Build GRCh38
rs760489873 Chr.X: 151743191-151743191 on Build GRCh38
rs753169347 Chr.X: 151743315-151743315 on Build GRCh38
rs942380316 Chr.X: 151743158-151743158 on Build GRCh38
rs371630210 Chr.X: 151743109-151743109 on Build GRCh38
rs776563648 Chr.X: 151743312-151743312 on Build GRCh38
rs370940957 Chr.X: 151742974-151742974 on Build GRCh38
rs142147240 Chr.X: 151743124-151743124 on Build GRCh38
rs776985518 Chr.X: 151742926-151742927 on Build GRCh38
rs868519016 Chr.X: 151743050-151743050 on Build GRCh38
rs201870506 Chr.X: 151743243-151743243 on Build GRCh38
rs5924687 Chr.X: 151743041-151743041 on Build GRCh38
rs866331249 Chr.X: 151742990-151742990 on Build GRCh38
rs746065892 Chr.X: 151742927-151742928 on Build GRCh38
rs143915577 Chr.X: 151743100-151743100 on Build GRCh38
rs187014401 Chr.X: 151743299-151743299 on Build GRCh38
rs778945457 Chr.X: 151743051-151743052 on Build GRCh38
rs751721587 Chr.X: 151743228-151743228 on Build GRCh38
rs375835366 Chr.X: 151742991-151742994 on Build GRCh38
rs779452392 Chr.X: 151742996-151742996 on Build GRCh38
rs5924686 Chr.X: 151743040-151743040 on Build GRCh38
rs866062595 Chr.X: 151743036-151743036 on Build GRCh38
rs1057519072 Chr.X: 151743137-151743137 on Build GRCh38
rs947321075 Chr.X: 151743292-151743292 on Build GRCh38
rs758741001 Chr.X: 151743006-151743009 on Build GRCh38
rs199883231 Chr.X: 151743013-151743028 on Build GRCh38
rs865808351 Chr.X: 151742995-151742995 on Build GRCh38
rs757491877 Chr.X: 151743236-151743236 on Build GRCh38
rs371376202 Chr.X: 151743023-151743023 on Build GRCh38
rs868154503 Chr.X: 151743038-151743038 on Build GRCh38
rs771939174 Chr.X: 151743183-151743183 on Build GRCh38
rs755223188 Chr.X: 151743048-151743049 on Build GRCh38
rs373871680 Chr.X: 151742969-151742972 on Build GRCh38
rs368441177 Chr.X: 151742992-151742992 on Build GRCh38
rs200747674 Chr.X: 151743011-151743011 on Build GRCh38
rs376038112 Chr.X: 151743025-151743025 on Build GRCh38
rs369307379 Chr.X: 151743027-151743027 on Build GRCh38
rs867766776 Chr.X: 151742946-151742946 on Build GRCh38
rs747179226 Chr.X: 151743009-151743009 on Build GRCh38
rs865917864 Chr.X: 151742929-151742929 on Build GRCh38
rs751915665 Chr.X: 151743106-151743106 on Build GRCh38
rs761332883 Chr.X: 151743154-151743154 on Build GRCh38
rs1024324726 Chr.X: 151742927-151742927 on Build GRCh38
rs752938539 Chr.X: 151743000-151743000 on Build GRCh38
rs868041337 Chr.X: 151742935-151742935 on Build GRCh38
rs765430192 Chr.X: 151743005-151743006 on Build GRCh38
rs111801834 Chr.X: 151742942-151742942 on Build GRCh38
rs148670521 Chr.X: 151743119-151743119 on Build GRCh38
rs770920130 Chr.X: 151743016-151743016 on Build GRCh38
rs773164303 Chr.X: 151743189-151743189 on Build GRCh38
rs763157685 Chr.X: 151743223-151743223 on Build GRCh38
rs758687932 Chr.X: 151743316-151743316 on Build GRCh38
rs56348162 Chr.X: 151743018-151743018 on Build GRCh38
rs780670790 Chr.X: 151743248-151743248 on Build GRCh38
rs377750886 Chr.X: 151742971-151742972 on Build GRCh38
rs374298377 Chr.X: 151742976-151742976 on Build GRCh38
rs866975344 Chr.X: 151743051-151743051 on Build GRCh38
rs767825547 Chr.X: 151743237-151743237 on Build GRCh38
rs756672330 Chr.X: 151743138-151743138 on Build GRCh38
rs960998062 Chr.X: 151742921-151742921 on Build GRCh38
rs768183199 Chr.X: 151743148-151743148 on Build GRCh38
rs374586764 Chr.X: 151743005-151743005 on Build GRCh38
rs746872122 Chr.X: 151743300-151743300 on Build GRCh38
rs764161862 Chr.X: 151743224-151743224 on Build GRCh38
rs868677496 Chr.X: 151743039-151743039 on Build GRCh38
rs748875319 Chr.X: 151743143-151743143 on Build GRCh38
rs866587569 Chr.X: 151742986-151742986 on Build GRCh38
rs766381419 Chr.X: 151743196-151743196 on Build GRCh38
rs111772081 Chr.X: 151743039-151743040 on Build GRCh38
rs55650700 Chr.X: 151743028-151743028 on Build GRCh38
rs866424112 Chr.X: 151742985-151742985 on Build GRCh38
rs747754116 Chr.X: 151743287-151743287 on Build GRCh38
rs771882154 Chr.X: 151743288-151743288 on Build GRCh38
rs986523225 Chr.X: 151743137-151743137 on Build GRCh38
rs775411517 Chr.X: 151743197-151743197 on Build GRCh38
rs35955120 Chr.X: 151742983-151742984 on Build GRCh38
rs56348525 Chr.X: 151742972-151742972 on Build GRCh38
rs34829464 Chr.X: 151742925-151742926 on Build GRCh38
rs376567084 Chr.X: 151742970-151742970 on Build GRCh38
rs375492989 Chr.X: 151743269-151743269 on Build GRCh38
rs766845667 Chr.X: 151742983-151742983 on Build GRCh38
rs369046832 Chr.X: 151742956-151742957 on Build GRCh38
rs1012470599 Chr.X: 151742922-151742922 on Build GRCh38
rs752854018 Chr.X: 151743088-151743088 on Build GRCh38
rs370968732 Chr.X: 151742982-151742983 on Build GRCh38

View Full Product Details