Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151955286-151955787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRE
Entrez Gene ID
2564
Gene Name
gamma-aminobutyric acid type A receptor epsilon subunit
Gene Aliases
-
UniGene
Hs.22785
Gene Chromosome Location
Chr.X: 151953124-151974679 on Build GRCh38
Transcripts
NM_004961.3 XM_011531140.2 XM_017029386.1 XM_017029387.1 XM_017029388.1 XM_017029389.1

SNP Detail

SNP ID Chromosome Location
rs752429085 Chr.X: 151955513-151955513 on Build GRCh38
rs141069947 Chr.X: 151955371-151955371 on Build GRCh38
rs77007790 Chr.X: 151955549-151955549 on Build GRCh38
rs755007870 Chr.X: 151955729-151955729 on Build GRCh38
rs188862502 Chr.X: 151955447-151955447 on Build GRCh38
rs147982495 Chr.X: 151955672-151955672 on Build GRCh38
rs961721530 Chr.X: 151955739-151955739 on Build GRCh38
rs758332176 Chr.X: 151955522-151955522 on Build GRCh38
rs775923936 Chr.X: 151955356-151955356 on Build GRCh38
rs147565446 Chr.X: 151955472-151955472 on Build GRCh38
rs367673721 Chr.X: 151955685-151955685 on Build GRCh38
rs61740527 Chr.X: 151955445-151955445 on Build GRCh38
rs753890768 Chr.X: 151955342-151955342 on Build GRCh38
rs749496766 Chr.X: 151955570-151955570 on Build GRCh38
rs779337081 Chr.X: 151955731-151955731 on Build GRCh38
rs924907083 Chr.X: 151955539-151955539 on Build GRCh38
rs951985435 Chr.X: 151955679-151955679 on Build GRCh38
rs483352728 Chr.X: 151955530-151955530 on Build GRCh38
rs61740516 Chr.X: 151955391-151955391 on Build GRCh38
rs752606074 Chr.X: 151955438-151955438 on Build GRCh38
rs766386512 Chr.X: 151955719-151955719 on Build GRCh38
rs753515807 Chr.X: 151955405-151955405 on Build GRCh38
rs765891212 Chr.X: 151955479-151955479 on Build GRCh38
rs141917924 Chr.X: 151955509-151955509 on Build GRCh38
rs77727101 Chr.X: 151955761-151955761 on Build GRCh38
rs756914368 Chr.X: 151955675-151955675 on Build GRCh38
rs181697394 Chr.X: 151955612-151955612 on Build GRCh38
rs774731784 Chr.X: 151955460-151955460 on Build GRCh38
rs748590514 Chr.X: 151955602-151955602 on Build GRCh38
rs201300022 Chr.X: 151955586-151955586 on Build GRCh38
rs760319267 Chr.X: 151955477-151955477 on Build GRCh38
rs773851811 Chr.X: 151955473-151955473 on Build GRCh38
rs773206156 Chr.X: 151955596-151955604 on Build GRCh38
rs888035563 Chr.X: 151955424-151955424 on Build GRCh38
rs778735662 Chr.X: 151955431-151955431 on Build GRCh38
rs749967424 Chr.X: 151955507-151955507 on Build GRCh38
rs749896675 Chr.X: 151955400-151955400 on Build GRCh38
rs374571425 Chr.X: 151955597-151955597 on Build GRCh38
rs779576822 Chr.X: 151955563-151955563 on Build GRCh38
rs74871311 Chr.X: 151955510-151955510 on Build GRCh38
rs781260349 Chr.X: 151955441-151955441 on Build GRCh38
rs55849806 Chr.X: 151955751-151955751 on Build GRCh38
rs936314042 Chr.X: 151955551-151955551 on Build GRCh38
rs78212023 Chr.X: 151955566-151955566 on Build GRCh38
rs780864713 Chr.X: 151955461-151955461 on Build GRCh38
rs758280870 Chr.X: 151955439-151955439 on Build GRCh38
rs776156895 Chr.X: 151955660-151955660 on Build GRCh38
rs369402853 Chr.X: 151955403-151955403 on Build GRCh38
rs760409209 Chr.X: 151955402-151955402 on Build GRCh38
rs774762959 Chr.X: 151955381-151955381 on Build GRCh38
rs892334242 Chr.X: 151955323-151955323 on Build GRCh38
rs778197680 Chr.X: 151955616-151955616 on Build GRCh38
rs759119898 Chr.X: 151955662-151955662 on Build GRCh38
rs768865712 Chr.X: 151955579-151955579 on Build GRCh38
rs372706803 Chr.X: 151955446-151955446 on Build GRCh38
rs187011945 Chr.X: 151955686-151955686 on Build GRCh38
rs769047265 Chr.X: 151955373-151955373 on Build GRCh38
rs112646382 Chr.X: 151955538-151955538 on Build GRCh38
rs776648638 Chr.X: 151955324-151955324 on Build GRCh38
rs753889158 Chr.X: 151955726-151955726 on Build GRCh38
rs763810305 Chr.X: 151955394-151955394 on Build GRCh38
rs751530972 Chr.X: 151955689-151955689 on Build GRCh38
rs768935553 Chr.X: 151955456-151955456 on Build GRCh38
rs371442646 Chr.X: 151955619-151955619 on Build GRCh38
rs867877489 Chr.X: 151955523-151955523 on Build GRCh38
rs745335766 Chr.X: 151955702-151955702 on Build GRCh38
rs746108157 Chr.X: 151955330-151955330 on Build GRCh38
rs748359641 Chr.X: 151955733-151955733 on Build GRCh38
rs753074937 Chr.X: 151955558-151955558 on Build GRCh38
rs751257441 Chr.X: 151955426-151955426 on Build GRCh38
rs775117386 Chr.X: 151955674-151955674 on Build GRCh38
rs896303707 Chr.X: 151955661-151955661 on Build GRCh38
rs376970481 Chr.X: 151955325-151955325 on Build GRCh38
rs772552823 Chr.X: 151955609-151955609 on Build GRCh38

View Full Product Details