Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153495289-153495789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BGN
Entrez Gene ID
633
Gene Name
biglycan
Gene Aliases
DSPG1, PG-S1, PGI, SLRR1A
UniGene
Hs.821
Gene Chromosome Location
Chr.X: 153494889-153509554 on Build GRCh38
Transcripts
NM_001711.5 NR_073156.1

Gene Symbol
HAUS7
Entrez Gene ID
55559
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
UCHL5IP, UIP1
UniGene
Hs.170835
Gene Chromosome Location
Chr.X: 153447665-153495525 on Build GRCh38
Transcripts
NM_001711.5 NR_073156.1

SNP Detail

SNP ID Chromosome Location
rs933190032 Chr.X: 153495321-153495321 on Build GRCh38
rs974511660 Chr.X: 153495631-153495631 on Build GRCh38
rs1042282649 Chr.X: 153495323-153495323 on Build GRCh38
rs970876769 Chr.X: 153495487-153495487 on Build GRCh38
rs782501744 Chr.X: 153495619-153495619 on Build GRCh38
rs1025112579 Chr.X: 153495480-153495480 on Build GRCh38
rs782201878 Chr.X: 153495737-153495737 on Build GRCh38
rs781784171 Chr.X: 153495417-153495417 on Build GRCh38
rs984842254 Chr.X: 153495581-153495581 on Build GRCh38
rs782754199 Chr.X: 153495572-153495572 on Build GRCh38
rs782536039 Chr.X: 153495646-153495646 on Build GRCh38
rs192607151 Chr.X: 153495535-153495535 on Build GRCh38
rs1053805162 Chr.X: 153495421-153495421 on Build GRCh38
rs782757791 Chr.X: 153495734-153495734 on Build GRCh38
rs530054889 Chr.X: 153495583-153495583 on Build GRCh38
rs55896261 Chr.X: 153495686-153495686 on Build GRCh38
rs933137485 Chr.X: 153495723-153495723 on Build GRCh38
rs782542051 Chr.X: 153495473-153495473 on Build GRCh38
rs782605608 Chr.X: 153495638-153495638 on Build GRCh38
rs936619536 Chr.X: 153495742-153495742 on Build GRCh38
rs143934714 Chr.X: 153495416-153495416 on Build GRCh38
rs964784852 Chr.X: 153495587-153495587 on Build GRCh38
rs2980045 Chr.X: 153495413-153495413 on Build GRCh38
rs994374413 Chr.X: 153495515-153495515 on Build GRCh38
rs895196653 Chr.X: 153495423-153495423 on Build GRCh38
rs781881934 Chr.X: 153495513-153495513 on Build GRCh38

View Full Product Details