Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153737050-153737538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD1
Entrez Gene ID
215
Gene Name
ATP binding cassette subfamily D member 1
Gene Aliases
ABC42, ALD, ALDP, AMN
UniGene
Hs.159546
Gene Chromosome Location
Chr.X: 153724851-153744762 on Build GRCh38
Transcripts
NM_000033.3

SNP Detail

SNP ID Chromosome Location
rs1003591837 Chr.X: 153737387-153737387 on Build GRCh38
rs782459996 Chr.X: 153737470-153737470 on Build GRCh38
rs1023611284 Chr.X: 153737355-153737355 on Build GRCh38
rs1035031015 Chr.X: 153737401-153737401 on Build GRCh38
rs782035584 Chr.X: 153737176-153737176 on Build GRCh38
rs782189180 Chr.X: 153737250-153737250 on Build GRCh38
rs782336998 Chr.X: 153737152-153737152 on Build GRCh38
rs192525493 Chr.X: 153737162-153737162 on Build GRCh38
rs782242003 Chr.X: 153737219-153737219 on Build GRCh38
rs782546330 Chr.X: 153737215-153737215 on Build GRCh38
rs1017091356 Chr.X: 153737474-153737474 on Build GRCh38
rs782516659 Chr.X: 153737230-153737230 on Build GRCh38
rs188345651 Chr.X: 153737274-153737274 on Build GRCh38
rs74315277 Chr.X: 153737155-153737155 on Build GRCh38
rs387906494 Chr.X: 153737178-153737179 on Build GRCh38
rs181175045 Chr.X: 153737477-153737477 on Build GRCh38
rs199690988 Chr.X: 153737115-153737115 on Build GRCh38
rs963313453 Chr.X: 153737484-153737484 on Build GRCh38
rs781962534 Chr.X: 153737111-153737111 on Build GRCh38
rs1052272241 Chr.X: 153737312-153737312 on Build GRCh38
rs782072629 Chr.X: 153737167-153737167 on Build GRCh38
rs782003554 Chr.X: 153737272-153737272 on Build GRCh38
rs951495369 Chr.X: 153737450-153737450 on Build GRCh38
rs128624214 Chr.X: 153737214-153737214 on Build GRCh38
rs782243754 Chr.X: 153737118-153737118 on Build GRCh38
rs142236904 Chr.X: 153737385-153737385 on Build GRCh38
rs983391406 Chr.X: 153737473-153737473 on Build GRCh38
rs781918062 Chr.X: 153737281-153737281 on Build GRCh38
rs782257884 Chr.X: 153737139-153737139 on Build GRCh38
rs782271412 Chr.X: 153737129-153737129 on Build GRCh38
rs128624222 Chr.X: 153737192-153737192 on Build GRCh38
rs782024993 Chr.X: 153737267-153737267 on Build GRCh38
rs781806403 Chr.X: 153737207-153737207 on Build GRCh38
rs975637209 Chr.X: 153737510-153737510 on Build GRCh38
rs782223032 Chr.X: 153737107-153737107 on Build GRCh38
rs782794762 Chr.X: 153737169-153737169 on Build GRCh38
rs183629718 Chr.X: 153737271-153737271 on Build GRCh38
rs782326465 Chr.X: 153737262-153737262 on Build GRCh38
rs782298500 Chr.X: 153737270-153737270 on Build GRCh38
rs1010807349 Chr.X: 153737352-153737352 on Build GRCh38
rs781981881 Chr.X: 153737138-153737138 on Build GRCh38
rs373127758 Chr.X: 153737280-153737280 on Build GRCh38
rs893747438 Chr.X: 153737313-153737313 on Build GRCh38
rs141446687 Chr.X: 153737191-153737191 on Build GRCh38
rs782596655 Chr.X: 153737234-153737234 on Build GRCh38
rs782704819 Chr.X: 153737206-153737206 on Build GRCh38
rs539250018 Chr.X: 153737083-153737083 on Build GRCh38
rs782429090 Chr.X: 153737110-153737110 on Build GRCh38
rs6643781 Chr.X: 153737085-153737085 on Build GRCh38
rs782723231 Chr.X: 153737080-153737080 on Build GRCh38
rs782415984 Chr.X: 153737127-153737127 on Build GRCh38
rs398123100 Chr.X: 153737159-153737159 on Build GRCh38
rs782118961 Chr.X: 153737189-153737189 on Build GRCh38
rs150878397 Chr.X: 153737218-153737218 on Build GRCh38

View Full Product Details