Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153740000-153740500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD1
Entrez Gene ID
215
Gene Name
ATP binding cassette subfamily D member 1
Gene Aliases
ABC42, ALD, ALDP, AMN
UniGene
Hs.159546
Gene Chromosome Location
Chr.X: 153724851-153744762 on Build GRCh38
Transcripts
NM_000033.3

SNP Detail

SNP ID Chromosome Location
rs993433741 Chr.X: 153740435-153740435 on Build GRCh38
rs782626071 Chr.X: 153740242-153740242 on Build GRCh38
rs184954481 Chr.X: 153740263-153740263 on Build GRCh38
rs782323354 Chr.X: 153740083-153740083 on Build GRCh38
rs128624224 Chr.X: 153740155-153740155 on Build GRCh38
rs4148036 Chr.X: 153740449-153740449 on Build GRCh38
rs387906495 Chr.X: 153740155-153740155 on Build GRCh38
rs372559385 Chr.X: 153740069-153740069 on Build GRCh38
rs56174192 Chr.X: 153740360-153740360 on Build GRCh38
rs782790134 Chr.X: 153740454-153740454 on Build GRCh38
rs782419093 Chr.X: 153740241-153740241 on Build GRCh38
rs782178203 Chr.X: 153740080-153740080 on Build GRCh38
rs782622708 Chr.X: 153740079-153740079 on Build GRCh38
rs398123101 Chr.X: 153740086-153740086 on Build GRCh38
rs368572593 Chr.X: 153740282-153740282 on Build GRCh38
rs149643908 Chr.X: 153740145-153740145 on Build GRCh38
rs927479436 Chr.X: 153740220-153740220 on Build GRCh38
rs782362712 Chr.X: 153740291-153740291 on Build GRCh38
rs128624223 Chr.X: 153740147-153740147 on Build GRCh38
rs782581290 Chr.X: 153740223-153740223 on Build GRCh38
rs781965097 Chr.X: 153740251-153740251 on Build GRCh38
rs7059198 Chr.X: 153740295-153740295 on Build GRCh38
rs904503753 Chr.X: 153740375-153740375 on Build GRCh38
rs398123103 Chr.X: 153740189-153740189 on Build GRCh38
rs1034357061 Chr.X: 153740395-153740395 on Build GRCh38
rs782440686 Chr.X: 153740218-153740218 on Build GRCh38
rs139023922 Chr.X: 153740283-153740283 on Build GRCh38
rs911005858 Chr.X: 153740466-153740466 on Build GRCh38
rs782370364 Chr.X: 153740136-153740136 on Build GRCh38
rs782041728 Chr.X: 153740253-153740253 on Build GRCh38
rs369538219 Chr.X: 153740249-153740249 on Build GRCh38
rs145245018 Chr.X: 153740328-153740328 on Build GRCh38
rs140996847 Chr.X: 153740154-153740154 on Build GRCh38
rs781838947 Chr.X: 153740216-153740216 on Build GRCh38
rs782158792 Chr.X: 153740129-153740129 on Build GRCh38
rs375632296 Chr.X: 153740075-153740075 on Build GRCh38
rs41314153 Chr.X: 153740151-153740151 on Build GRCh38
rs370136009 Chr.X: 153740078-153740078 on Build GRCh38
rs781790745 Chr.X: 153740453-153740453 on Build GRCh38
rs145073701 Chr.X: 153740184-153740184 on Build GRCh38
rs188120924 Chr.X: 153740475-153740475 on Build GRCh38
rs781969484 Chr.X: 153740405-153740405 on Build GRCh38
rs142496200 Chr.X: 153740390-153740390 on Build GRCh38
rs972314833 Chr.X: 153740450-153740450 on Build GRCh38
rs782514164 Chr.X: 153740214-153740214 on Build GRCh38
rs149185761 Chr.X: 153740068-153740068 on Build GRCh38
rs376472029 Chr.X: 153740185-153740185 on Build GRCh38
rs782488568 Chr.X: 153740047-153740047 on Build GRCh38
rs782726532 Chr.X: 153740436-153740436 on Build GRCh38
rs782611445 Chr.X: 153740215-153740215 on Build GRCh38
rs193922094 Chr.X: 153740195-153740195 on Build GRCh38
rs398123102 Chr.X: 153740156-153740156 on Build GRCh38
rs782399305 Chr.X: 153740250-153740250 on Build GRCh38
rs375499311 Chr.X: 153740261-153740261 on Build GRCh38
rs782694108 Chr.X: 153740286-153740286 on Build GRCh38
rs375422416 Chr.X: 153740058-153740058 on Build GRCh38
rs373442692 Chr.X: 153740124-153740124 on Build GRCh38
rs781789877 Chr.X: 153740438-153740438 on Build GRCh38
rs781863403 Chr.X: 153740270-153740270 on Build GRCh38
rs782499874 Chr.X: 153740476-153740476 on Build GRCh38
rs781862879 Chr.X: 153740200-153740200 on Build GRCh38
rs139343560 Chr.X: 153740139-153740139 on Build GRCh38
rs782152974 Chr.X: 153740376-153740376 on Build GRCh38
rs797044766 Chr.X: 153740119-153740119 on Build GRCh38
rs983782630 Chr.X: 153740465-153740465 on Build GRCh38
rs183184507 Chr.X: 153740408-153740408 on Build GRCh38
rs782412002 Chr.X: 153740254-153740254 on Build GRCh38
rs190868914 Chr.X: 153740298-153740298 on Build GRCh38
rs782278443 Chr.X: 153740232-153740232 on Build GRCh38
rs201753687 Chr.X: 153740059-153740059 on Build GRCh38
rs782484271 Chr.X: 153740061-153740061 on Build GRCh38
rs781903537 Chr.X: 153740045-153740045 on Build GRCh38
rs930928628 Chr.X: 153740456-153740456 on Build GRCh38
rs896926825 Chr.X: 153740051-153740051 on Build GRCh38

View Full Product Details