Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229322418-229322960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSAP
Entrez Gene ID
126731
Gene Name
centriole, cilia and spindle associated protein
Gene Aliases
C1orf96, CSAP
UniGene
Hs.585011
Gene Chromosome Location
Chr.1: 229321005-229342941 on Build GRCh38
Transcripts
NM_145257.4

SNP Detail

SNP ID Chromosome Location
rs933908939 Chr.1: 229322453-229322453 on Build GRCh38
rs983288488 Chr.1: 229322690-229322690 on Build GRCh38
rs963869076 Chr.1: 229322756-229322756 on Build GRCh38
rs1051004801 Chr.1: 229322739-229322739 on Build GRCh38
rs547224598 Chr.1: 229322686-229322686 on Build GRCh38
rs893992658 Chr.1: 229322556-229322556 on Build GRCh38
rs771263760 Chr.1: 229322596-229322597 on Build GRCh38
rs893626349 Chr.1: 229322617-229322617 on Build GRCh38
rs187858745 Chr.1: 229322711-229322711 on Build GRCh38
rs547593670 Chr.1: 229322808-229322810 on Build GRCh38
rs1051239540 Chr.1: 229322486-229322486 on Build GRCh38
rs1040022075 Chr.1: 229322675-229322675 on Build GRCh38
rs951392606 Chr.1: 229322685-229322685 on Build GRCh38
rs1088 Chr.1: 229322709-229322709 on Build GRCh38
rs539038487 Chr.1: 229322827-229322827 on Build GRCh38
rs771932422 Chr.1: 229322725-229322725 on Build GRCh38
rs934316745 Chr.1: 229322750-229322750 on Build GRCh38
rs949574824 Chr.1: 229322858-229322858 on Build GRCh38
rs1035458004 Chr.1: 229322620-229322620 on Build GRCh38
rs1046405950 Chr.1: 229322492-229322492 on Build GRCh38
rs183926279 Chr.1: 229322598-229322598 on Build GRCh38
rs531348755 Chr.1: 229322480-229322480 on Build GRCh38
rs1054188607 Chr.1: 229322552-229322552 on Build GRCh38
rs746172562 Chr.1: 229322737-229322737 on Build GRCh38
rs114787809 Chr.1: 229322836-229322836 on Build GRCh38
rs1054261598 Chr.1: 229322930-229322930 on Build GRCh38
rs747289702 Chr.1: 229322576-229322576 on Build GRCh38
rs569910459 Chr.1: 229322789-229322789 on Build GRCh38
rs556240446 Chr.1: 229322728-229322728 on Build GRCh38
rs990956662 Chr.1: 229322758-229322758 on Build GRCh38
rs746009788 Chr.1: 229322611-229322611 on Build GRCh38
rs564097602 Chr.1: 229322543-229322543 on Build GRCh38
rs902838079 Chr.1: 229322531-229322531 on Build GRCh38
rs924290218 Chr.1: 229322745-229322745 on Build GRCh38
rs1006747425 Chr.1: 229322650-229322650 on Build GRCh38
rs949627062 Chr.1: 229322456-229322456 on Build GRCh38
rs902512911 Chr.1: 229322529-229322529 on Build GRCh38
rs935384836 Chr.1: 229322886-229322886 on Build GRCh38
rs965243505 Chr.1: 229322705-229322705 on Build GRCh38
rs1043188 Chr.1: 229322482-229322482 on Build GRCh38
rs969667529 Chr.1: 229322599-229322599 on Build GRCh38
rs879538186 Chr.1: 229322818-229322822 on Build GRCh38
rs951115970 Chr.1: 229322716-229322716 on Build GRCh38
rs237799 Chr.1: 229322885-229322885 on Build GRCh38
rs975258326 Chr.1: 229322712-229322712 on Build GRCh38
rs774475303 Chr.1: 229322710-229322710 on Build GRCh38
rs572723288 Chr.1: 229322880-229322880 on Build GRCh38

View Full Product Details