Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229325020-229325546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSAP
Entrez Gene ID
126731
Gene Name
centriole, cilia and spindle associated protein
Gene Aliases
C1orf96, CSAP
UniGene
Hs.585011
Gene Chromosome Location
Chr.1: 229321005-229342941 on Build GRCh38
Transcripts
NM_145257.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5003843 c.333G>T p.Q111H Chr.1: 229325373-229325373 on Build GRCh38
299023 c.648A>C p.S216S Chr.1: 229325400-229325400 on Build GRCh38
297417 c.801G>A p.S267S Chr.1: 229325247-229325247 on Build GRCh38
7270663 c.794G>A p.C265Y Chr.1: 229325254-229325254 on Build GRCh38
5845792 c.337G>A p.E113K Chr.1: 229325369-229325369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749896092 Chr.1: 229325206-229325206 on Build GRCh38
rs777436638 Chr.1: 229325428-229325428 on Build GRCh38
rs997858958 Chr.1: 229325194-229325205 on Build GRCh38
rs199755543 Chr.1: 229325222-229325222 on Build GRCh38
rs1004486800 Chr.1: 229325405-229325405 on Build GRCh38
rs550995888 Chr.1: 229325058-229325058 on Build GRCh38
rs770979799 Chr.1: 229325309-229325309 on Build GRCh38
rs774326765 Chr.1: 229325313-229325313 on Build GRCh38
rs200643344 Chr.1: 229325217-229325217 on Build GRCh38
rs963093784 Chr.1: 229325081-229325081 on Build GRCh38
rs749211088 Chr.1: 229325444-229325444 on Build GRCh38
rs749384189 Chr.1: 229325234-229325234 on Build GRCh38
rs749199176 Chr.1: 229325307-229325307 on Build GRCh38
rs142888796 Chr.1: 229325475-229325475 on Build GRCh38
rs981244763 Chr.1: 229325514-229325514 on Build GRCh38
rs558201743 Chr.1: 229325151-229325151 on Build GRCh38
rs781062232 Chr.1: 229325380-229325380 on Build GRCh38
rs927323560 Chr.1: 229325073-229325073 on Build GRCh38
rs988861790 Chr.1: 229325240-229325240 on Build GRCh38
rs898848942 Chr.1: 229325210-229325210 on Build GRCh38
rs769853432 Chr.1: 229325278-229325278 on Build GRCh38
rs978851473 Chr.1: 229325061-229325061 on Build GRCh38
rs917087014 Chr.1: 229325496-229325496 on Build GRCh38
rs565465454 Chr.1: 229325154-229325154 on Build GRCh38
rs559873142 Chr.1: 229325369-229325369 on Build GRCh38
rs917077355 Chr.1: 229325167-229325167 on Build GRCh38
rs368875795 Chr.1: 229325397-229325397 on Build GRCh38
rs755918335 Chr.1: 229325419-229325419 on Build GRCh38
rs767776558 Chr.1: 229325329-229325329 on Build GRCh38
rs1004650344 Chr.1: 229325047-229325047 on Build GRCh38
rs992909079 Chr.1: 229325478-229325478 on Build GRCh38
rs940505551 Chr.1: 229325500-229325500 on Build GRCh38
rs746943810 Chr.1: 229325424-229325424 on Build GRCh38
rs974156426 Chr.1: 229325086-229325086 on Build GRCh38
rs184794144 Chr.1: 229325109-229325109 on Build GRCh38
rs747946855 Chr.1: 229325382-229325382 on Build GRCh38
rs981338475 Chr.1: 229325178-229325178 on Build GRCh38
rs969867088 Chr.1: 229325511-229325511 on Build GRCh38
rs562017044 Chr.1: 229325503-229325503 on Build GRCh38
rs900006557 Chr.1: 229325302-229325302 on Build GRCh38
rs570983272 Chr.1: 229325098-229325098 on Build GRCh38
rs780293534 Chr.1: 229325223-229325223 on Build GRCh38
rs189175208 Chr.1: 229325220-229325220 on Build GRCh38
rs778319465 Chr.1: 229325199-229325199 on Build GRCh38
rs748692796 Chr.1: 229325425-229325425 on Build GRCh38
rs376862814 Chr.1: 229325457-229325457 on Build GRCh38
rs756599545 Chr.1: 229325197-229325197 on Build GRCh38
rs995588369 Chr.1: 229325485-229325485 on Build GRCh38
rs1051272147 Chr.1: 229325363-229325363 on Build GRCh38
rs757707620 Chr.1: 229325338-229325338 on Build GRCh38
rs1020229896 Chr.1: 229325243-229325243 on Build GRCh38
rs375812703 Chr.1: 229325297-229325297 on Build GRCh38
rs200618445 Chr.1: 229325455-229325455 on Build GRCh38
rs761093354 Chr.1: 229325335-229325335 on Build GRCh38
rs775800838 Chr.1: 229325334-229325334 on Build GRCh38
rs768451855 Chr.1: 229325227-229325227 on Build GRCh38
rs978797507 Chr.1: 229325448-229325448 on Build GRCh38
rs759756701 Chr.1: 229325320-229325320 on Build GRCh38
rs750701768 Chr.1: 229325117-229325117 on Build GRCh38
rs150524317 Chr.1: 229325336-229325336 on Build GRCh38
rs576943262 Chr.1: 229325339-229325339 on Build GRCh38
rs180702303 Chr.1: 229325228-229325228 on Build GRCh38
rs2282083 Chr.1: 229325277-229325277 on Build GRCh38
rs201879768 Chr.1: 229325350-229325350 on Build GRCh38
rs753072962 Chr.1: 229325184-229325184 on Build GRCh38

View Full Product Details