Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229342021-229342529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSAP
Entrez Gene ID
126731
Gene Name
centriole, cilia and spindle associated protein
Gene Aliases
C1orf96, CSAP
UniGene
Hs.585011
Gene Chromosome Location
Chr.1: 229321005-229342941 on Build GRCh38
Transcripts
NM_145257.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3804265 c.361C>A p.L121M Chr.1: 229342105-229342105 on Build GRCh38
5795280 c.216C>T p.P72P Chr.1: 229342250-229342250 on Build GRCh38
3771648 c.287G>A p.R96Q Chr.1: 229342179-229342179 on Build GRCh38
5701258 c.298_299insG p.A100fs*33 Chr.1: 229342167-229342168 on Build GRCh38
6391060 c.151G>A p.D51N Chr.1: 229342315-229342315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765999981 Chr.1: 229342502-229342511 on Build GRCh38
rs988224014 Chr.1: 229342087-229342087 on Build GRCh38
rs755505435 Chr.1: 229342484-229342484 on Build GRCh38
rs996774679 Chr.1: 229342176-229342176 on Build GRCh38
rs772653642 Chr.1: 229342257-229342257 on Build GRCh38
rs778686530 Chr.1: 229342497-229342497 on Build GRCh38
rs761753575 Chr.1: 229342107-229342108 on Build GRCh38
rs748894195 Chr.1: 229342391-229342391 on Build GRCh38
rs999027346 Chr.1: 229342133-229342133 on Build GRCh38
rs752024632 Chr.1: 229342471-229342471 on Build GRCh38
rs980643127 Chr.1: 229342466-229342466 on Build GRCh38
rs780291980 Chr.1: 229342469-229342469 on Build GRCh38
rs753993383 Chr.1: 229342431-229342431 on Build GRCh38
rs772904629 Chr.1: 229342298-229342298 on Build GRCh38
rs770486890 Chr.1: 229342488-229342488 on Build GRCh38
rs1003880723 Chr.1: 229342213-229342213 on Build GRCh38
rs368125854 Chr.1: 229342353-229342353 on Build GRCh38
rs749657408 Chr.1: 229342127-229342127 on Build GRCh38
rs1029544265 Chr.1: 229342278-229342278 on Build GRCh38
rs976224328 Chr.1: 229342501-229342501 on Build GRCh38
rs747732001 Chr.1: 229342286-229342286 on Build GRCh38
rs762102520 Chr.1: 229342440-229342440 on Build GRCh38
rs771892927 Chr.1: 229342394-229342394 on Build GRCh38
rs1029776524 Chr.1: 229342214-229342228 on Build GRCh38
rs754978160 Chr.1: 229342109-229342109 on Build GRCh38
rs551475820 Chr.1: 229342055-229342055 on Build GRCh38
rs1056150789 Chr.1: 229342163-229342163 on Build GRCh38
rs894869719 Chr.1: 229342101-229342101 on Build GRCh38
rs762656944 Chr.1: 229342447-229342449 on Build GRCh38
rs759373424 Chr.1: 229342311-229342311 on Build GRCh38
rs757639581 Chr.1: 229342128-229342128 on Build GRCh38
rs752716539 Chr.1: 229342315-229342315 on Build GRCh38
rs112195774 Chr.1: 229342415-229342415 on Build GRCh38
rs759664214 Chr.1: 229342066-229342066 on Build GRCh38
rs774227192 Chr.1: 229342306-229342306 on Build GRCh38
rs780674778 Chr.1: 229342365-229342365 on Build GRCh38
rs1004010852 Chr.1: 229342453-229342453 on Build GRCh38
rs772957610 Chr.1: 229342069-229342069 on Build GRCh38
rs112574153 Chr.1: 229342284-229342284 on Build GRCh38
rs371974930 Chr.1: 229342383-229342383 on Build GRCh38
rs757584591 Chr.1: 229342346-229342346 on Build GRCh38
rs750848038 Chr.1: 229342354-229342354 on Build GRCh38
rs750746454 Chr.1: 229342449-229342449 on Build GRCh38
rs751388996 Chr.1: 229342105-229342105 on Build GRCh38
rs764662601 Chr.1: 229342116-229342117 on Build GRCh38
rs112041170 Chr.1: 229342108-229342108 on Build GRCh38
rs775979513 Chr.1: 229342274-229342274 on Build GRCh38
rs747594363 Chr.1: 229342372-229342372 on Build GRCh38
rs745520527 Chr.1: 229342500-229342500 on Build GRCh38
rs891172042 Chr.1: 229342446-229342446 on Build GRCh38
rs764915886 Chr.1: 229342062-229342062 on Build GRCh38
rs1031904661 Chr.1: 229342174-229342174 on Build GRCh38
rs752874136 Chr.1: 229342112-229342112 on Build GRCh38
rs776503110 Chr.1: 229342093-229342093 on Build GRCh38
rs778208826 Chr.1: 229342121-229342121 on Build GRCh38
rs771834257 Chr.1: 229342509-229342509 on Build GRCh38
rs1019151695 Chr.1: 229342309-229342309 on Build GRCh38
rs766126122 Chr.1: 229342304-229342304 on Build GRCh38
rs754046436 Chr.1: 229342341-229342341 on Build GRCh38
rs571590141 Chr.1: 229342088-229342088 on Build GRCh38
rs374954717 Chr.1: 229342205-229342205 on Build GRCh38
rs758851743 Chr.1: 229342454-229342454 on Build GRCh38
rs756375007 Chr.1: 229342118-229342118 on Build GRCh38
rs779423761 Chr.1: 229342129-229342129 on Build GRCh38
rs367794347 Chr.1: 229342254-229342254 on Build GRCh38
rs886737304 Chr.1: 229342172-229342172 on Build GRCh38
rs201982587 Chr.1: 229342425-229342425 on Build GRCh38
rs760694000 Chr.1: 229342412-229342412 on Build GRCh38
rs775441272 Chr.1: 229342405-229342405 on Build GRCh38
rs887696787 Chr.1: 229342081-229342081 on Build GRCh38
rs765535172 Chr.1: 229342448-229342448 on Build GRCh38
rs777299196 Chr.1: 229342382-229342382 on Build GRCh38
rs769232689 Chr.1: 229342378-229342378 on Build GRCh38
rs746321005 Chr.1: 229342246-229342246 on Build GRCh38
rs375851502 Chr.1: 229342093-229342093 on Build GRCh38
rs760895958 Chr.1: 229342316-229342316 on Build GRCh38
rs1029721179 Chr.1: 229342220-229342220 on Build GRCh38
rs527713891 Chr.1: 229342110-229342110 on Build GRCh38
rs939222088 Chr.1: 229342142-229342142 on Build GRCh38
rs769428896 Chr.1: 229342293-229342293 on Build GRCh38
rs78093414 Chr.1: 229342059-229342059 on Build GRCh38
rs773989155 Chr.1: 229342392-229342392 on Build GRCh38
rs772778716 Chr.1: 229342437-229342438 on Build GRCh38
rs748839087 Chr.1: 229342487-229342487 on Build GRCh38
rs957621933 Chr.1: 229342208-229342208 on Build GRCh38
rs767532308 Chr.1: 229342312-229342312 on Build GRCh38
rs12736443 Chr.1: 229342363-229342363 on Build GRCh38

View Full Product Details