Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230066842-230067361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT2
Entrez Gene ID
2590
Gene Name
polypeptide N-acetylgalactosaminyltransferase 2
Gene Aliases
GalNAc-T2
UniGene
Hs.743964
Gene Chromosome Location
Chr.1: 230057789-230282130 on Build GRCh38
Transcripts
NM_001291866.1 NM_004481.4 NR_120373.1 XM_017000963.1 XM_017000965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143498 c.12C>A p.R4R Chr.1: 230067292-230067292 on Build GRCh38
1997798 c.75delG p.G27fs*115 Chr.1: 230067355-230067355 on Build GRCh38
397098 c.14C>T p.S5L Chr.1: 230067294-230067294 on Build GRCh38
7316746 c.57C>G p.I19M Chr.1: 230067337-230067337 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1019005409 Chr.1: 230067132-230067132 on Build GRCh38
rs773281082 Chr.1: 230067319-230067319 on Build GRCh38
rs953601031 Chr.1: 230066865-230066865 on Build GRCh38
rs944525425 Chr.1: 230067039-230067039 on Build GRCh38
rs1036201036 Chr.1: 230066978-230066978 on Build GRCh38
rs914142432 Chr.1: 230066936-230066936 on Build GRCh38
rs966160902 Chr.1: 230067036-230067036 on Build GRCh38
rs986247909 Chr.1: 230066902-230066902 on Build GRCh38
rs958327709 Chr.1: 230067191-230067191 on Build GRCh38
rs978845783 Chr.1: 230066938-230066938 on Build GRCh38
rs949855871 Chr.1: 230067097-230067097 on Build GRCh38
rs866225707 Chr.1: 230067038-230067038 on Build GRCh38
rs541927864 Chr.1: 230066933-230066933 on Build GRCh38
rs769786670 Chr.1: 230067315-230067315 on Build GRCh38
rs865947899 Chr.1: 230067314-230067314 on Build GRCh38
rs991240455 Chr.1: 230067087-230067087 on Build GRCh38
rs546906676 Chr.1: 230067265-230067265 on Build GRCh38
rs889189072 Chr.1: 230066999-230066999 on Build GRCh38
rs564710739 Chr.1: 230067216-230067216 on Build GRCh38
rs988429978 Chr.1: 230066908-230066908 on Build GRCh38
rs949970007 Chr.1: 230066970-230066970 on Build GRCh38
rs763087504 Chr.1: 230067320-230067320 on Build GRCh38
rs937086358 Chr.1: 230067073-230067073 on Build GRCh38
rs968313405 Chr.1: 230067168-230067168 on Build GRCh38
rs1028088778 Chr.1: 230066996-230066996 on Build GRCh38
rs971905739 Chr.1: 230067219-230067219 on Build GRCh38
rs1036618992 Chr.1: 230067104-230067104 on Build GRCh38
rs1055538883 Chr.1: 230066957-230066957 on Build GRCh38
rs953355077 Chr.1: 230066864-230066864 on Build GRCh38
rs937114278 Chr.1: 230066945-230066945 on Build GRCh38
rs544518954 Chr.1: 230067201-230067201 on Build GRCh38
rs911945235 Chr.1: 230066912-230066912 on Build GRCh38
rs910479087 Chr.1: 230067237-230067237 on Build GRCh38
rs966274575 Chr.1: 230066878-230066878 on Build GRCh38
rs1009712959 Chr.1: 230067159-230067159 on Build GRCh38
rs572889253 Chr.1: 230066877-230066877 on Build GRCh38
rs745520668 Chr.1: 230067258-230067258 on Build GRCh38
rs778442919 Chr.1: 230067241-230067241 on Build GRCh38
rs566513446 Chr.1: 230067270-230067270 on Build GRCh38
rs1032678013 Chr.1: 230067183-230067183 on Build GRCh38
rs1007680811 Chr.1: 230067026-230067026 on Build GRCh38
rs1049047037 Chr.1: 230067122-230067122 on Build GRCh38
rs962051508 Chr.1: 230067089-230067126 on Build GRCh38
rs925637394 Chr.1: 230067067-230067067 on Build GRCh38
rs1021561591 Chr.1: 230067160-230067160 on Build GRCh38
rs925700295 Chr.1: 230066942-230066942 on Build GRCh38
rs986275852 Chr.1: 230066872-230066872 on Build GRCh38
rs897718604 Chr.1: 230067105-230067105 on Build GRCh38
rs991125873 Chr.1: 230067200-230067200 on Build GRCh38
rs945698018 Chr.1: 230066937-230066937 on Build GRCh38
rs746866392 Chr.1: 230067271-230067271 on Build GRCh38
rs768416580 Chr.1: 230067282-230067282 on Build GRCh38
rs533432248 Chr.1: 230067233-230067233 on Build GRCh38
rs779679053 Chr.1: 230067269-230067269 on Build GRCh38
rs897745108 Chr.1: 230066987-230066987 on Build GRCh38
rs1019120416 Chr.1: 230067035-230067035 on Build GRCh38
rs776589215 Chr.1: 230067284-230067284 on Build GRCh38
rs771249582 Chr.1: 230067337-230067337 on Build GRCh38
rs552974853 Chr.1: 230066868-230066868 on Build GRCh38
rs1019148655 Chr.1: 230066877-230066878 on Build GRCh38
rs1010093746 Chr.1: 230067289-230067289 on Build GRCh38
rs1040413981 Chr.1: 230067261-230067261 on Build GRCh38
rs181092443 Chr.1: 230066969-230066969 on Build GRCh38
rs1007567661 Chr.1: 230067129-230067129 on Build GRCh38
rs971139760 Chr.1: 230067211-230067211 on Build GRCh38
rs943322825 Chr.1: 230067239-230067239 on Build GRCh38
rs930579910 Chr.1: 230067117-230067117 on Build GRCh38
rs183724411 Chr.1: 230067172-230067172 on Build GRCh38
rs889159088 Chr.1: 230067125-230067125 on Build GRCh38
rs919114396 Chr.1: 230067220-230067220 on Build GRCh38
rs901941819 Chr.1: 230067152-230067152 on Build GRCh38
rs866780552 Chr.1: 230067338-230067338 on Build GRCh38
rs978430436 Chr.1: 230067047-230067047 on Build GRCh38
rs1049409931 Chr.1: 230067234-230067234 on Build GRCh38
rs994820491 Chr.1: 230066991-230066991 on Build GRCh38
rs370099426 Chr.1: 230067292-230067292 on Build GRCh38
rs968721856 Chr.1: 230067043-230067043 on Build GRCh38

View Full Product Details