Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230767436-230767843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN9
Entrez Gene ID
10753
Gene Name
calpain 9
Gene Aliases
GC36, nCL-4
UniGene
Hs.498021
Gene Chromosome Location
Chr.1: 230747289-230803356 on Build GRCh38
Transcripts
NM_001319676.1 NM_006615.2 NM_016452.2 XM_011544017.1 XM_011544018.1 XM_011544019.2 XM_011544020.1 XM_017000098.1 XM_017000099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5380041 c.699C>T p.F233F Chr.1: 230767703-230767703 on Build GRCh38
3710639 c.565G>T p.G189* Chr.1: 230767569-230767569 on Build GRCh38
1626888 c.657T>G p.I219M Chr.1: 230767661-230767661 on Build GRCh38
1146224 c.561G>A p.L187L Chr.1: 230767565-230767565 on Build GRCh38
3485018 c.588G>A p.M196I Chr.1: 230767592-230767592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766970963 Chr.1: 230767613-230767613 on Build GRCh38
rs765102262 Chr.1: 230767586-230767586 on Build GRCh38
rs946470316 Chr.1: 230767480-230767480 on Build GRCh38
rs1012217126 Chr.1: 230767589-230767589 on Build GRCh38
rs60100843 Chr.1: 230767667-230767667 on Build GRCh38
rs757044891 Chr.1: 230767654-230767654 on Build GRCh38
rs763721514 Chr.1: 230767643-230767643 on Build GRCh38
rs372361365 Chr.1: 230767583-230767583 on Build GRCh38
rs370962935 Chr.1: 230767515-230767516 on Build GRCh38
rs370440769 Chr.1: 230767762-230767762 on Build GRCh38
rs11579577 Chr.1: 230767487-230767487 on Build GRCh38
rs200639798 Chr.1: 230767510-230767511 on Build GRCh38
rs865913369 Chr.1: 230767516-230767516 on Build GRCh38
rs148953435 Chr.1: 230767596-230767596 on Build GRCh38
rs771580006 Chr.1: 230767731-230767731 on Build GRCh38
rs751244678 Chr.1: 230767593-230767593 on Build GRCh38
rs1012966495 Chr.1: 230767530-230767530 on Build GRCh38
rs879133780 Chr.1: 230767795-230767795 on Build GRCh38
rs936214239 Chr.1: 230767787-230767787 on Build GRCh38
rs777474196 Chr.1: 230767523-230767523 on Build GRCh38
rs558264332 Chr.1: 230767749-230767750 on Build GRCh38
rs568824112 Chr.1: 230767462-230767462 on Build GRCh38
rs149766737 Chr.1: 230767577-230767577 on Build GRCh38
rs560253000 Chr.1: 230767793-230767793 on Build GRCh38
rs909510246 Chr.1: 230767665-230767665 on Build GRCh38
rs760259417 Chr.1: 230767640-230767640 on Build GRCh38
rs911803618 Chr.1: 230767710-230767710 on Build GRCh38
rs1039862957 Chr.1: 230767473-230767473 on Build GRCh38
rs369955125 Chr.1: 230767720-230767720 on Build GRCh38
rs556429467 Chr.1: 230767587-230767587 on Build GRCh38
rs773742359 Chr.1: 230767608-230767608 on Build GRCh38
rs1025761017 Chr.1: 230767672-230767672 on Build GRCh38
rs527480684 Chr.1: 230767466-230767466 on Build GRCh38
rs962765916 Chr.1: 230767609-230767609 on Build GRCh38
rs780217323 Chr.1: 230767669-230767669 on Build GRCh38
rs553903946 Chr.1: 230767550-230767550 on Build GRCh38
rs755310866 Chr.1: 230767693-230767693 on Build GRCh38
rs781562865 Chr.1: 230767695-230767695 on Build GRCh38
rs372367329 Chr.1: 230767722-230767722 on Build GRCh38
rs536743386 Chr.1: 230767578-230767578 on Build GRCh38
rs11122590 Chr.1: 230767488-230767488 on Build GRCh38
rs776322706 Chr.1: 230767757-230767757 on Build GRCh38
rs747249590 Chr.1: 230767683-230767683 on Build GRCh38
rs914926123 Chr.1: 230767751-230767751 on Build GRCh38
rs368899138 Chr.1: 230767760-230767760 on Build GRCh38
rs531252850 Chr.1: 230767484-230767484 on Build GRCh38
rs3828127 Chr.1: 230767749-230767749 on Build GRCh38
rs559006925 Chr.1: 230767737-230767737 on Build GRCh38
rs924787416 Chr.1: 230767782-230767782 on Build GRCh38
rs1003354168 Chr.1: 230767599-230767599 on Build GRCh38
rs763777063 Chr.1: 230767748-230767748 on Build GRCh38
rs771569749 Chr.1: 230767510-230767516 on Build GRCh38
rs879641566 Chr.1: 230767487-230767488 on Build GRCh38
rs760171267 Chr.1: 230767738-230767738 on Build GRCh38
rs3828126 Chr.1: 230767604-230767604 on Build GRCh38
rs115285515 Chr.1: 230767525-230767525 on Build GRCh38
rs386640213 Chr.1: 230767487-230767488 on Build GRCh38
rs199946717 Chr.1: 230767552-230767552 on Build GRCh38
rs752347458 Chr.1: 230767639-230767639 on Build GRCh38
rs760299883 Chr.1: 230767730-230767761 on Build GRCh38
rs754842123 Chr.1: 230767499-230767499 on Build GRCh38
rs28359644 Chr.1: 230767705-230767705 on Build GRCh38
rs775043847 Chr.1: 230767576-230767576 on Build GRCh38
rs758439855 Chr.1: 230767663-230767663 on Build GRCh38
rs950560110 Chr.1: 230767687-230767687 on Build GRCh38
rs375577407 Chr.1: 230767644-230767644 on Build GRCh38
rs147002278 Chr.1: 230767662-230767662 on Build GRCh38
rs893522422 Chr.1: 230767471-230767471 on Build GRCh38
rs759021638 Chr.1: 230767611-230767611 on Build GRCh38
rs946439210 Chr.1: 230767775-230767775 on Build GRCh38
rs984018565 Chr.1: 230767656-230767656 on Build GRCh38
rs1042103194 Chr.1: 230767820-230767820 on Build GRCh38
rs755931030 Chr.1: 230767515-230767515 on Build GRCh38
rs748504285 Chr.1: 230767703-230767703 on Build GRCh38

View Full Product Details