Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230778818-230779315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN9
Entrez Gene ID
10753
Gene Name
calpain 9
Gene Aliases
GC36, nCL-4
UniGene
Hs.498021
Gene Chromosome Location
Chr.1: 230747289-230803356 on Build GRCh38
Transcripts
NM_001319676.1 NM_006615.2 NM_016452.2 XM_011544017.1 XM_011544018.1 XM_011544019.2 XM_011544020.1 XM_017000098.1 XM_017000099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1146227 c.1066G>T p.G356* Chr.1: 230779085-230779085 on Build GRCh38
1601877 c.1049A>T p.E350V Chr.1: 230779068-230779068 on Build GRCh38
1601876 c.961C>T p.H321Y Chr.1: 230779058-230779058 on Build GRCh38
1146225 c.1065G>T p.Q355H Chr.1: 230779084-230779084 on Build GRCh38
1601875 c.1039C>T p.H347Y Chr.1: 230779058-230779058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762381875 Chr.1: 230778962-230778962 on Build GRCh38
rs776047394 Chr.1: 230778953-230778953 on Build GRCh38
rs769714574 Chr.1: 230778908-230778908 on Build GRCh38
rs763956045 Chr.1: 230778995-230778995 on Build GRCh38
rs143810036 Chr.1: 230779023-230779023 on Build GRCh38
rs895640377 Chr.1: 230779081-230779081 on Build GRCh38
rs1933630 Chr.1: 230778960-230778960 on Build GRCh38
rs200401992 Chr.1: 230779051-230779051 on Build GRCh38
rs373768301 Chr.1: 230778950-230778950 on Build GRCh38
rs770366418 Chr.1: 230779034-230779034 on Build GRCh38
rs745399830 Chr.1: 230779109-230779109 on Build GRCh38
rs1042464540 Chr.1: 230778902-230778902 on Build GRCh38
rs575112353 Chr.1: 230779100-230779100 on Build GRCh38
rs183093849 Chr.1: 230779237-230779237 on Build GRCh38
rs747037534 Chr.1: 230779189-230779189 on Build GRCh38
rs569927031 Chr.1: 230778847-230778847 on Build GRCh38
rs530416336 Chr.1: 230779234-230779235 on Build GRCh38
rs199593400 Chr.1: 230779075-230779075 on Build GRCh38
rs368728974 Chr.1: 230778944-230778944 on Build GRCh38
rs191352967 Chr.1: 230778927-230778927 on Build GRCh38
rs533848130 Chr.1: 230778878-230778878 on Build GRCh38
rs765846394 Chr.1: 230778968-230778968 on Build GRCh38
rs780436701 Chr.1: 230779097-230779097 on Build GRCh38
rs148341318 Chr.1: 230779107-230779107 on Build GRCh38
rs771508321 Chr.1: 230779116-230779116 on Build GRCh38
rs145648359 Chr.1: 230779052-230779052 on Build GRCh38
rs377045480 Chr.1: 230779292-230779292 on Build GRCh38
rs145226005 Chr.1: 230779074-230779074 on Build GRCh38
rs150463113 Chr.1: 230779121-230779121 on Build GRCh38
rs759012112 Chr.1: 230779037-230779037 on Build GRCh38
rs763129836 Chr.1: 230779156-230779156 on Build GRCh38
rs377721311 Chr.1: 230778951-230778951 on Build GRCh38
rs943621576 Chr.1: 230778844-230778844 on Build GRCh38
rs779010670 Chr.1: 230778948-230778948 on Build GRCh38
rs752525122 Chr.1: 230778981-230778981 on Build GRCh38
rs1022533849 Chr.1: 230779210-230779210 on Build GRCh38
rs183059984 Chr.1: 230778939-230778939 on Build GRCh38
rs765323395 Chr.1: 230779149-230779149 on Build GRCh38
rs745922166 Chr.1: 230779011-230779011 on Build GRCh38
rs144137595 Chr.1: 230779122-230779122 on Build GRCh38
rs974912164 Chr.1: 230779182-230779182 on Build GRCh38
rs140902814 Chr.1: 230779099-230779099 on Build GRCh38
rs760395648 Chr.1: 230779044-230779044 on Build GRCh38
rs1933631 Chr.1: 230778983-230778983 on Build GRCh38
rs377433609 Chr.1: 230779008-230779008 on Build GRCh38
rs777730830 Chr.1: 230778933-230778933 on Build GRCh38
rs201665088 Chr.1: 230779137-230779137 on Build GRCh38
rs1008646106 Chr.1: 230779141-230779141 on Build GRCh38
rs368103164 Chr.1: 230779140-230779140 on Build GRCh38
rs140805668 Chr.1: 230779025-230779025 on Build GRCh38
rs753743813 Chr.1: 230779053-230779053 on Build GRCh38
rs750327770 Chr.1: 230779153-230779153 on Build GRCh38
rs1046768011 Chr.1: 230778911-230778911 on Build GRCh38
rs1022869624 Chr.1: 230778959-230778959 on Build GRCh38
rs1031619524 Chr.1: 230779260-230779260 on Build GRCh38
rs191588472 Chr.1: 230779108-230779108 on Build GRCh38
rs759032118 Chr.1: 230779098-230779099 on Build GRCh38
rs774020558 Chr.1: 230778969-230778969 on Build GRCh38
rs759137023 Chr.1: 230778972-230778972 on Build GRCh38
rs370629822 Chr.1: 230779036-230779036 on Build GRCh38
rs1019144205 Chr.1: 230779167-230779167 on Build GRCh38
rs766525002 Chr.1: 230779159-230779159 on Build GRCh38
rs186852645 Chr.1: 230779033-230779033 on Build GRCh38
rs553509357 Chr.1: 230778899-230778899 on Build GRCh38
rs370787528 Chr.1: 230779176-230779176 on Build GRCh38
rs776180464 Chr.1: 230779283-230779283 on Build GRCh38
rs748436760 Chr.1: 230779105-230779105 on Build GRCh38
rs749390266 Chr.1: 230778936-230778936 on Build GRCh38
rs751751022 Chr.1: 230779161-230779161 on Build GRCh38
rs532569955 Chr.1: 230778861-230778861 on Build GRCh38
rs767220461 Chr.1: 230779039-230779039 on Build GRCh38
rs745458820 Chr.1: 230778875-230778875 on Build GRCh38
rs978360858 Chr.1: 230779261-230779261 on Build GRCh38
rs750599570 Chr.1: 230779092-230779092 on Build GRCh38
rs28359684 Chr.1: 230779000-230779000 on Build GRCh38
rs750889415 Chr.1: 230779080-230779080 on Build GRCh38
rs923960716 Chr.1: 230779287-230779287 on Build GRCh38
rs374397007 Chr.1: 230779180-230779180 on Build GRCh38
rs758734708 Chr.1: 230779021-230779021 on Build GRCh38
rs886834051 Chr.1: 230778926-230778926 on Build GRCh38
rs774791882 Chr.1: 230779205-230779205 on Build GRCh38
rs747435521 Chr.1: 230779024-230779024 on Build GRCh38
rs16852652 Chr.1: 230779043-230779043 on Build GRCh38
rs571490494 Chr.1: 230778872-230778872 on Build GRCh38
rs373024313 Chr.1: 230779004-230779004 on Build GRCh38
rs767130719 Chr.1: 230778973-230778973 on Build GRCh38
rs769809064 Chr.1: 230778862-230778862 on Build GRCh38
rs12130847 Chr.1: 230778866-230778866 on Build GRCh38

View Full Product Details