Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231339684-231340084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRTN
Entrez Gene ID
83932
Gene Name
SprT-like N-terminal domain
Gene Aliases
C1orf124, DVC1, PRO4323, spartan
UniGene
Hs.554892
Gene Chromosome Location
Chr.1: 231337936-231355029 on Build GRCh38
Transcripts
NM_001010984.3 NM_001261462.2 NM_032018.6 XM_006711818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125382 c.253G>A p.G85S Chr.1: 231339800-231339800 on Build GRCh38
6789327 c.264T>C p.C88C Chr.1: 231339811-231339811 on Build GRCh38
1748219 c.319G>A p.E107K Chr.1: 231339866-231339866 on Build GRCh38
1998430 c.245A>G p.E82G Chr.1: 231339792-231339792 on Build GRCh38
905483 c.321G>T p.E107D Chr.1: 231339868-231339868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780454862 Chr.1: 231339733-231339733 on Build GRCh38
rs752309622 Chr.1: 231339819-231339819 on Build GRCh38
rs370771864 Chr.1: 231339786-231339786 on Build GRCh38
rs777860971 Chr.1: 231340025-231340026 on Build GRCh38
rs202096749 Chr.1: 231339856-231339856 on Build GRCh38
rs535278554 Chr.1: 231340055-231340055 on Build GRCh38
rs919696678 Chr.1: 231339930-231339930 on Build GRCh38
rs543194459 Chr.1: 231339790-231339790 on Build GRCh38
rs981043823 Chr.1: 231339939-231339939 on Build GRCh38
rs780114530 Chr.1: 231339872-231339872 on Build GRCh38
rs3071954 Chr.1: 231340047-231340048 on Build GRCh38
rs910861529 Chr.1: 231339945-231339945 on Build GRCh38
rs527637254 Chr.1: 231340026-231340026 on Build GRCh38
rs762287724 Chr.1: 231339755-231339755 on Build GRCh38
rs1041294818 Chr.1: 231339968-231339968 on Build GRCh38
rs771578058 Chr.1: 231339920-231339920 on Build GRCh38
rs552027191 Chr.1: 231339887-231339887 on Build GRCh38
rs375199797 Chr.1: 231339873-231339873 on Build GRCh38
rs763397666 Chr.1: 231339903-231339903 on Build GRCh38
rs892023725 Chr.1: 231340032-231340032 on Build GRCh38
rs758566090 Chr.1: 231339865-231339865 on Build GRCh38
rs757314351 Chr.1: 231339853-231339853 on Build GRCh38
rs1043583443 Chr.1: 231340055-231340056 on Build GRCh38
rs772463254 Chr.1: 231339730-231339730 on Build GRCh38
rs548769054 Chr.1: 231340056-231340057 on Build GRCh38
rs763864399 Chr.1: 231339842-231339842 on Build GRCh38
rs946221355 Chr.1: 231340054-231340055 on Build GRCh38
rs952898625 Chr.1: 231339958-231339958 on Build GRCh38
rs565651311 Chr.1: 231339955-231339955 on Build GRCh38
rs774740638 Chr.1: 231339881-231339882 on Build GRCh38
rs760311031 Chr.1: 231339826-231339826 on Build GRCh38
rs777051202 Chr.1: 231339744-231339744 on Build GRCh38
rs771522308 Chr.1: 231339807-231339808 on Build GRCh38
rs1014232910 Chr.1: 231340058-231340059 on Build GRCh38
rs781456117 Chr.1: 231339882-231339882 on Build GRCh38
rs895805286 Chr.1: 231340057-231340058 on Build GRCh38
rs766960947 Chr.1: 231339802-231339802 on Build GRCh38
rs770100207 Chr.1: 231339893-231339893 on Build GRCh38
rs1049446829 Chr.1: 231339944-231339944 on Build GRCh38
rs773894329 Chr.1: 231339789-231339789 on Build GRCh38
rs1061065 Chr.1: 231339871-231339871 on Build GRCh38
rs768908695 Chr.1: 231339740-231339740 on Build GRCh38
rs753655266 Chr.1: 231339848-231339848 on Build GRCh38
rs992055507 Chr.1: 231339711-231339711 on Build GRCh38
rs200071813 Chr.1: 231339736-231339736 on Build GRCh38
rs750346304 Chr.1: 231339862-231339862 on Build GRCh38
rs773726644 Chr.1: 231339898-231339898 on Build GRCh38
rs201683566 Chr.1: 231339855-231339855 on Build GRCh38
rs768908236 Chr.1: 231339880-231339880 on Build GRCh38

View Full Product Details