Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231347676-231348198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRTN
Entrez Gene ID
83932
Gene Name
SprT-like N-terminal domain
Gene Aliases
C1orf124, DVC1, PRO4323, spartan
UniGene
Hs.554892
Gene Chromosome Location
Chr.1: 231337936-231355029 on Build GRCh38
Transcripts
NM_001010984.3 NM_001261462.2 NM_032018.6 XM_006711818.2 XM_011544289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5886101 c.323C>T p.T108I Chr.1: 231347798-231347798 on Build GRCh38
5886102 c.11C>T p.T4I Chr.1: 231347798-231347798 on Build GRCh38
5437859 c.113A>G p.N38S Chr.1: 231347900-231347900 on Build GRCh38
5437858 c.425A>G p.N142S Chr.1: 231347900-231347900 on Build GRCh38
905484 c.418C>T p.R140C Chr.1: 231347893-231347893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752516186 Chr.1: 231348027-231348027 on Build GRCh38
rs779507874 Chr.1: 231347973-231347973 on Build GRCh38
rs761491064 Chr.1: 231347865-231347865 on Build GRCh38
rs1021233074 Chr.1: 231347871-231347871 on Build GRCh38
rs758404830 Chr.1: 231347756-231347756 on Build GRCh38
rs145060091 Chr.1: 231347881-231347881 on Build GRCh38
rs942759055 Chr.1: 231348157-231348157 on Build GRCh38
rs149447916 Chr.1: 231347802-231347802 on Build GRCh38
rs193027176 Chr.1: 231347979-231347979 on Build GRCh38
rs755191834 Chr.1: 231347770-231347770 on Build GRCh38
rs766305995 Chr.1: 231347894-231347894 on Build GRCh38
rs959040794 Chr.1: 231347983-231347983 on Build GRCh38
rs370828341 Chr.1: 231347768-231347768 on Build GRCh38
rs142934903 Chr.1: 231347845-231347845 on Build GRCh38
rs145745024 Chr.1: 231348132-231348132 on Build GRCh38
rs372844006 Chr.1: 231347945-231347945 on Build GRCh38
rs530129511 Chr.1: 231347997-231347997 on Build GRCh38
rs202193347 Chr.1: 231347951-231347951 on Build GRCh38
rs539250304 Chr.1: 231348075-231348075 on Build GRCh38
rs140400037 Chr.1: 231347841-231347841 on Build GRCh38
rs368573828 Chr.1: 231347855-231347855 on Build GRCh38
rs776369037 Chr.1: 231347857-231347857 on Build GRCh38
rs142300636 Chr.1: 231348047-231348047 on Build GRCh38
rs149759874 Chr.1: 231347924-231347924 on Build GRCh38
rs997895109 Chr.1: 231347996-231347998 on Build GRCh38
rs990483330 Chr.1: 231348043-231348043 on Build GRCh38
rs546932307 Chr.1: 231347708-231347708 on Build GRCh38
rs773127484 Chr.1: 231347889-231347889 on Build GRCh38
rs200366145 Chr.1: 231347852-231347852 on Build GRCh38
rs767434540 Chr.1: 231347900-231347900 on Build GRCh38
rs918765054 Chr.1: 231348092-231348092 on Build GRCh38
rs755104564 Chr.1: 231347899-231347899 on Build GRCh38
rs767501891 Chr.1: 231347776-231347776 on Build GRCh38
rs539151489 Chr.1: 231348065-231348065 on Build GRCh38
rs759243080 Chr.1: 231347939-231347939 on Build GRCh38
rs764165838 Chr.1: 231348072-231348072 on Build GRCh38
rs527236213 Chr.1: 231347825-231347825 on Build GRCh38
rs147506920 Chr.1: 231347844-231347844 on Build GRCh38
rs375528421 Chr.1: 231347946-231347946 on Build GRCh38
rs763031758 Chr.1: 231347893-231347893 on Build GRCh38
rs190272738 Chr.1: 231347706-231347706 on Build GRCh38
rs552862622 Chr.1: 231348119-231348119 on Build GRCh38
rs781504202 Chr.1: 231347794-231347794 on Build GRCh38
rs893121385 Chr.1: 231348162-231348162 on Build GRCh38
rs929879094 Chr.1: 231347887-231347887 on Build GRCh38
rs113359952 Chr.1: 231347761-231347761 on Build GRCh38
rs751634081 Chr.1: 231347769-231347769 on Build GRCh38
rs545662080 Chr.1: 231347856-231347856 on Build GRCh38
rs574447237 Chr.1: 231347752-231347752 on Build GRCh38
rs1001235256 Chr.1: 231348086-231348086 on Build GRCh38
rs1033159101 Chr.1: 231347777-231347777 on Build GRCh38
rs184987420 Chr.1: 231348124-231348124 on Build GRCh38
rs934153106 Chr.1: 231348095-231348095 on Build GRCh38
rs972996428 Chr.1: 231348070-231348070 on Build GRCh38
rs1000404617 Chr.1: 231347735-231347735 on Build GRCh38
rs1054481530 Chr.1: 231348158-231348158 on Build GRCh38
rs765075867 Chr.1: 231347748-231347748 on Build GRCh38

View Full Product Details