Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 47312898-47312395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF10
Entrez Gene ID
2662
Gene Name
growth differentiation factor 10
Gene Aliases
BMP-3b, BMP3B
UniGene
Hs.2171
Gene Chromosome Location
Chr.10: 47300224-47313577 on Build GRCh38
Transcripts
NM_004962.3

View Full Product Details