Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49165598-49166120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf128
Entrez Gene ID
170371
Gene Name
chromosome 10 open reading frame 128
Gene Aliases
-
UniGene
Hs.385493
Gene Chromosome Location
Chr.10: 49154724-49188580 on Build GRCh38
Transcripts
NM_001010863.2 NM_001288740.1 NM_001288741.1 NM_001288742.1 NM_001288743.1 XM_011539378.2 XM_011539379.1 XM_011539380.1 XM_011539382.1 XM_011539383.2 XM_011539384.1 XM_011539385.1 XM_017015792.1 XM_017015793.1 XM_017015794.1 XM_017015795.1 XM_017015796.1 XM_017015797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1348051 c.249G>A p.P83P Chr.10: 49165786-49165786 on Build GRCh38
6215668 c.248C>T p.P83L Chr.10: 49165787-49165787 on Build GRCh38
5050204 c.249G>A p.P83P Chr.10: 49165786-49165786 on Build GRCh38
5370749 c.266C>T p.P89L Chr.10: 49165769-49165769 on Build GRCh38
6215666 c.248C>T p.P83L Chr.10: 49165787-49165787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781064904 Chr.10: 49165767-49165767 on Build GRCh38
rs1026468720 Chr.10: 49165980-49165980 on Build GRCh38
rs12257132 Chr.10: 49165787-49165787 on Build GRCh38
rs936131661 Chr.10: 49166065-49166065 on Build GRCh38
rs375710736 Chr.10: 49165812-49165812 on Build GRCh38
rs200403793 Chr.10: 49165777-49165777 on Build GRCh38
rs773145171 Chr.10: 49165797-49165798 on Build GRCh38
rs139773718 Chr.10: 49165969-49165969 on Build GRCh38
rs747955473 Chr.10: 49165833-49165833 on Build GRCh38
rs569201583 Chr.10: 49165899-49165899 on Build GRCh38
rs1034591748 Chr.10: 49165773-49165773 on Build GRCh38
rs764570719 Chr.10: 49165791-49165791 on Build GRCh38
rs79009803 Chr.10: 49165813-49165813 on Build GRCh38
rs202117693 Chr.10: 49165770-49165770 on Build GRCh38
rs12243014 Chr.10: 49166041-49166041 on Build GRCh38
rs749247058 Chr.10: 49165845-49165845 on Build GRCh38
rs951854568 Chr.10: 49166023-49166023 on Build GRCh38
rs757385173 Chr.10: 49165734-49165734 on Build GRCh38
rs545765624 Chr.10: 49166033-49166033 on Build GRCh38
rs368560115 Chr.10: 49165946-49165947 on Build GRCh38
rs77059741 Chr.10: 49165693-49165693 on Build GRCh38
rs548261313 Chr.10: 49165916-49165916 on Build GRCh38
rs767427299 Chr.10: 49165745-49165746 on Build GRCh38
rs769577592 Chr.10: 49166028-49166028 on Build GRCh38
rs369694733 Chr.10: 49165702-49165702 on Build GRCh38
rs984796307 Chr.10: 49165628-49165628 on Build GRCh38
rs924606906 Chr.10: 49165905-49165905 on Build GRCh38
rs1045636316 Chr.10: 49166083-49166083 on Build GRCh38
rs1054828437 Chr.10: 49166069-49166069 on Build GRCh38
rs148203576 Chr.10: 49165640-49165640 on Build GRCh38
rs934099754 Chr.10: 49165646-49165646 on Build GRCh38
rs147342689 Chr.10: 49165937-49165937 on Build GRCh38
rs1012791970 Chr.10: 49165955-49165955 on Build GRCh38
rs769773402 Chr.10: 49165843-49165843 on Build GRCh38
rs762542092 Chr.10: 49165776-49165776 on Build GRCh38
rs1000095704 Chr.10: 49165708-49165708 on Build GRCh38
rs945844278 Chr.10: 49165924-49165924 on Build GRCh38
rs533735223 Chr.10: 49165919-49165919 on Build GRCh38
rs977209228 Chr.10: 49166032-49166032 on Build GRCh38
rs566368083 Chr.10: 49165917-49165917 on Build GRCh38
rs924685686 Chr.10: 49166055-49166055 on Build GRCh38
rs977648272 Chr.10: 49166054-49166054 on Build GRCh38
rs768279430 Chr.10: 49165766-49165766 on Build GRCh38
rs753072705 Chr.10: 49165789-49165789 on Build GRCh38
rs749704609 Chr.10: 49165626-49165626 on Build GRCh38
rs528683467 Chr.10: 49165717-49165717 on Build GRCh38
rs931255239 Chr.10: 49166062-49166062 on Build GRCh38
rs901257985 Chr.10: 49165645-49165645 on Build GRCh38
rs567599398 Chr.10: 49165724-49165724 on Build GRCh38
rs763569267 Chr.10: 49165729-49165729 on Build GRCh38
rs975666851 Chr.10: 49166043-49166043 on Build GRCh38
rs971086838 Chr.10: 49165782-49165782 on Build GRCh38
rs959144771 Chr.10: 49165974-49165975 on Build GRCh38
rs760553067 Chr.10: 49165825-49165825 on Build GRCh38
rs993717551 Chr.10: 49166087-49166087 on Build GRCh38
rs1041685468 Chr.10: 49165670-49165670 on Build GRCh38
rs1045501399 Chr.10: 49165967-49165967 on Build GRCh38
rs74322768 Chr.10: 49165908-49165908 on Build GRCh38
rs990302407 Chr.10: 49166020-49166020 on Build GRCh38
rs202171289 Chr.10: 49165786-49165786 on Build GRCh38
rs966692026 Chr.10: 49165838-49165838 on Build GRCh38
rs760862290 Chr.10: 49165858-49165858 on Build GRCh38
rs753092591 Chr.10: 49165722-49165722 on Build GRCh38
rs953494922 Chr.10: 49166009-49166009 on Build GRCh38
rs559419978 Chr.10: 49166086-49166086 on Build GRCh38
rs555906689 Chr.10: 49166029-49166029 on Build GRCh38
rs777852122 Chr.10: 49165844-49165844 on Build GRCh38
rs750563527 Chr.10: 49165814-49165814 on Build GRCh38
rs746107350 Chr.10: 49165764-49165764 on Build GRCh38
rs754408858 Chr.10: 49165804-49165804 on Build GRCh38
rs779104165 Chr.10: 49165759-49165759 on Build GRCh38
rs573527862 Chr.10: 49166071-49166071 on Build GRCh38
rs773336529 Chr.10: 49165772-49165772 on Build GRCh38
rs374736423 Chr.10: 49165832-49165832 on Build GRCh38
rs143681973 Chr.10: 49166072-49166072 on Build GRCh38
rs1055718690 Chr.10: 49165941-49165941 on Build GRCh38
rs143075829 Chr.10: 49165634-49165634 on Build GRCh38
rs370592323 Chr.10: 49165730-49165730 on Build GRCh38
rs147204570 Chr.10: 49166091-49166091 on Build GRCh38
rs541597474 Chr.10: 49166042-49166042 on Build GRCh38
rs564703130 Chr.10: 49165978-49165978 on Build GRCh38
rs200624169 Chr.10: 49166053-49166053 on Build GRCh38
rs866503078 Chr.10: 49165856-49165856 on Build GRCh38
rs373571429 Chr.10: 49165732-49165732 on Build GRCh38
rs760992125 Chr.10: 49165790-49165790 on Build GRCh38
rs117774116 Chr.10: 49165633-49165633 on Build GRCh38
rs773220245 Chr.10: 49165818-49165818 on Build GRCh38
rs922743920 Chr.10: 49166052-49166052 on Build GRCh38
rs990880117 Chr.10: 49165975-49165990 on Build GRCh38
rs1054498762 Chr.10: 49165931-49165931 on Build GRCh38
rs934025397 Chr.10: 49165876-49165876 on Build GRCh38

View Full Product Details