Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49325453-49325945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf71
Entrez Gene ID
118461
Gene Name
chromosome 10 open reading frame 71
Gene Aliases
-
UniGene
Hs.585480
Gene Chromosome Location
Chr.10: 49297009-49330145 on Build GRCh38
Transcripts
NM_001135196.1 XM_005269476.4 XM_005269477.3 XM_005269478.4 XM_017015619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3769126 c.3060C>T p.P1020P Chr.10: 49325605-49325605 on Build GRCh38
5628222 c.3389C>T p.S1130L Chr.10: 49325934-49325934 on Build GRCh38
5606364 c.3305C>T p.S1102F Chr.10: 49325850-49325850 on Build GRCh38
1348055 c.2941G>A p.V981M Chr.10: 49325486-49325486 on Build GRCh38
6530830 c.3271G>C p.E1091Q Chr.10: 49325816-49325816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039010763 Chr.10: 49325769-49325769 on Build GRCh38
rs368250068 Chr.10: 49325722-49325722 on Build GRCh38
rs371013291 Chr.10: 49325631-49325631 on Build GRCh38
rs746622246 Chr.10: 49325613-49325613 on Build GRCh38
rs925179387 Chr.10: 49325672-49325672 on Build GRCh38
rs950034709 Chr.10: 49325712-49325712 on Build GRCh38
rs971001162 Chr.10: 49325909-49325909 on Build GRCh38
rs909913986 Chr.10: 49325728-49325728 on Build GRCh38
rs751668217 Chr.10: 49325720-49325720 on Build GRCh38
rs748444986 Chr.10: 49325891-49325891 on Build GRCh38
rs868200891 Chr.10: 49325694-49325694 on Build GRCh38
rs769445302 Chr.10: 49325705-49325705 on Build GRCh38
rs866082565 Chr.10: 49325506-49325506 on Build GRCh38
rs963959821 Chr.10: 49325634-49325634 on Build GRCh38
rs778976495 Chr.10: 49325573-49325573 on Build GRCh38
rs551539055 Chr.10: 49325725-49325725 on Build GRCh38
rs766572381 Chr.10: 49325485-49325485 on Build GRCh38
rs746547019 Chr.10: 49325836-49325836 on Build GRCh38
rs773763574 Chr.10: 49325707-49325707 on Build GRCh38
rs546884682 Chr.10: 49325920-49325920 on Build GRCh38
rs906075875 Chr.10: 49325503-49325503 on Build GRCh38
rs751827587 Chr.10: 49325507-49325507 on Build GRCh38
rs918753234 Chr.10: 49325711-49325711 on Build GRCh38
rs755904916 Chr.10: 49325533-49325533 on Build GRCh38
rs376629653 Chr.10: 49325776-49325776 on Build GRCh38
rs955052415 Chr.10: 49325593-49325593 on Build GRCh38
rs767880029 Chr.10: 49325727-49325727 on Build GRCh38
rs935140987 Chr.10: 49325689-49325689 on Build GRCh38
rs1007856006 Chr.10: 49325816-49325816 on Build GRCh38
rs1023900263 Chr.10: 49325908-49325908 on Build GRCh38
rs187842723 Chr.10: 49325567-49325567 on Build GRCh38
rs11101094 Chr.10: 49325796-49325796 on Build GRCh38
rs541315612 Chr.10: 49325696-49325696 on Build GRCh38
rs111416939 Chr.10: 49325863-49325863 on Build GRCh38
rs1012546449 Chr.10: 49325897-49325897 on Build GRCh38
rs190338188 Chr.10: 49325780-49325780 on Build GRCh38
rs750427970 Chr.10: 49325785-49325785 on Build GRCh38
rs779213214 Chr.10: 49325730-49325730 on Build GRCh38
rs763007049 Chr.10: 49325714-49325714 on Build GRCh38
rs979749435 Chr.10: 49325661-49325661 on Build GRCh38
rs866006410 Chr.10: 49325850-49325850 on Build GRCh38
rs71471341 Chr.10: 49325730-49325731 on Build GRCh38
rs866092737 Chr.10: 49325604-49325604 on Build GRCh38
rs937649951 Chr.10: 49325764-49325764 on Build GRCh38
rs766484687 Chr.10: 49325719-49325719 on Build GRCh38
rs866542893 Chr.10: 49325735-49325735 on Build GRCh38
rs545241202 Chr.10: 49325566-49325566 on Build GRCh38
rs1041977194 Chr.10: 49325815-49325815 on Build GRCh38
rs779665299 Chr.10: 49325819-49325819 on Build GRCh38
rs10857471 Chr.10: 49325731-49325731 on Build GRCh38
rs770899878 Chr.10: 49325864-49325864 on Build GRCh38
rs1046033025 Chr.10: 49325486-49325486 on Build GRCh38
rs1002119156 Chr.10: 49325520-49325520 on Build GRCh38
rs761308417 Chr.10: 49325843-49325843 on Build GRCh38
rs778974852 Chr.10: 49325779-49325779 on Build GRCh38
rs770240050 Chr.10: 49325918-49325918 on Build GRCh38
rs745360386 Chr.10: 49325595-49325595 on Build GRCh38
rs1000379148 Chr.10: 49325820-49325820 on Build GRCh38
rs2377940 Chr.10: 49325572-49325572 on Build GRCh38
rs770192781 Chr.10: 49325695-49325695 on Build GRCh38
rs532189362 Chr.10: 49325710-49325710 on Build GRCh38
rs567505616 Chr.10: 49325821-49325821 on Build GRCh38
rs762239726 Chr.10: 49325647-49325647 on Build GRCh38
rs115787597 Chr.10: 49325605-49325605 on Build GRCh38
rs563008657 Chr.10: 49325606-49325606 on Build GRCh38

View Full Product Details