Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49679546-49680035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf53
Entrez Gene ID
282966
Gene Name
chromosome 10 open reading frame 53
Gene Aliases
-
UniGene
Hs.131287
Gene Chromosome Location
Chr.10: 49679638-49708910 on Build GRCh38
Transcripts
NM_001042427.1 NM_182554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370768 c.83T>C p.L28P Chr.10: 49679780-49679780 on Build GRCh38
6411957 c.40T>C p.Y14H Chr.10: 49679737-49679737 on Build GRCh38
3686958 c.79C>T p.R27C Chr.10: 49679776-49679776 on Build GRCh38
4955250 c.19G>C p.V7L Chr.10: 49679716-49679716 on Build GRCh38
4955251 c.19G>C p.V7L Chr.10: 49679716-49679716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008427620 Chr.10: 49679772-49679772 on Build GRCh38
rs779398271 Chr.10: 49679672-49679672 on Build GRCh38
rs920124414 Chr.10: 49679752-49679752 on Build GRCh38
rs932397860 Chr.10: 49680000-49680000 on Build GRCh38
rs558431917 Chr.10: 49679746-49679746 on Build GRCh38
rs760695647 Chr.10: 49679833-49679833 on Build GRCh38
rs762004569 Chr.10: 49679763-49679763 on Build GRCh38
rs767354624 Chr.10: 49679826-49679826 on Build GRCh38
rs533772421 Chr.10: 49679912-49679912 on Build GRCh38
rs779096776 Chr.10: 49679590-49679590 on Build GRCh38
rs934291456 Chr.10: 49679897-49679897 on Build GRCh38
rs543381535 Chr.10: 49679647-49679647 on Build GRCh38
rs918962923 Chr.10: 49679958-49679958 on Build GRCh38
rs759380407 Chr.10: 49679817-49679817 on Build GRCh38
rs548197754 Chr.10: 49679873-49679873 on Build GRCh38
rs528678218 Chr.10: 49679668-49679668 on Build GRCh38
rs1047738863 Chr.10: 49679591-49679591 on Build GRCh38
rs116463656 Chr.10: 49679858-49679858 on Build GRCh38
rs774124565 Chr.10: 49679815-49679815 on Build GRCh38
rs961659568 Chr.10: 49679775-49679775 on Build GRCh38
rs763462062 Chr.10: 49679836-49679836 on Build GRCh38
rs750163553 Chr.10: 49679680-49679680 on Build GRCh38
rs767875589 Chr.10: 49679635-49679635 on Build GRCh38
rs902834084 Chr.10: 49679685-49679685 on Build GRCh38
rs752507814 Chr.10: 49679832-49679832 on Build GRCh38
rs959536199 Chr.10: 49679812-49679812 on Build GRCh38
rs908533564 Chr.10: 49679624-49679624 on Build GRCh38
rs898645549 Chr.10: 49679583-49679583 on Build GRCh38
rs917790072 Chr.10: 49679684-49679685 on Build GRCh38
rs547143816 Chr.10: 49679702-49679702 on Build GRCh38
rs377321406 Chr.10: 49679726-49679726 on Build GRCh38
rs930101936 Chr.10: 49679754-49679754 on Build GRCh38
rs891136519 Chr.10: 49679600-49679600 on Build GRCh38
rs540145478 Chr.10: 49679588-49679588 on Build GRCh38
rs982592873 Chr.10: 49679630-49679630 on Build GRCh38
rs901025591 Chr.10: 49679713-49679713 on Build GRCh38
rs11367300 Chr.10: 49679851-49679851 on Build GRCh38
rs550656341 Chr.10: 49679734-49679734 on Build GRCh38
rs775803976 Chr.10: 49679738-49679738 on Build GRCh38
rs1040805364 Chr.10: 49679901-49679901 on Build GRCh38
rs1000925449 Chr.10: 49679810-49679810 on Build GRCh38
rs566713074 Chr.10: 49679906-49679906 on Build GRCh38
rs149069015 Chr.10: 49679757-49679757 on Build GRCh38
rs1041168483 Chr.10: 49679703-49679703 on Build GRCh38
rs980669969 Chr.10: 49679882-49679882 on Build GRCh38
rs1038581537 Chr.10: 49679576-49679576 on Build GRCh38
rs1028197220 Chr.10: 49679729-49679729 on Build GRCh38
rs561585871 Chr.10: 49679658-49679658 on Build GRCh38
rs1037837931 Chr.10: 49679747-49679747 on Build GRCh38
rs1153781 Chr.10: 49679605-49679605 on Build GRCh38
rs888402282 Chr.10: 49679735-49679735 on Build GRCh38
rs374622847 Chr.10: 49679736-49679736 on Build GRCh38
rs928179660 Chr.10: 49679634-49679634 on Build GRCh38
rs998637859 Chr.10: 49679691-49679691 on Build GRCh38
rs746301424 Chr.10: 49679676-49679676 on Build GRCh38
rs996735701 Chr.10: 49679588-49679588 on Build GRCh38
rs973189395 Chr.10: 49679916-49679916 on Build GRCh38
rs536237267 Chr.10: 49679862-49679862 on Build GRCh38
rs924317654 Chr.10: 49679894-49679894 on Build GRCh38
rs961516211 Chr.10: 49679629-49679629 on Build GRCh38
rs1042769676 Chr.10: 49679684-49679684 on Build GRCh38
rs1032200846 Chr.10: 49679667-49679667 on Build GRCh38
rs978731162 Chr.10: 49679861-49679861 on Build GRCh38
rs144285786 Chr.10: 49679781-49679781 on Build GRCh38
rs956723914 Chr.10: 49679674-49679675 on Build GRCh38
rs1035488898 Chr.10: 49679802-49679802 on Build GRCh38
rs988090599 Chr.10: 49679676-49679678 on Build GRCh38
rs899828485 Chr.10: 49679807-49679807 on Build GRCh38
rs571664551 Chr.10: 49679744-49679744 on Build GRCh38
rs1028166504 Chr.10: 49679595-49679595 on Build GRCh38
rs970382705 Chr.10: 49679688-49679688 on Build GRCh38
rs984046285 Chr.10: 49679623-49679623 on Build GRCh38
rs1055398343 Chr.10: 49679648-49679648 on Build GRCh38

View Full Product Details