Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49694453-49694947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf53
Entrez Gene ID
282966
Gene Name
chromosome 10 open reading frame 53
Gene Aliases
-
UniGene
Hs.131287
Gene Chromosome Location
Chr.10: 49679638-49708910 on Build GRCh38
Transcripts
NM_001042427.1 NM_182554.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
685076 c.218-10G>A p.? Chr.10: 49694528-49694528 on Build GRCh38
6129566 c.254C>A p.A85D Chr.10: 49694574-49694574 on Build GRCh38
6517858 c.263C>A p.A88D Chr.10: 49694583-49694583 on Build GRCh38
5836953 c.218G>A p.G73E Chr.10: 49694538-49694538 on Build GRCh38
5703627 c.265G>A p.V89M Chr.10: 49694585-49694585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922740612 Chr.10: 49694801-49694801 on Build GRCh38
rs376184767 Chr.10: 49694586-49694586 on Build GRCh38
rs995476597 Chr.10: 49694517-49694517 on Build GRCh38
rs868708085 Chr.10: 49694770-49694770 on Build GRCh38
rs1033520175 Chr.10: 49694814-49694814 on Build GRCh38
rs749424513 Chr.10: 49694488-49694488 on Build GRCh38
rs966605929 Chr.10: 49694863-49694863 on Build GRCh38
rs1003362517 Chr.10: 49694595-49694595 on Build GRCh38
rs1049973768 Chr.10: 49694828-49694828 on Build GRCh38
rs777704145 Chr.10: 49694598-49694598 on Build GRCh38
rs11101213 Chr.10: 49694604-49694604 on Build GRCh38
rs943406228 Chr.10: 49694661-49694661 on Build GRCh38
rs775829298 Chr.10: 49694628-49694628 on Build GRCh38
rs369526864 Chr.10: 49694561-49694561 on Build GRCh38
rs940210957 Chr.10: 49694476-49694476 on Build GRCh38
rs145249659 Chr.10: 49694547-49694547 on Build GRCh38
rs763455967 Chr.10: 49694728-49694728 on Build GRCh38
rs1007756048 Chr.10: 49694495-49694495 on Build GRCh38
rs756581082 Chr.10: 49694557-49694557 on Build GRCh38
rs1042770973 Chr.10: 49694881-49694881 on Build GRCh38
rs769845045 Chr.10: 49694596-49694596 on Build GRCh38
rs771753672 Chr.10: 49694626-49694626 on Build GRCh38
rs1024581784 Chr.10: 49694749-49694749 on Build GRCh38
rs1055421 Chr.10: 49694638-49694638 on Build GRCh38
rs906302461 Chr.10: 49694763-49694763 on Build GRCh38
rs907609575 Chr.10: 49694475-49694475 on Build GRCh38
rs902620304 Chr.10: 49694883-49694883 on Build GRCh38
rs375700971 Chr.10: 49694537-49694537 on Build GRCh38
rs922877276 Chr.10: 49694528-49694528 on Build GRCh38
rs779138998 Chr.10: 49694496-49694496 on Build GRCh38
rs201121438 Chr.10: 49694635-49694635 on Build GRCh38
rs771177398 Chr.10: 49694588-49694588 on Build GRCh38
rs765816348 Chr.10: 49694542-49694542 on Build GRCh38
rs899292744 Chr.10: 49694687-49694687 on Build GRCh38
rs775358317 Chr.10: 49694695-49694695 on Build GRCh38
rs866616179 Chr.10: 49694484-49694484 on Build GRCh38
rs958076638 Chr.10: 49694815-49694815 on Build GRCh38
rs757295118 Chr.10: 49694579-49694579 on Build GRCh38
rs772597208 Chr.10: 49694531-49694531 on Build GRCh38
rs149157578 Chr.10: 49694860-49694860 on Build GRCh38
rs550422267 Chr.10: 49694862-49694862 on Build GRCh38
rs1041215586 Chr.10: 49694478-49694478 on Build GRCh38
rs746083829 Chr.10: 49694497-49694497 on Build GRCh38
rs759169510 Chr.10: 49694622-49694622 on Build GRCh38
rs911873135 Chr.10: 49694867-49694867 on Build GRCh38
rs781106770 Chr.10: 49694587-49694587 on Build GRCh38
rs987483091 Chr.10: 49694865-49694865 on Build GRCh38
rs532892675 Chr.10: 49694688-49694688 on Build GRCh38
rs965772251 Chr.10: 49694760-49694760 on Build GRCh38
rs748079329 Chr.10: 49694515-49694515 on Build GRCh38
rs761507691 Chr.10: 49694891-49694891 on Build GRCh38
rs778892175 Chr.10: 49694580-49694580 on Build GRCh38
rs975200200 Chr.10: 49694907-49694907 on Build GRCh38
rs754329632 Chr.10: 49694631-49694631 on Build GRCh38
rs372537686 Chr.10: 49694610-49694610 on Build GRCh38
rs7069935 Chr.10: 49694572-49694572 on Build GRCh38
rs185091658 Chr.10: 49694694-49694694 on Build GRCh38
rs1011414318 Chr.10: 49694575-49694575 on Build GRCh38
rs746055820 Chr.10: 49694585-49694585 on Build GRCh38
rs1039224345 Chr.10: 49694684-49694684 on Build GRCh38
rs758685961 Chr.10: 49694646-49694646 on Build GRCh38
rs769895982 Chr.10: 49694520-49694520 on Build GRCh38
rs1002338230 Chr.10: 49694772-49694772 on Build GRCh38
rs777917968 Chr.10: 49694574-49694574 on Build GRCh38
rs747890674 Chr.10: 49694525-49694526 on Build GRCh38
rs375603494 Chr.10: 49694630-49694630 on Build GRCh38
rs1042508398 Chr.10: 49694576-49694576 on Build GRCh38
rs753107732 Chr.10: 49694556-49694556 on Build GRCh38
rs764652108 Chr.10: 49694565-49694565 on Build GRCh38
rs768533193 Chr.10: 49694503-49694503 on Build GRCh38
rs780477616 Chr.10: 49694653-49694653 on Build GRCh38
rs899428070 Chr.10: 49694504-49694504 on Build GRCh38
rs776592651 Chr.10: 49694510-49694510 on Build GRCh38
rs769741284 Chr.10: 49694639-49694639 on Build GRCh38
rs41281963 Chr.10: 49694719-49694719 on Build GRCh38
rs751110375 Chr.10: 49694748-49694748 on Build GRCh38
rs369717123 Chr.10: 49694543-49694543 on Build GRCh38
rs1048486570 Chr.10: 49694698-49694698 on Build GRCh38
rs976930628 Chr.10: 49694799-49694799 on Build GRCh38
rs989318863 Chr.10: 49694820-49694820 on Build GRCh38
rs757415199 Chr.10: 49694492-49694492 on Build GRCh38
rs749191013 Chr.10: 49694600-49694600 on Build GRCh38
rs943439130 Chr.10: 49694875-49694875 on Build GRCh38

View Full Product Details