Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110008166-110008693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
UniGene
Hs.501012
Gene Chromosome Location
Chr.10: 109996350-110135565 on Build GRCh38
Transcripts
NM_001121.3 NM_001320591.1 NM_001320592.1 NM_001320593.1 NM_001320594.1 NM_016824.4 NM_019903.4 NR_038943.1

Gene Symbol
ADD3-AS1
Entrez Gene ID
100505933
Gene Name
ADD3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.604103
Gene Chromosome Location
Chr.10: 109945559-110008381 on Build GRCh38
Transcripts
NM_001121.3 NM_001320591.1 NM_001320592.1 NM_001320593.1 NM_001320594.1 NM_016824.4 NM_019903.4 NR_038943.1

SNP Detail

SNP ID Chromosome Location
rs1018762258 Chr.10: 110008391-110008391 on Build GRCh38
rs904275573 Chr.10: 110008262-110008262 on Build GRCh38
rs962802094 Chr.10: 110008558-110008558 on Build GRCh38
rs989883857 Chr.10: 110008618-110008618 on Build GRCh38
rs994860702 Chr.10: 110008514-110008514 on Build GRCh38
rs933117451 Chr.10: 110008536-110008536 on Build GRCh38
rs895635036 Chr.10: 110008245-110008245 on Build GRCh38
rs1018372773 Chr.10: 110008286-110008286 on Build GRCh38
rs1029914553 Chr.10: 110008530-110008530 on Build GRCh38
rs964654767 Chr.10: 110008189-110008189 on Build GRCh38
rs4918474 Chr.10: 110008631-110008631 on Build GRCh38
rs1045268804 Chr.10: 110008232-110008232 on Build GRCh38
rs373204485 Chr.10: 110008655-110008655 on Build GRCh38
rs1020030794 Chr.10: 110008252-110008252 on Build GRCh38
rs745991726 Chr.10: 110008239-110008239 on Build GRCh38
rs752622560 Chr.10: 110008586-110008586 on Build GRCh38
rs111423248 Chr.10: 110008300-110008300 on Build GRCh38
rs1056969794 Chr.10: 110008427-110008427 on Build GRCh38
rs1031898382 Chr.10: 110008612-110008612 on Build GRCh38
rs577071118 Chr.10: 110008202-110008202 on Build GRCh38
rs897913477 Chr.10: 110008509-110008509 on Build GRCh38
rs541225233 Chr.10: 110008255-110008255 on Build GRCh38
rs1004058545 Chr.10: 110008244-110008244 on Build GRCh38
rs111247100 Chr.10: 110008596-110008596 on Build GRCh38
rs772703022 Chr.10: 110008340-110008340 on Build GRCh38
rs546766081 Chr.10: 110008580-110008580 on Build GRCh38
rs939525784 Chr.10: 110008466-110008466 on Build GRCh38
rs976891231 Chr.10: 110008192-110008192 on Build GRCh38
rs992123188 Chr.10: 110008575-110008575 on Build GRCh38
rs954007517 Chr.10: 110008238-110008255 on Build GRCh38
rs915550729 Chr.10: 110008438-110008438 on Build GRCh38
rs976832702 Chr.10: 110008227-110008228 on Build GRCh38
rs909989130 Chr.10: 110008213-110008213 on Build GRCh38
rs926635174 Chr.10: 110008398-110008398 on Build GRCh38
rs964306416 Chr.10: 110008318-110008318 on Build GRCh38
rs905745488 Chr.10: 110008223-110008223 on Build GRCh38
rs982849552 Chr.10: 110008208-110008208 on Build GRCh38
rs139466147 Chr.10: 110008487-110008487 on Build GRCh38
rs1017531953 Chr.10: 110008557-110008557 on Build GRCh38
rs997883054 Chr.10: 110008504-110008504 on Build GRCh38
rs904245329 Chr.10: 110008457-110008457 on Build GRCh38
rs982985526 Chr.10: 110008371-110008371 on Build GRCh38
rs922695730 Chr.10: 110008228-110008245 on Build GRCh38
rs759277552 Chr.10: 110008574-110008574 on Build GRCh38
rs953316480 Chr.10: 110008328-110008328 on Build GRCh38
rs944467409 Chr.10: 110008205-110008205 on Build GRCh38
rs1041530664 Chr.10: 110008585-110008585 on Build GRCh38
rs552921230 Chr.10: 110008294-110008294 on Build GRCh38
rs575239353 Chr.10: 110008458-110008458 on Build GRCh38
rs994858025 Chr.10: 110008290-110008290 on Build GRCh38
rs1004349052 Chr.10: 110008546-110008546 on Build GRCh38
rs535151209 Chr.10: 110008226-110008226 on Build GRCh38
rs551012263 Chr.10: 110008597-110008597 on Build GRCh38
rs558687615 Chr.10: 110008201-110008201 on Build GRCh38
rs1056870159 Chr.10: 110008230-110008230 on Build GRCh38
rs965889101 Chr.10: 110008392-110008392 on Build GRCh38
rs935881060 Chr.10: 110008588-110008588 on Build GRCh38
rs980767247 Chr.10: 110008395-110008395 on Build GRCh38
rs563675266 Chr.10: 110008450-110008450 on Build GRCh38
rs780454897 Chr.10: 110008656-110008656 on Build GRCh38
rs977184975 Chr.10: 110008206-110008206 on Build GRCh38
rs751153117 Chr.10: 110008543-110008543 on Build GRCh38
rs184112420 Chr.10: 110008638-110008638 on Build GRCh38
rs536105963 Chr.10: 110008622-110008622 on Build GRCh38
rs957308205 Chr.10: 110008411-110008411 on Build GRCh38
rs999832280 Chr.10: 110008266-110008266 on Build GRCh38
rs1045473882 Chr.10: 110008222-110008222 on Build GRCh38
rs545854688 Chr.10: 110008459-110008459 on Build GRCh38
rs972259204 Chr.10: 110008272-110008272 on Build GRCh38
rs574661671 Chr.10: 110008347-110008347 on Build GRCh38
rs1010058835 Chr.10: 110008248-110008248 on Build GRCh38
rs750548481 Chr.10: 110008616-110008616 on Build GRCh38
rs909928063 Chr.10: 110008387-110008387 on Build GRCh38
rs939377915 Chr.10: 110008417-110008417 on Build GRCh38
rs951610149 Chr.10: 110008534-110008534 on Build GRCh38
rs7076609 Chr.10: 110008393-110008393 on Build GRCh38
rs981245149 Chr.10: 110008565-110008565 on Build GRCh38
rs899621294 Chr.10: 110008614-110008614 on Build GRCh38
rs937271482 Chr.10: 110008225-110008225 on Build GRCh38
rs569586780 Chr.10: 110008623-110008623 on Build GRCh38
rs941366473 Chr.10: 110008219-110008219 on Build GRCh38
rs945844227 Chr.10: 110008435-110008435 on Build GRCh38
rs976986122 Chr.10: 110008325-110008325 on Build GRCh38
rs765474733 Chr.10: 110008613-110008613 on Build GRCh38
rs767291067 Chr.10: 110008584-110008584 on Build GRCh38
rs895778157 Chr.10: 110008434-110008434 on Build GRCh38
rs897857193 Chr.10: 110008281-110008281 on Build GRCh38
rs562175782 Chr.10: 110008587-110008587 on Build GRCh38
rs528608459 Chr.10: 110008567-110008567 on Build GRCh38

View Full Product Details