Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110100543-110101043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
UniGene
Hs.501012
Gene Chromosome Location
Chr.10: 109996350-110135565 on Build GRCh38
Transcripts
NM_001121.3 NM_001320591.1 NM_001320592.1 NM_001320593.1 NM_001320594.1 NM_016824.4 NM_019903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011539 c.27G>A p.V9V Chr.10: 110100680-110100680 on Build GRCh38
3985412 c.32C>G p.T11S Chr.10: 110100685-110100685 on Build GRCh38
1474341 c.18C>T p.S6S Chr.10: 110100671-110100671 on Build GRCh38
242853 c.25G>A p.V9M Chr.10: 110100678-110100678 on Build GRCh38
1193828 c.56C>T p.P19L Chr.10: 110100709-110100709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201508110 Chr.10: 110100858-110100858 on Build GRCh38
rs777166861 Chr.10: 110100676-110100676 on Build GRCh38
rs764083258 Chr.10: 110100629-110100629 on Build GRCh38
rs551499537 Chr.10: 110100633-110100633 on Build GRCh38
rs981069213 Chr.10: 110100873-110100873 on Build GRCh38
rs761201144 Chr.10: 110100774-110100774 on Build GRCh38
rs957291036 Chr.10: 110100713-110100713 on Build GRCh38
rs376781474 Chr.10: 110100836-110100836 on Build GRCh38
rs745720198 Chr.10: 110100683-110100683 on Build GRCh38
rs1041581889 Chr.10: 110100976-110100976 on Build GRCh38
rs868119118 Chr.10: 110100733-110100733 on Build GRCh38
rs41291884 Chr.10: 110100647-110100647 on Build GRCh38
rs769661014 Chr.10: 110100842-110100842 on Build GRCh38
rs1055052157 Chr.10: 110100928-110100928 on Build GRCh38
rs1006752741 Chr.10: 110100704-110100704 on Build GRCh38
rs775197377 Chr.10: 110100696-110100696 on Build GRCh38
rs749078661 Chr.10: 110100845-110100845 on Build GRCh38
rs370819901 Chr.10: 110100609-110100609 on Build GRCh38
rs769573055 Chr.10: 110100694-110100694 on Build GRCh38
rs772284791 Chr.10: 110100887-110100887 on Build GRCh38
rs770809843 Chr.10: 110100678-110100678 on Build GRCh38
rs761931587 Chr.10: 110100772-110100772 on Build GRCh38
rs756745987 Chr.10: 110100829-110100829 on Build GRCh38
rs118066002 Chr.10: 110101002-110101002 on Build GRCh38
rs185480644 Chr.10: 110100668-110100668 on Build GRCh38
rs958530450 Chr.10: 110100667-110100667 on Build GRCh38
rs975652096 Chr.10: 110100737-110100737 on Build GRCh38
rs945886114 Chr.10: 110100932-110100932 on Build GRCh38
rs761868855 Chr.10: 110100865-110100865 on Build GRCh38
rs962525828 Chr.10: 110100722-110100722 on Build GRCh38
rs766867209 Chr.10: 110100786-110100786 on Build GRCh38
rs754899382 Chr.10: 110100907-110100907 on Build GRCh38
rs569697030 Chr.10: 110100985-110100985 on Build GRCh38
rs750965183 Chr.10: 110100773-110100773 on Build GRCh38
rs554589265 Chr.10: 110100996-110100996 on Build GRCh38
rs150532955 Chr.10: 110100739-110100739 on Build GRCh38
rs372253017 Chr.10: 110100815-110100815 on Build GRCh38
rs937055499 Chr.10: 110100913-110100913 on Build GRCh38
rs776523907 Chr.10: 110100680-110100680 on Build GRCh38
rs773371621 Chr.10: 110100897-110100897 on Build GRCh38
rs775676054 Chr.10: 110100625-110100625 on Build GRCh38
rs770969233 Chr.10: 110100836-110100837 on Build GRCh38
rs757535114 Chr.10: 110100904-110100904 on Build GRCh38
rs367830044 Chr.10: 110100677-110100677 on Build GRCh38
rs967127374 Chr.10: 110100679-110100679 on Build GRCh38
rs937129133 Chr.10: 110100590-110100590 on Build GRCh38
rs898508318 Chr.10: 110100600-110100600 on Build GRCh38
rs758014351 Chr.10: 110100790-110100790 on Build GRCh38
rs967641759 Chr.10: 110100998-110100998 on Build GRCh38
rs546910906 Chr.10: 110100903-110100903 on Build GRCh38
rs142566562 Chr.10: 110100707-110100707 on Build GRCh38
rs371176610 Chr.10: 110100670-110100670 on Build GRCh38
rs751609758 Chr.10: 110100631-110100631 on Build GRCh38
rs563622821 Chr.10: 110100632-110100632 on Build GRCh38
rs373971493 Chr.10: 110100618-110100618 on Build GRCh38
rs972313098 Chr.10: 110100726-110100726 on Build GRCh38
rs759292355 Chr.10: 110100615-110100615 on Build GRCh38
rs779768563 Chr.10: 110100688-110100690 on Build GRCh38
rs781170581 Chr.10: 110100831-110100831 on Build GRCh38
rs766133787 Chr.10: 110100729-110100729 on Build GRCh38
rs368066210 Chr.10: 110100661-110100661 on Build GRCh38
rs763157652 Chr.10: 110100626-110100626 on Build GRCh38
rs756536764 Chr.10: 110100643-110100643 on Build GRCh38
rs189792852 Chr.10: 110100851-110100851 on Build GRCh38
rs139486472 Chr.10: 110100827-110100827 on Build GRCh38
rs373682804 Chr.10: 110100673-110100673 on Build GRCh38
rs894362840 Chr.10: 110100960-110100960 on Build GRCh38
rs762838979 Chr.10: 110100708-110100708 on Build GRCh38
rs764111338 Chr.10: 110100908-110100908 on Build GRCh38
rs7090030 Chr.10: 110100915-110100915 on Build GRCh38
rs768885657 Chr.10: 110100732-110100732 on Build GRCh38
rs1014141596 Chr.10: 110100978-110100978 on Build GRCh38

View Full Product Details