Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114043860-114044389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRB1
Entrez Gene ID
153
Gene Name
adrenoceptor beta 1
Gene Aliases
ADRB1R, B1AR, BETA1AR, RHR
UniGene
Hs.99913
Gene Chromosome Location
Chr.10: 114044047-114046908 on Build GRCh38
Transcripts
NM_000684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6586375 c.50C>A p.S17* Chr.10: 114044182-114044182 on Build GRCh38
6045529 c.158T>C p.L53P Chr.10: 114044290-114044290 on Build GRCh38
5049902 c.239T>C p.I80T Chr.10: 114044371-114044371 on Build GRCh38
5796998 c.202A>C p.I68L Chr.10: 114044334-114044334 on Build GRCh38
4480326 c.240C>T p.I80I Chr.10: 114044372-114044372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373125312 Chr.10: 114044364-114044364 on Build GRCh38
rs754908383 Chr.10: 114044230-114044230 on Build GRCh38
rs753765187 Chr.10: 114044228-114044228 on Build GRCh38
rs560283907 Chr.10: 114044176-114044176 on Build GRCh38
rs35476499 Chr.10: 114044237-114044237 on Build GRCh38
rs943949932 Chr.10: 114044040-114044040 on Build GRCh38
rs1041199660 Chr.10: 114044010-114044010 on Build GRCh38
rs1047376625 Chr.10: 114044347-114044347 on Build GRCh38
rs368596680 Chr.10: 114044255-114044255 on Build GRCh38
rs766122769 Chr.10: 114044218-114044218 on Build GRCh38
rs371084372 Chr.10: 114044249-114044249 on Build GRCh38
rs960191966 Chr.10: 114043921-114043921 on Build GRCh38
rs238743 Chr.10: 114043890-114043890 on Build GRCh38
rs375635406 Chr.10: 114044251-114044251 on Build GRCh38
rs34072930 Chr.10: 114044222-114044222 on Build GRCh38
rs775700146 Chr.10: 114044269-114044269 on Build GRCh38
rs768864377 Chr.10: 114044271-114044271 on Build GRCh38
rs764795066 Chr.10: 114044234-114044234 on Build GRCh38
rs987838185 Chr.10: 114043959-114043959 on Build GRCh38
rs530691478 Chr.10: 114043929-114043929 on Build GRCh38
rs752633662 Chr.10: 114044288-114044288 on Build GRCh38
rs369867546 Chr.10: 114044246-114044246 on Build GRCh38
rs943761104 Chr.10: 114044006-114044006 on Build GRCh38
rs577981525 Chr.10: 114044141-114044141 on Build GRCh38
rs757591980 Chr.10: 114044254-114044254 on Build GRCh38
rs1014314304 Chr.10: 114044087-114044087 on Build GRCh38
rs35720093 Chr.10: 114044217-114044217 on Build GRCh38
rs1023492154 Chr.10: 114044124-114044124 on Build GRCh38
rs72556521 Chr.10: 114044250-114044250 on Build GRCh38
rs537853447 Chr.10: 114043968-114043968 on Build GRCh38
rs1014696477 Chr.10: 114044316-114044316 on Build GRCh38
rs879056623 Chr.10: 114043940-114043945 on Build GRCh38
rs1801252 Chr.10: 114044277-114044277 on Build GRCh38
rs901569872 Chr.10: 114044290-114044290 on Build GRCh38
rs749325026 Chr.10: 114044270-114044270 on Build GRCh38
rs1036995848 Chr.10: 114044070-114044070 on Build GRCh38
rs1006052166 Chr.10: 114044361-114044361 on Build GRCh38
rs116403933 Chr.10: 114044339-114044339 on Build GRCh38
rs888733720 Chr.10: 114044048-114044048 on Build GRCh38
rs1024775815 Chr.10: 114043922-114043930 on Build GRCh38
rs750812061 Chr.10: 114044333-114044333 on Build GRCh38
rs1014758367 Chr.10: 114043922-114043924 on Build GRCh38
rs1004167974 Chr.10: 114044060-114044060 on Build GRCh38
rs545019520 Chr.10: 114044146-114044146 on Build GRCh38
rs768811340 Chr.10: 114044363-114044363 on Build GRCh38
rs759501745 Chr.10: 114044287-114044287 on Build GRCh38
rs925216195 Chr.10: 114043934-114043934 on Build GRCh38
rs556229079 Chr.10: 114044041-114044041 on Build GRCh38
rs895813743 Chr.10: 114044075-114044075 on Build GRCh38
rs751191292 Chr.10: 114044309-114044309 on Build GRCh38
rs780643195 Chr.10: 114044342-114044342 on Build GRCh38
rs1002068782 Chr.10: 114044171-114044171 on Build GRCh38
rs34844626 Chr.10: 114044209-114044209 on Build GRCh38
rs1015625816 Chr.10: 114043888-114043888 on Build GRCh38
rs776353088 Chr.10: 114044285-114044285 on Build GRCh38
rs555170500 Chr.10: 114043923-114043923 on Build GRCh38
rs35230616 Chr.10: 114044224-114044224 on Build GRCh38
rs770272324 Chr.10: 114044268-114044268 on Build GRCh38
rs921413698 Chr.10: 114044028-114044028 on Build GRCh38
rs763615545 Chr.10: 114044305-114044305 on Build GRCh38
rs980663243 Chr.10: 114043927-114043927 on Build GRCh38
rs957978969 Chr.10: 114044207-114044207 on Build GRCh38
rs34947712 Chr.10: 114044258-114044258 on Build GRCh38
rs573648222 Chr.10: 114043954-114043954 on Build GRCh38
rs913532543 Chr.10: 114044243-114044243 on Build GRCh38
rs781616595 Chr.10: 114044329-114044329 on Build GRCh38
rs746694631 Chr.10: 114044252-114044252 on Build GRCh38
rs1037704013 Chr.10: 114044306-114044306 on Build GRCh38
rs140442073 Chr.10: 114044360-114044360 on Build GRCh38
rs976656253 Chr.10: 114044264-114044264 on Build GRCh38
rs746355133 Chr.10: 114044267-114044267 on Build GRCh38

View Full Product Details