Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114122324-114122874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

SNP Detail

SNP ID Chromosome Location
rs925177109 Chr.10: 114122767-114122767 on Build GRCh38
rs1027206016 Chr.10: 114122596-114122596 on Build GRCh38
rs555060531 Chr.10: 114122387-114122387 on Build GRCh38
rs999797427 Chr.10: 114122442-114122442 on Build GRCh38
rs916295730 Chr.10: 114122745-114122745 on Build GRCh38
rs1028334965 Chr.10: 114122529-114122529 on Build GRCh38
rs373517014 Chr.10: 114122588-114122588 on Build GRCh38
rs992923632 Chr.10: 114122714-114122714 on Build GRCh38
rs201794970 Chr.10: 114122539-114122539 on Build GRCh38
rs1011601465 Chr.10: 114122519-114122519 on Build GRCh38
rs1032132241 Chr.10: 114122465-114122465 on Build GRCh38
rs1046059882 Chr.10: 114122806-114122806 on Build GRCh38
rs912813285 Chr.10: 114122647-114122647 on Build GRCh38
rs556352465 Chr.10: 114122680-114122680 on Build GRCh38
rs78122524 Chr.10: 114122661-114122661 on Build GRCh38
rs894818950 Chr.10: 114122363-114122363 on Build GRCh38
rs936355733 Chr.10: 114122772-114122772 on Build GRCh38
rs931441002 Chr.10: 114122738-114122738 on Build GRCh38
rs913552321 Chr.10: 114122819-114122819 on Build GRCh38
rs986778188 Chr.10: 114122636-114122636 on Build GRCh38
rs543776594 Chr.10: 114122466-114122466 on Build GRCh38
rs899998010 Chr.10: 114122477-114122477 on Build GRCh38
rs1040792371 Chr.10: 114122845-114122845 on Build GRCh38
rs1052080276 Chr.10: 114122793-114122793 on Build GRCh38
rs1052152504 Chr.10: 114122369-114122369 on Build GRCh38
rs936299912 Chr.10: 114122367-114122367 on Build GRCh38
rs1007967028 Chr.10: 114122405-114122405 on Build GRCh38
rs575096538 Chr.10: 114122830-114122830 on Build GRCh38
rs1041634330 Chr.10: 114122473-114122473 on Build GRCh38
rs990588759 Chr.10: 114122740-114122740 on Build GRCh38
rs541794757 Chr.10: 114122713-114122713 on Build GRCh38
rs1013231170 Chr.10: 114122370-114122370 on Build GRCh38
rs954203509 Chr.10: 114122564-114122564 on Build GRCh38
rs943827114 Chr.10: 114122832-114122832 on Build GRCh38
rs997135578 Chr.10: 114122567-114122567 on Build GRCh38
rs576329792 Chr.10: 114122445-114122445 on Build GRCh38
rs140513403 Chr.10: 114122798-114122798 on Build GRCh38
rs892182965 Chr.10: 114122376-114122376 on Build GRCh38
rs980657282 Chr.10: 114122764-114122764 on Build GRCh38
rs778303388 Chr.10: 114122375-114122375 on Build GRCh38
rs543160461 Chr.10: 114122394-114122394 on Build GRCh38
rs902395751 Chr.10: 114122420-114122420 on Build GRCh38

View Full Product Details