Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114124237-114124741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

SNP Detail

SNP ID Chromosome Location
rs78598149 Chr.10: 114124567-114124567 on Build GRCh38
rs888460877 Chr.10: 114124311-114124311 on Build GRCh38
rs971916652 Chr.10: 114124549-114124549 on Build GRCh38
rs901008157 Chr.10: 114124351-114124351 on Build GRCh38
rs919077430 Chr.10: 114124560-114124560 on Build GRCh38
rs528634127 Chr.10: 114124593-114124593 on Build GRCh38
rs913206786 Chr.10: 114124305-114124305 on Build GRCh38
rs557451192 Chr.10: 114124341-114124341 on Build GRCh38
rs529184013 Chr.10: 114124504-114124504 on Build GRCh38
rs923318827 Chr.10: 114124612-114124612 on Build GRCh38
rs941293549 Chr.10: 114124702-114124702 on Build GRCh38
rs758036202 Chr.10: 114124364-114124364 on Build GRCh38
rs889619337 Chr.10: 114124670-114124670 on Build GRCh38
rs565951392 Chr.10: 114124581-114124581 on Build GRCh38
rs976211072 Chr.10: 114124594-114124594 on Build GRCh38
rs1012635547 Chr.10: 114124317-114124317 on Build GRCh38
rs892654265 Chr.10: 114124654-114124654 on Build GRCh38
rs570938579 Chr.10: 114124592-114124592 on Build GRCh38
rs532998803 Chr.10: 114124580-114124580 on Build GRCh38
rs150856333 Chr.10: 114124362-114124362 on Build GRCh38
rs993942239 Chr.10: 114124439-114124439 on Build GRCh38
rs185517957 Chr.10: 114124508-114124508 on Build GRCh38
rs539825479 Chr.10: 114124367-114124367 on Build GRCh38
rs1018899141 Chr.10: 114124521-114124521 on Build GRCh38
rs1024437657 Chr.10: 114124443-114124443 on Build GRCh38
rs906824115 Chr.10: 114124610-114124610 on Build GRCh38
rs1005453550 Chr.10: 114124384-114124384 on Build GRCh38
rs953076951 Chr.10: 114124430-114124430 on Build GRCh38
rs188183531 Chr.10: 114124282-114124282 on Build GRCh38
rs1049611370 Chr.10: 114124647-114124647 on Build GRCh38
rs1045464061 Chr.10: 114124606-114124606 on Build GRCh38
rs945999277 Chr.10: 114124589-114124589 on Build GRCh38
rs145175627 Chr.10: 114124309-114124310 on Build GRCh38
rs896808516 Chr.10: 114124401-114124402 on Build GRCh38
rs756626268 Chr.10: 114124637-114124637 on Build GRCh38
rs575755846 Chr.10: 114124272-114124272 on Build GRCh38
rs778015183 Chr.10: 114124690-114124690 on Build GRCh38
rs906743701 Chr.10: 114124345-114124345 on Build GRCh38
rs966356907 Chr.10: 114124535-114124535 on Build GRCh38
rs971608151 Chr.10: 114124454-114124454 on Build GRCh38
rs1003747074 Chr.10: 114124354-114124354 on Build GRCh38
rs1048453480 Chr.10: 114124295-114124295 on Build GRCh38
rs35369963 Chr.10: 114124283-114124284 on Build GRCh38
rs193162602 Chr.10: 114124432-114124432 on Build GRCh38
rs1010625004 Chr.10: 114124386-114124386 on Build GRCh38
rs936017347 Chr.10: 114124274-114124274 on Build GRCh38
rs116601974 Chr.10: 114124387-114124387 on Build GRCh38
rs1010991645 Chr.10: 114124716-114124716 on Build GRCh38
rs200274201 Chr.10: 114124310-114124311 on Build GRCh38
rs60191874 Chr.10: 114124313-114124314 on Build GRCh38
rs67841380 Chr.10: 114124314-114124315 on Build GRCh38
rs187937463 Chr.10: 114124624-114124624 on Build GRCh38
rs568466356 Chr.10: 114124630-114124630 on Build GRCh38

View Full Product Details