Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114124577-114125122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

SNP Detail

SNP ID Chromosome Location
rs1007267466 Chr.10: 114124741-114124741 on Build GRCh38
rs984587003 Chr.10: 114124858-114124858 on Build GRCh38
rs946922208 Chr.10: 114124994-114124994 on Build GRCh38
rs941293549 Chr.10: 114124702-114124702 on Build GRCh38
rs923318827 Chr.10: 114124612-114124612 on Build GRCh38
rs538817900 Chr.10: 114124720-114124720 on Build GRCh38
rs780274825 Chr.10: 114125092-114125092 on Build GRCh38
rs889619337 Chr.10: 114124670-114124670 on Build GRCh38
rs939344123 Chr.10: 114124934-114124934 on Build GRCh38
rs539984632 Chr.10: 114124787-114124787 on Build GRCh38
rs41293048 Chr.10: 114124768-114124768 on Build GRCh38
rs892654265 Chr.10: 114124654-114124654 on Build GRCh38
rs149993067 Chr.10: 114124905-114124905 on Build GRCh38
rs1024072095 Chr.10: 114124759-114124759 on Build GRCh38
rs929897841 Chr.10: 114125003-114125003 on Build GRCh38
rs1043872700 Chr.10: 114125028-114125028 on Build GRCh38
rs1020931859 Chr.10: 114124856-114124856 on Build GRCh38
rs918470553 Chr.10: 114124974-114124974 on Build GRCh38
rs910000216 Chr.10: 114124874-114124874 on Build GRCh38
rs980819561 Chr.10: 114124926-114124926 on Build GRCh38
rs536103225 Chr.10: 114124726-114124726 on Build GRCh38
rs892685680 Chr.10: 114124977-114124977 on Build GRCh38
rs201498373 Chr.10: 114125006-114125007 on Build GRCh38
rs906824115 Chr.10: 114124610-114124610 on Build GRCh38
rs1049611370 Chr.10: 114124647-114124647 on Build GRCh38
rs557384763 Chr.10: 114124727-114124727 on Build GRCh38
rs1045464061 Chr.10: 114124606-114124606 on Build GRCh38
rs957500402 Chr.10: 114124829-114124829 on Build GRCh38
rs1038661456 Chr.10: 114124719-114124719 on Build GRCh38
rs139286873 Chr.10: 114124740-114124740 on Build GRCh38
rs994217292 Chr.10: 114124753-114124753 on Build GRCh38
rs1038311839 Chr.10: 114124969-114124969 on Build GRCh38
rs971985417 Chr.10: 114124770-114124770 on Build GRCh38
rs533287706 Chr.10: 114125081-114125081 on Build GRCh38
rs756626268 Chr.10: 114124637-114124637 on Build GRCh38
rs562662703 Chr.10: 114124965-114124965 on Build GRCh38
rs778015183 Chr.10: 114124690-114124690 on Build GRCh38
rs569403939 Chr.10: 114124739-114124739 on Build GRCh38
rs951860492 Chr.10: 114124823-114124823 on Build GRCh38
rs927968591 Chr.10: 114124930-114124930 on Build GRCh38
rs1026210096 Chr.10: 114124802-114124802 on Build GRCh38
rs1010991645 Chr.10: 114124716-114124716 on Build GRCh38
rs990363693 Chr.10: 114124839-114124840 on Build GRCh38
rs554897294 Chr.10: 114124791-114124791 on Build GRCh38
rs554341504 Chr.10: 114124847-114124847 on Build GRCh38
rs375433776 Chr.10: 114124764-114124764 on Build GRCh38
rs911067934 Chr.10: 114124949-114124949 on Build GRCh38
rs1001245663 Chr.10: 114125082-114125083 on Build GRCh38
rs573565808 Chr.10: 114124859-114124859 on Build GRCh38
rs187937463 Chr.10: 114124624-114124624 on Build GRCh38
rs773753016 Chr.10: 114124809-114124813 on Build GRCh38
rs976516088 Chr.10: 114124887-114124887 on Build GRCh38
rs568466356 Chr.10: 114124630-114124630 on Build GRCh38

View Full Product Details