Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6621249-6621750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

SNP Detail

SNP ID Chromosome Location
rs956097228 Chr.11: 6621705-6621705 on Build GRCh38
rs571950521 Chr.11: 6621600-6621600 on Build GRCh38
rs144267622 Chr.11: 6621530-6621532 on Build GRCh38
rs894603196 Chr.11: 6621510-6621510 on Build GRCh38
rs1040107618 Chr.11: 6621332-6621332 on Build GRCh38
rs954200375 Chr.11: 6621502-6621502 on Build GRCh38
rs1031498519 Chr.11: 6621457-6621457 on Build GRCh38
rs796945257 Chr.11: 6621447-6621447 on Build GRCh38
rs574642256 Chr.11: 6621429-6621429 on Build GRCh38
rs570787200 Chr.11: 6621458-6621458 on Build GRCh38
rs914200765 Chr.11: 6621715-6621715 on Build GRCh38
rs574619994 Chr.11: 6621683-6621683 on Build GRCh38
rs181411626 Chr.11: 6621425-6621425 on Build GRCh38
rs563523926 Chr.11: 6621731-6621731 on Build GRCh38
rs1025571486 Chr.11: 6621618-6621618 on Build GRCh38
rs747427764 Chr.11: 6621593-6621593 on Build GRCh38
rs763598131 Chr.11: 6621649-6621649 on Build GRCh38
rs530647625 Chr.11: 6621729-6621729 on Build GRCh38
rs764105625 Chr.11: 6621648-6621648 on Build GRCh38
rs892815390 Chr.11: 6621397-6621397 on Build GRCh38
rs1048704583 Chr.11: 6621380-6621381 on Build GRCh38
rs528543158 Chr.11: 6621362-6621362 on Build GRCh38
rs77307773 Chr.11: 6621604-6621604 on Build GRCh38
rs1016184096 Chr.11: 6621571-6621571 on Build GRCh38
rs752029422 Chr.11: 6621723-6621723 on Build GRCh38
rs775073456 Chr.11: 6621608-6621608 on Build GRCh38
rs897582726 Chr.11: 6621346-6621346 on Build GRCh38
rs1012537218 Chr.11: 6621673-6621673 on Build GRCh38
rs1050801029 Chr.11: 6621306-6621306 on Build GRCh38
rs543645531 Chr.11: 6621512-6621512 on Build GRCh38
rs755066856 Chr.11: 6621336-6621336 on Build GRCh38
rs1034394501 Chr.11: 6621470-6621470 on Build GRCh38
rs1054150316 Chr.11: 6621367-6621367 on Build GRCh38
rs762136959 Chr.11: 6621678-6621678 on Build GRCh38
rs185406127 Chr.11: 6621328-6621328 on Build GRCh38
rs762654594 Chr.11: 6621632-6621632 on Build GRCh38
rs887467552 Chr.11: 6621381-6621386 on Build GRCh38
rs1050036001 Chr.11: 6621307-6621307 on Build GRCh38
rs868236455 Chr.11: 6621527-6621529 on Build GRCh38
rs910968333 Chr.11: 6621675-6621675 on Build GRCh38
rs554384031 Chr.11: 6621680-6621680 on Build GRCh38
rs891272800 Chr.11: 6621315-6621315 on Build GRCh38
rs776815179 Chr.11: 6621659-6621659 on Build GRCh38
rs962071343 Chr.11: 6621588-6621588 on Build GRCh38
rs759315005 Chr.11: 6621567-6621567 on Build GRCh38
rs766957948 Chr.11: 6621711-6621711 on Build GRCh38
rs1044255646 Chr.11: 6621677-6621677 on Build GRCh38
rs189069517 Chr.11: 6621360-6621360 on Build GRCh38
rs750879934 Chr.11: 6621694-6621694 on Build GRCh38
rs956148060 Chr.11: 6621463-6621463 on Build GRCh38
rs369163933 Chr.11: 6621448-6621448 on Build GRCh38
rs61735329 Chr.11: 6621514-6621514 on Build GRCh38
rs777150562 Chr.11: 6621730-6621730 on Build GRCh38
rs532480223 Chr.11: 6621313-6621313 on Build GRCh38
rs1022853381 Chr.11: 6621436-6621436 on Build GRCh38
rs1003029994 Chr.11: 6621465-6621465 on Build GRCh38
rs932901735 Chr.11: 6621273-6621273 on Build GRCh38
rs758656034 Chr.11: 6621707-6621707 on Build GRCh38
rs61875879 Chr.11: 6621451-6621451 on Build GRCh38
rs548394513 Chr.11: 6621402-6621402 on Build GRCh38
rs778975451 Chr.11: 6621351-6621351 on Build GRCh38
rs770502784 Chr.11: 6621505-6621505 on Build GRCh38
rs569203772 Chr.11: 6621544-6621544 on Build GRCh38
rs539790233 Chr.11: 6621481-6621481 on Build GRCh38
rs1037195893 Chr.11: 6621558-6621558 on Build GRCh38
rs907330763 Chr.11: 6621464-6621464 on Build GRCh38
rs1031486706 Chr.11: 6621704-6621704 on Build GRCh38

View Full Product Details