Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7672373-7672879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R2
Entrez Gene ID
51700
Gene Name
cytochrome b5 reductase 2
Gene Aliases
B5R.2
UniGene
Hs.414362
Gene Chromosome Location
Chr.11: 7665095-7678568 on Build GRCh38
Transcripts
NM_001302826.1 NM_001302827.1 NM_016229.4 NR_126508.1 XM_005252975.4 XM_006718251.2 XM_011520184.2 XM_011520185.2 XM_017017920.1 XM_017017921.1 XM_017017922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136818 c.76G>C p.E26Q Chr.11: 7672750-7672750 on Build GRCh38
7275359 c.151+1G>C p.? Chr.11: 7672450-7672450 on Build GRCh38
4200982 c.125C>T p.P42L Chr.11: 7672477-7672477 on Build GRCh38
1628230 c.38A>T p.D13V Chr.11: 7672788-7672788 on Build GRCh38
4617219 c.150A>T p.V50V Chr.11: 7672452-7672452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540755199 Chr.11: 7672606-7672606 on Build GRCh38
rs770775296 Chr.11: 7672400-7672400 on Build GRCh38
rs371895643 Chr.11: 7672846-7672846 on Build GRCh38
rs766444251 Chr.11: 7672757-7672757 on Build GRCh38
rs985455757 Chr.11: 7672546-7672546 on Build GRCh38
rs762743838 Chr.11: 7672849-7672849 on Build GRCh38
rs990265937 Chr.11: 7672529-7672533 on Build GRCh38
rs377368796 Chr.11: 7672602-7672602 on Build GRCh38
rs778000025 Chr.11: 7672824-7672824 on Build GRCh38
rs568822682 Chr.11: 7672623-7672626 on Build GRCh38
rs765329574 Chr.11: 7672539-7672540 on Build GRCh38
rs183671644 Chr.11: 7672586-7672586 on Build GRCh38
rs1003484554 Chr.11: 7672489-7672489 on Build GRCh38
rs927351967 Chr.11: 7672415-7672415 on Build GRCh38
rs201145173 Chr.11: 7672477-7672477 on Build GRCh38
rs971634024 Chr.11: 7672613-7672613 on Build GRCh38
rs200550068 Chr.11: 7672758-7672758 on Build GRCh38
rs765982299 Chr.11: 7672856-7672856 on Build GRCh38
rs190479584 Chr.11: 7672649-7672649 on Build GRCh38
rs551869923 Chr.11: 7672510-7672510 on Build GRCh38
rs367743285 Chr.11: 7672830-7672830 on Build GRCh38
rs775828749 Chr.11: 7672420-7672420 on Build GRCh38
rs554088885 Chr.11: 7672625-7672626 on Build GRCh38
rs185210779 Chr.11: 7672843-7672843 on Build GRCh38
rs367691614 Chr.11: 7672714-7672714 on Build GRCh38
rs565308821 Chr.11: 7672524-7672525 on Build GRCh38
rs760259338 Chr.11: 7672534-7672534 on Build GRCh38
rs373643933 Chr.11: 7672788-7672788 on Build GRCh38
rs529321266 Chr.11: 7672674-7672674 on Build GRCh38
rs572529720 Chr.11: 7672479-7672479 on Build GRCh38
rs958694747 Chr.11: 7672818-7672818 on Build GRCh38
rs764260138 Chr.11: 7672437-7672437 on Build GRCh38
rs756239047 Chr.11: 7672816-7672816 on Build GRCh38
rs371724541 Chr.11: 7672711-7672711 on Build GRCh38
rs200124380 Chr.11: 7672715-7672715 on Build GRCh38
rs763673733 Chr.11: 7672539-7672539 on Build GRCh38
rs773600046 Chr.11: 7672691-7672691 on Build GRCh38
rs376133963 Chr.11: 7672441-7672441 on Build GRCh38
rs761031257 Chr.11: 7672426-7672426 on Build GRCh38
rs556333364 Chr.11: 7672626-7672627 on Build GRCh38
rs752549050 Chr.11: 7672807-7672807 on Build GRCh38
rs773501192 Chr.11: 7672404-7672404 on Build GRCh38
rs758730472 Chr.11: 7672450-7672450 on Build GRCh38
rs139087887 Chr.11: 7672575-7672575 on Build GRCh38
rs11041525 Chr.11: 7672782-7672782 on Build GRCh38
rs752331164 Chr.11: 7672455-7672455 on Build GRCh38
rs113660947 Chr.11: 7672799-7672799 on Build GRCh38
rs374021416 Chr.11: 7672844-7672844 on Build GRCh38
rs944576947 Chr.11: 7672401-7672401 on Build GRCh38
rs750751977 Chr.11: 7672444-7672444 on Build GRCh38
rs374508689 Chr.11: 7672499-7672499 on Build GRCh38
rs754699921 Chr.11: 7672760-7672760 on Build GRCh38
rs985271586 Chr.11: 7672646-7672646 on Build GRCh38
rs768875775 Chr.11: 7672703-7672703 on Build GRCh38
rs57207415 Chr.11: 7672623-7672624 on Build GRCh38
rs919115303 Chr.11: 7672623-7672634 on Build GRCh38
rs778537314 Chr.11: 7672564-7672564 on Build GRCh38
rs575405160 Chr.11: 7672505-7672505 on Build GRCh38
rs749961856 Chr.11: 7672736-7672736 on Build GRCh38
rs989033294 Chr.11: 7672592-7672592 on Build GRCh38
rs371538362 Chr.11: 7672838-7672838 on Build GRCh38
rs144823333 Chr.11: 7672622-7672623 on Build GRCh38
rs752366768 Chr.11: 7672706-7672707 on Build GRCh38
rs761821782 Chr.11: 7672498-7672498 on Build GRCh38
rs981383721 Chr.11: 7672631-7672631 on Build GRCh38
rs369595286 Chr.11: 7672754-7672754 on Build GRCh38
rs749548535 Chr.11: 7672826-7672826 on Build GRCh38
rs753344409 Chr.11: 7672541-7672541 on Build GRCh38
rs779931158 Chr.11: 7672570-7672570 on Build GRCh38
rs753413559 Chr.11: 7672722-7672722 on Build GRCh38
rs367986020 Chr.11: 7672509-7672509 on Build GRCh38
rs907447269 Chr.11: 7672480-7672480 on Build GRCh38
rs1036230122 Chr.11: 7672689-7672689 on Build GRCh38
rs760622631 Chr.11: 7672410-7672423 on Build GRCh38
rs887614105 Chr.11: 7672453-7672453 on Build GRCh38
rs748313980 Chr.11: 7672764-7672764 on Build GRCh38
rs151216178 Chr.11: 7672458-7672458 on Build GRCh38
rs779026569 Chr.11: 7672829-7672829 on Build GRCh38
rs193208545 Chr.11: 7672797-7672797 on Build GRCh38
rs927299567 Chr.11: 7672636-7672636 on Build GRCh38
rs750376937 Chr.11: 7672566-7672566 on Build GRCh38
rs549356450 Chr.11: 7672681-7672681 on Build GRCh38
rs577894211 Chr.11: 7672604-7672604 on Build GRCh38
rs535051265 Chr.11: 7672496-7672496 on Build GRCh38
rs774804725 Chr.11: 7672772-7672772 on Build GRCh38
rs371577976 Chr.11: 7672786-7672786 on Build GRCh38
rs767148574 Chr.11: 7672805-7672805 on Build GRCh38
rs539181818 Chr.11: 7672467-7672467 on Build GRCh38
rs71059117 Chr.11: 7672642-7672643 on Build GRCh38
rs1003146820 Chr.11: 7672688-7672688 on Build GRCh38
rs766583278 Chr.11: 7672624-7672625 on Build GRCh38
rs763630271 Chr.11: 7672725-7672725 on Build GRCh38
rs766598641 Chr.11: 7672510-7672510 on Build GRCh38
rs756361351 Chr.11: 7672769-7672769 on Build GRCh38
rs367580620 Chr.11: 7672460-7672460 on Build GRCh38
rs944648666 Chr.11: 7672585-7672585 on Build GRCh38
rs555646323 Chr.11: 7672500-7672500 on Build GRCh38
rs368655867 Chr.11: 7672639-7672642 on Build GRCh38
rs748443710 Chr.11: 7672705-7672705 on Build GRCh38
rs774135673 Chr.11: 7672406-7672406 on Build GRCh38
rs142188175 Chr.11: 7672451-7672451 on Build GRCh38
rs147551491 Chr.11: 7672785-7672785 on Build GRCh38
rs745669414 Chr.11: 7672407-7672407 on Build GRCh38
rs937390947 Chr.11: 7672643-7672643 on Build GRCh38
rs371380902 Chr.11: 7672487-7672487 on Build GRCh38
rs370655207 Chr.11: 7672531-7672531 on Build GRCh38
rs552628590 Chr.11: 7672476-7672476 on Build GRCh38
rs774647327 Chr.11: 7672719-7672719 on Build GRCh38
rs767961875 Chr.11: 7672721-7672721 on Build GRCh38
rs888846180 Chr.11: 7672508-7672508 on Build GRCh38
rs772801957 Chr.11: 7672791-7672791 on Build GRCh38
rs141959668 Chr.11: 7672801-7672801 on Build GRCh38
rs761588515 Chr.11: 7672735-7672735 on Build GRCh38
rs766723853 Chr.11: 7672513-7672513 on Build GRCh38
rs756688143 Chr.11: 7672550-7672550 on Build GRCh38
rs754945035 Chr.11: 7672528-7672529 on Build GRCh38
rs777770734 Chr.11: 7672770-7672770 on Build GRCh38
rs775225523 Chr.11: 7672641-7672642 on Build GRCh38
rs781097966 Chr.11: 7672763-7672763 on Build GRCh38
rs61729556 Chr.11: 7672771-7672771 on Build GRCh38
rs772639433 Chr.11: 7672837-7672837 on Build GRCh38
rs563198235 Chr.11: 7672704-7672704 on Build GRCh38
rs888793713 Chr.11: 7672746-7672746 on Build GRCh38
rs909335254 Chr.11: 7672671-7672671 on Build GRCh38
rs758478543 Chr.11: 7672569-7672569 on Build GRCh38
rs751923969 Chr.11: 7672521-7672521 on Build GRCh38
rs796923238 Chr.11: 7672641-7672642 on Build GRCh38
rs772376001 Chr.11: 7672418-7672418 on Build GRCh38
rs1040200495 Chr.11: 7672410-7672410 on Build GRCh38
rs769749812 Chr.11: 7672707-7672707 on Build GRCh38
rs772968293 Chr.11: 7672847-7672847 on Build GRCh38
rs758225455 Chr.11: 7672754-7672755 on Build GRCh38
rs778743032 Chr.11: 7672470-7672470 on Build GRCh38
rs777242160 Chr.11: 7672843-7672844 on Build GRCh38
rs757301907 Chr.11: 7672827-7672827 on Build GRCh38

View Full Product Details