Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8912344-8912840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIP1
Entrez Gene ID
56672
Gene Name
A-kinase interacting protein 1
Gene Aliases
BCA3, C11orf17
UniGene
Hs.131180
Gene Chromosome Location
Chr.11: 8911117-8920079 on Build GRCh38
Transcripts
NM_001206646.1 NM_001206647.1 NM_001206648.1 NM_020642.3 NR_045417.1 NR_045418.1 XM_017018011.1 XM_017018012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2222438 c.226G>C p.E76Q Chr.11: 8912456-8912456 on Build GRCh38
5905830 c.244C>T p.L82F Chr.11: 8912474-8912474 on Build GRCh38
4850903 c.224G>C p.R75T Chr.11: 8912454-8912454 on Build GRCh38
6284308 c.246T>C p.L82L Chr.11: 8912476-8912476 on Build GRCh38
4836328 c.260G>C p.R87T Chr.11: 8912490-8912490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887021168 Chr.11: 8912642-8912642 on Build GRCh38
rs771242599 Chr.11: 8912457-8912458 on Build GRCh38
rs776374830 Chr.11: 8912422-8912422 on Build GRCh38
rs1021688766 Chr.11: 8912711-8912711 on Build GRCh38
rs868535266 Chr.11: 8912686-8912686 on Build GRCh38
rs191305501 Chr.11: 8912633-8912633 on Build GRCh38
rs1022128787 Chr.11: 8912413-8912413 on Build GRCh38
rs574047902 Chr.11: 8912639-8912639 on Build GRCh38
rs1034141598 Chr.11: 8912637-8912637 on Build GRCh38
rs977843889 Chr.11: 8912449-8912449 on Build GRCh38
rs11042122 Chr.11: 8912742-8912742 on Build GRCh38
rs577782364 Chr.11: 8912604-8912604 on Build GRCh38
rs533291247 Chr.11: 8912808-8912808 on Build GRCh38
rs146611149 Chr.11: 8912531-8912531 on Build GRCh38
rs751684945 Chr.11: 8912450-8912450 on Build GRCh38
rs371556773 Chr.11: 8912566-8912566 on Build GRCh38
rs564354136 Chr.11: 8912795-8912795 on Build GRCh38
rs759742435 Chr.11: 8912554-8912558 on Build GRCh38
rs200956210 Chr.11: 8912579-8912579 on Build GRCh38
rs992518228 Chr.11: 8912649-8912649 on Build GRCh38
rs773071562 Chr.11: 8912572-8912572 on Build GRCh38
rs375674336 Chr.11: 8912575-8912575 on Build GRCh38
rs909712065 Chr.11: 8912692-8912707 on Build GRCh38
rs758649557 Chr.11: 8912437-8912437 on Build GRCh38
rs761641136 Chr.11: 8912423-8912423 on Build GRCh38
rs766313147 Chr.11: 8912446-8912446 on Build GRCh38
rs1054274823 Chr.11: 8912804-8912805 on Build GRCh38
rs555057623 Chr.11: 8912466-8912466 on Build GRCh38
rs922050614 Chr.11: 8912805-8912806 on Build GRCh38
rs1040944507 Chr.11: 8912662-8912662 on Build GRCh38
rs774820159 Chr.11: 8912536-8912536 on Build GRCh38
rs943999395 Chr.11: 8912648-8912648 on Build GRCh38
rs2742518 Chr.11: 8912554-8912554 on Build GRCh38
rs562700967 Chr.11: 8912782-8912782 on Build GRCh38
rs373583406 Chr.11: 8912549-8912549 on Build GRCh38
rs745449201 Chr.11: 8912532-8912532 on Build GRCh38
rs996905161 Chr.11: 8912409-8912409 on Build GRCh38
rs763038449 Chr.11: 8912573-8912573 on Build GRCh38
rs931962760 Chr.11: 8912805-8912805 on Build GRCh38
rs998853831 Chr.11: 8912726-8912726 on Build GRCh38
rs771775222 Chr.11: 8912534-8912534 on Build GRCh38
rs904629258 Chr.11: 8912722-8912722 on Build GRCh38
rs778625273 Chr.11: 8912517-8912517 on Build GRCh38
rs1051689159 Chr.11: 8912811-8912811 on Build GRCh38
rs1031710502 Chr.11: 8912789-8912789 on Build GRCh38
rs755501774 Chr.11: 8912459-8912459 on Build GRCh38
rs769981533 Chr.11: 8912560-8912567 on Build GRCh38
rs750676735 Chr.11: 8912433-8912433 on Build GRCh38
rs776998697 Chr.11: 8912531-8912531 on Build GRCh38
rs997098603 Chr.11: 8912707-8912707 on Build GRCh38
rs899643504 Chr.11: 8912685-8912685 on Build GRCh38
rs996169496 Chr.11: 8912381-8912381 on Build GRCh38
rs531743879 Chr.11: 8912791-8912791 on Build GRCh38
rs756608322 Chr.11: 8912497-8912497 on Build GRCh38
rs924839667 Chr.11: 8912463-8912463 on Build GRCh38
rs543149255 Chr.11: 8912720-8912720 on Build GRCh38
rs890394096 Chr.11: 8912430-8912430 on Build GRCh38
rs368557112 Chr.11: 8912429-8912429 on Build GRCh38
rs570373592 Chr.11: 8912815-8912815 on Build GRCh38
rs371516259 Chr.11: 8912506-8912506 on Build GRCh38
rs775623992 Chr.11: 8912749-8912749 on Build GRCh38
rs768554484 Chr.11: 8912553-8912553 on Build GRCh38
rs183602255 Chr.11: 8912794-8912794 on Build GRCh38
rs769510615 Chr.11: 8912568-8912568 on Build GRCh38
rs537025239 Chr.11: 8912451-8912454 on Build GRCh38
rs540412914 Chr.11: 8912607-8912607 on Build GRCh38
rs748504704 Chr.11: 8912483-8912483 on Build GRCh38

View Full Product Details