Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8919995-8920494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIP1
Entrez Gene ID
56672
Gene Name
A-kinase interacting protein 1
Gene Aliases
BCA3, C11orf17
UniGene
Hs.131180
Gene Chromosome Location
Chr.11: 8911117-8920079 on Build GRCh38
Transcripts
NM_001206646.1 NM_001206647.1 NM_001206648.1 NM_020642.3 NR_045417.1 NR_045418.1 XM_017018011.1 XM_017018012.1 NM_020643.2 XM_017018013.1

Gene Symbol
C11orf16
Entrez Gene ID
56673
Gene Name
chromosome 11 open reading frame 16
Gene Aliases
-
UniGene
Hs.277349
Gene Chromosome Location
Chr.11: 8920076-8933006 on Build GRCh38
Transcripts
NM_001206646.1 NM_001206647.1 NM_001206648.1 NM_020642.3 NR_045417.1 NR_045418.1 XM_017018011.1 XM_017018012.1 NM_020643.2 XM_017018013.1

SNP Detail

SNP ID Chromosome Location
rs75188962 Chr.11: 8920120-8920120 on Build GRCh38
rs897809948 Chr.11: 8920269-8920269 on Build GRCh38
rs553673408 Chr.11: 8920413-8920413 on Build GRCh38
rs1036766310 Chr.11: 8920384-8920384 on Build GRCh38
rs1037203575 Chr.11: 8920051-8920051 on Build GRCh38
rs570920391 Chr.11: 8920342-8920343 on Build GRCh38
rs773928204 Chr.11: 8920410-8920410 on Build GRCh38
rs1000079087 Chr.11: 8920464-8920464 on Build GRCh38
rs946919890 Chr.11: 8920422-8920422 on Build GRCh38
rs372597728 Chr.11: 8920324-8920324 on Build GRCh38
rs889293803 Chr.11: 8920214-8920214 on Build GRCh38
rs534060664 Chr.11: 8920322-8920322 on Build GRCh38
rs949798193 Chr.11: 8920171-8920171 on Build GRCh38
rs997873229 Chr.11: 8920081-8920081 on Build GRCh38
rs953976062 Chr.11: 8920121-8920121 on Build GRCh38
rs944915330 Chr.11: 8920446-8920446 on Build GRCh38
rs993473259 Chr.11: 8920287-8920287 on Build GRCh38
rs917339186 Chr.11: 8920255-8920255 on Build GRCh38
rs777636746 Chr.11: 8920201-8920201 on Build GRCh38
rs1008992823 Chr.11: 8920032-8920032 on Build GRCh38
rs538805626 Chr.11: 8920189-8920189 on Build GRCh38
rs930858993 Chr.11: 8920321-8920321 on Build GRCh38
rs891601377 Chr.11: 8920469-8920469 on Build GRCh38
rs115543642 Chr.11: 8920419-8920419 on Build GRCh38
rs373267880 Chr.11: 8920468-8920468 on Build GRCh38
rs1050740024 Chr.11: 8920424-8920424 on Build GRCh38
rs900814127 Chr.11: 8920059-8920059 on Build GRCh38
rs900945303 Chr.11: 8920418-8920418 on Build GRCh38
rs955810486 Chr.11: 8920402-8920402 on Build GRCh38
rs924389765 Chr.11: 8920358-8920358 on Build GRCh38
rs796268947 Chr.11: 8920133-8920133 on Build GRCh38
rs376490379 Chr.11: 8920266-8920266 on Build GRCh38
rs934358398 Chr.11: 8920191-8920191 on Build GRCh38
rs760516714 Chr.11: 8920449-8920449 on Build GRCh38
rs746715459 Chr.11: 8920396-8920396 on Build GRCh38
rs962070893 Chr.11: 8920085-8920085 on Build GRCh38
rs145379760 Chr.11: 8920312-8920312 on Build GRCh38
rs757770198 Chr.11: 8920436-8920437 on Build GRCh38
rs980107930 Chr.11: 8920274-8920274 on Build GRCh38
rs956074131 Chr.11: 8920200-8920200 on Build GRCh38
rs1049459288 Chr.11: 8920425-8920425 on Build GRCh38
rs534345395 Chr.11: 8920443-8920443 on Build GRCh38
rs940875415 Chr.11: 8920342-8920342 on Build GRCh38
rs947002760 Chr.11: 8920209-8920209 on Build GRCh38
rs1050997484 Chr.11: 8920211-8920211 on Build GRCh38
rs137922329 Chr.11: 8920159-8920160 on Build GRCh38
rs774162186 Chr.11: 8920310-8920310 on Build GRCh38
rs542397861 Chr.11: 8920436-8920436 on Build GRCh38
rs971287587 Chr.11: 8920327-8920327 on Build GRCh38
rs1009395577 Chr.11: 8920245-8920245 on Build GRCh38
rs1027596345 Chr.11: 8920107-8920108 on Build GRCh38
rs199812931 Chr.11: 8920161-8920161 on Build GRCh38
rs1032807079 Chr.11: 8920195-8920195 on Build GRCh38
rs534642688 Chr.11: 8920094-8920094 on Build GRCh38
rs780248298 Chr.11: 8920103-8920103 on Build GRCh38
rs915751239 Chr.11: 8920142-8920142 on Build GRCh38

View Full Product Details