Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11292900-11293408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT18
Entrez Gene ID
374378
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
UniGene
Hs.655152
Gene Chromosome Location
Chr.11: 11270874-11622045 on Build GRCh38
Transcripts
NM_198516.2 XM_006718224.2 XM_011520068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627790 c.1513-8G>A p.? Chr.11: 11293201-11293201 on Build GRCh38
269225 c.1650G>A p.R550R Chr.11: 11293056-11293056 on Build GRCh38
4428384 c.1656G>A p.K552K Chr.11: 11293050-11293050 on Build GRCh38
4017882 c.1516G>A p.V506M Chr.11: 11293190-11293190 on Build GRCh38
4424058 c.1665G>A p.W555* Chr.11: 11293041-11293041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762703454 Chr.11: 11293211-11293211 on Build GRCh38
rs1051732897 Chr.11: 11293280-11293280 on Build GRCh38
rs201537431 Chr.11: 11293230-11293230 on Build GRCh38
rs751588560 Chr.11: 11293091-11293091 on Build GRCh38
rs112598122 Chr.11: 11293131-11293131 on Build GRCh38
rs780816869 Chr.11: 11293125-11293127 on Build GRCh38
rs755509275 Chr.11: 11293096-11293096 on Build GRCh38
rs755705416 Chr.11: 11293211-11293212 on Build GRCh38
rs548962000 Chr.11: 11293140-11293140 on Build GRCh38
rs529247669 Chr.11: 11293139-11293139 on Build GRCh38
rs942518051 Chr.11: 11292950-11292950 on Build GRCh38
rs754105385 Chr.11: 11293183-11293183 on Build GRCh38
rs771187705 Chr.11: 11293215-11293215 on Build GRCh38
rs751676341 Chr.11: 11293167-11293167 on Build GRCh38
rs190037096 Chr.11: 11292990-11292990 on Build GRCh38
rs1032964380 Chr.11: 11293376-11293376 on Build GRCh38
rs779824658 Chr.11: 11293190-11293190 on Build GRCh38
rs768067191 Chr.11: 11293177-11293177 on Build GRCh38
rs200436772 Chr.11: 11293090-11293090 on Build GRCh38
rs148407434 Chr.11: 11293385-11293385 on Build GRCh38
rs999693567 Chr.11: 11292986-11292986 on Build GRCh38
rs753533517 Chr.11: 11292993-11292993 on Build GRCh38
rs761639425 Chr.11: 11293082-11293082 on Build GRCh38
rs573227011 Chr.11: 11293032-11293032 on Build GRCh38
rs773762313 Chr.11: 11292979-11292979 on Build GRCh38
rs773089871 Chr.11: 11293207-11293207 on Build GRCh38
rs374763409 Chr.11: 11293233-11293233 on Build GRCh38
rs386750685 Chr.11: 11293252-11293253 on Build GRCh38
rs765427511 Chr.11: 11293083-11293083 on Build GRCh38
rs769569442 Chr.11: 11293205-11293205 on Build GRCh38
rs182930467 Chr.11: 11293011-11293011 on Build GRCh38
rs925759620 Chr.11: 11293251-11293251 on Build GRCh38
rs568965427 Chr.11: 11292952-11292952 on Build GRCh38
rs369723200 Chr.11: 11293243-11293243 on Build GRCh38
rs774539122 Chr.11: 11293217-11293217 on Build GRCh38
rs113829057 Chr.11: 11293275-11293275 on Build GRCh38
rs1001743636 Chr.11: 11293299-11293299 on Build GRCh38
rs138326637 Chr.11: 11293252-11293252 on Build GRCh38
rs746479178 Chr.11: 11293121-11293121 on Build GRCh38
rs767399912 Chr.11: 11292982-11292982 on Build GRCh38
rs113918582 Chr.11: 11293128-11293128 on Build GRCh38
rs763988566 Chr.11: 11292992-11292992 on Build GRCh38
rs902793406 Chr.11: 11292972-11292972 on Build GRCh38
rs1021838041 Chr.11: 11293353-11293353 on Build GRCh38
rs372092468 Chr.11: 11293180-11293180 on Build GRCh38
rs150701186 Chr.11: 11293106-11293106 on Build GRCh38
rs578084309 Chr.11: 11293386-11293386 on Build GRCh38
rs10437598 Chr.11: 11292991-11292991 on Build GRCh38
rs376700751 Chr.11: 11293201-11293201 on Build GRCh38
rs887143673 Chr.11: 11293294-11293294 on Build GRCh38
rs139933398 Chr.11: 11293169-11293169 on Build GRCh38
rs758482976 Chr.11: 11293089-11293089 on Build GRCh38
rs150020084 Chr.11: 11293188-11293188 on Build GRCh38
rs914339591 Chr.11: 11293127-11293127 on Build GRCh38
rs781557639 Chr.11: 11293033-11293033 on Build GRCh38
rs754270512 Chr.11: 11293240-11293240 on Build GRCh38
rs747518549 Chr.11: 11293031-11293031 on Build GRCh38
rs199611116 Chr.11: 11293102-11293102 on Build GRCh38
rs201889204 Chr.11: 11293037-11293037 on Build GRCh38
rs1005067086 Chr.11: 11293287-11293287 on Build GRCh38
rs763258008 Chr.11: 11293143-11293143 on Build GRCh38
rs959777482 Chr.11: 11293307-11293307 on Build GRCh38
rs750010154 Chr.11: 11293210-11293210 on Build GRCh38
rs551200358 Chr.11: 11293191-11293191 on Build GRCh38
rs772824227 Chr.11: 11293141-11293141 on Build GRCh38
rs774696071 Chr.11: 11293126-11293128 on Build GRCh38
rs1025724833 Chr.11: 11293282-11293282 on Build GRCh38
rs35037993 Chr.11: 11293107-11293107 on Build GRCh38
rs112108154 Chr.11: 11293241-11293241 on Build GRCh38
rs898581004 Chr.11: 11293262-11293262 on Build GRCh38
rs17855075 Chr.11: 11293084-11293084 on Build GRCh38
rs781639055 Chr.11: 11293097-11293097 on Build GRCh38
rs766192930 Chr.11: 11293242-11293242 on Build GRCh38
rs759051779 Chr.11: 11292981-11292981 on Build GRCh38
rs764076553 Chr.11: 11293066-11293066 on Build GRCh38
rs76712184 Chr.11: 11292999-11292999 on Build GRCh38
rs1045614537 Chr.11: 11293259-11293259 on Build GRCh38
rs1034511859 Chr.11: 11293305-11293305 on Build GRCh38
rs759676464 Chr.11: 11293218-11293218 on Build GRCh38
rs947100986 Chr.11: 11292963-11292963 on Build GRCh38
rs368789058 Chr.11: 11293202-11293202 on Build GRCh38
rs964274529 Chr.11: 11293321-11293321 on Build GRCh38
rs544628048 Chr.11: 11293327-11293327 on Build GRCh38
rs968506260 Chr.11: 11293367-11293367 on Build GRCh38
rs576653157 Chr.11: 11292923-11292923 on Build GRCh38
rs745424620 Chr.11: 11293049-11293049 on Build GRCh38
rs185402461 Chr.11: 11293219-11293219 on Build GRCh38
rs187101191 Chr.11: 11292944-11292944 on Build GRCh38
rs993140205 Chr.11: 11293029-11293029 on Build GRCh38
rs112243920 Chr.11: 11293018-11293018 on Build GRCh38
rs539244744 Chr.11: 11292959-11292959 on Build GRCh38
rs148153149 Chr.11: 11293035-11293035 on Build GRCh38
rs758483374 Chr.11: 11293013-11293013 on Build GRCh38
rs748503160 Chr.11: 11293034-11293034 on Build GRCh38
rs1023746354 Chr.11: 11292994-11292994 on Build GRCh38
rs752665485 Chr.11: 11293223-11293223 on Build GRCh38
rs753239923 Chr.11: 11293179-11293179 on Build GRCh38
rs755750886 Chr.11: 11293308-11293308 on Build GRCh38
rs535419857 Chr.11: 11292947-11292947 on Build GRCh38
rs16909352 Chr.11: 11293071-11293071 on Build GRCh38
rs1030613232 Chr.11: 11292929-11292929 on Build GRCh38
rs1037938768 Chr.11: 11293290-11293291 on Build GRCh38
rs377654983 Chr.11: 11292938-11292938 on Build GRCh38
rs771611553 Chr.11: 11293059-11293059 on Build GRCh38
rs113081593 Chr.11: 11292951-11292951 on Build GRCh38
rs760111423 Chr.11: 11292966-11292966 on Build GRCh38
rs546593721 Chr.11: 11292927-11292927 on Build GRCh38
rs771795980 Chr.11: 11293116-11293116 on Build GRCh38
rs1008213568 Chr.11: 11293298-11293298 on Build GRCh38
rs750645578 Chr.11: 11293088-11293088 on Build GRCh38
rs767078208 Chr.11: 11293349-11293349 on Build GRCh38
rs760706102 Chr.11: 11293061-11293061 on Build GRCh38
rs886452088 Chr.11: 11293286-11293286 on Build GRCh38
rs770565303 Chr.11: 11292978-11292978 on Build GRCh38
rs992545379 Chr.11: 11293212-11293212 on Build GRCh38
rs568876193 Chr.11: 11293155-11293155 on Build GRCh38
rs757445547 Chr.11: 11293010-11293010 on Build GRCh38
rs555380948 Chr.11: 11293060-11293060 on Build GRCh38
rs561697544 Chr.11: 11293309-11293309 on Build GRCh38
rs149641477 Chr.11: 11293253-11293253 on Build GRCh38
rs950879412 Chr.11: 11292934-11292934 on Build GRCh38
rs779735158 Chr.11: 11293120-11293120 on Build GRCh38

View Full Product Details