Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11962612-11963132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK3
Entrez Gene ID
27122
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Gene Aliases
REIC, RIG
UniGene
Hs.292156
Gene Chromosome Location
Chr.11: 11962996-12009370 on Build GRCh38
Transcripts
NM_001018057.1 NM_013253.4 NM_015881.5 XM_006718178.2 XM_017017553.1 XM_017017554.1 XM_017017555.1

SNP Detail

SNP ID Chromosome Location
rs781680513 Chr.11: 11962802-11962802 on Build GRCh38
rs746334803 Chr.11: 11962889-11962889 on Build GRCh38
rs569198887 Chr.11: 11962819-11962819 on Build GRCh38
rs901133702 Chr.11: 11962811-11962811 on Build GRCh38
rs565023754 Chr.11: 11962808-11962808 on Build GRCh38
rs1041984286 Chr.11: 11962824-11962824 on Build GRCh38
rs1057228829 Chr.11: 11962728-11962728 on Build GRCh38
rs1021722789 Chr.11: 11962750-11962750 on Build GRCh38
rs775776075 Chr.11: 11962986-11962986 on Build GRCh38
rs1043638942 Chr.11: 11963015-11963015 on Build GRCh38
rs779049846 Chr.11: 11962845-11962846 on Build GRCh38
rs998213378 Chr.11: 11962957-11962957 on Build GRCh38
rs965280492 Chr.11: 11962780-11962780 on Build GRCh38
rs901176443 Chr.11: 11963041-11963041 on Build GRCh38
rs1034553801 Chr.11: 11962888-11962888 on Build GRCh38
rs904813139 Chr.11: 11962885-11962885 on Build GRCh38
rs567014829 Chr.11: 11963101-11963101 on Build GRCh38
rs531446736 Chr.11: 11962746-11962746 on Build GRCh38
rs947825697 Chr.11: 11962907-11962907 on Build GRCh38
rs757341844 Chr.11: 11962762-11962762 on Build GRCh38
rs1025495626 Chr.11: 11963020-11963020 on Build GRCh38
rs547387817 Chr.11: 11962980-11962980 on Build GRCh38
rs1000753289 Chr.11: 11962886-11962886 on Build GRCh38
rs770150974 Chr.11: 11962914-11962914 on Build GRCh38
rs185081957 Chr.11: 11962754-11962754 on Build GRCh38
rs998074419 Chr.11: 11963061-11963061 on Build GRCh38
rs952687659 Chr.11: 11963048-11963048 on Build GRCh38
rs1025585956 Chr.11: 11963100-11963100 on Build GRCh38
rs1048245415 Chr.11: 11962758-11962758 on Build GRCh38
rs972410044 Chr.11: 11963014-11963014 on Build GRCh38
rs927674002 Chr.11: 11962931-11962931 on Build GRCh38
rs909614922 Chr.11: 11962873-11962873 on Build GRCh38
rs1039724189 Chr.11: 11963040-11963040 on Build GRCh38
rs1006871934 Chr.11: 11962763-11962763 on Build GRCh38
rs918513798 Chr.11: 11962733-11962733 on Build GRCh38
rs891934020 Chr.11: 11962990-11962990 on Build GRCh38
rs143680578 Chr.11: 11962656-11962656 on Build GRCh38
rs946253305 Chr.11: 11963000-11963000 on Build GRCh38
rs942769163 Chr.11: 11962775-11962775 on Build GRCh38
rs972588408 Chr.11: 11962699-11962699 on Build GRCh38
rs1007418732 Chr.11: 11963036-11963036 on Build GRCh38
rs984254447 Chr.11: 11962862-11962862 on Build GRCh38
rs970386942 Chr.11: 11962668-11962668 on Build GRCh38
rs1025443568 Chr.11: 11962832-11962834 on Build GRCh38
rs1010700887 Chr.11: 11962749-11962749 on Build GRCh38

View Full Product Details