Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11965386-11965891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK3
Entrez Gene ID
27122
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Gene Aliases
REIC, RIG
UniGene
Hs.292156
Gene Chromosome Location
Chr.11: 11962996-12009370 on Build GRCh38
Transcripts
NM_001018057.1 NM_013253.4 NM_015881.5 XM_006718178.2 XM_017017553.1 XM_017017554.1 XM_017017555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371826 c.762G>A p.W254* Chr.11: 11965877-11965877 on Build GRCh38
5371825 c.830G>A p.S277N Chr.11: 11965809-11965809 on Build GRCh38
5771488 c.796C>G p.P266A Chr.11: 11965843-11965843 on Build GRCh38
7163380 c.824C>T p.P275L Chr.11: 11965815-11965815 on Build GRCh38
6644504 c.788A>G p.D263G Chr.11: 11965851-11965851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574670771 Chr.11: 11965517-11965517 on Build GRCh38
rs529362006 Chr.11: 11965622-11965622 on Build GRCh38
rs761319141 Chr.11: 11965858-11965858 on Build GRCh38
rs1035282663 Chr.11: 11965766-11965766 on Build GRCh38
rs147589185 Chr.11: 11965642-11965642 on Build GRCh38
rs973569550 Chr.11: 11965750-11965750 on Build GRCh38
rs770086823 Chr.11: 11965788-11965788 on Build GRCh38
rs144453521 Chr.11: 11965749-11965749 on Build GRCh38
rs1055315793 Chr.11: 11965535-11965535 on Build GRCh38
rs541318958 Chr.11: 11965508-11965508 on Build GRCh38
rs1035567953 Chr.11: 11965574-11965574 on Build GRCh38
rs563769256 Chr.11: 11965598-11965598 on Build GRCh38
rs748289346 Chr.11: 11965775-11965775 on Build GRCh38
rs962131161 Chr.11: 11965480-11965480 on Build GRCh38
rs753259246 Chr.11: 11965769-11965770 on Build GRCh38
rs969681527 Chr.11: 11965538-11965538 on Build GRCh38
rs1054294517 Chr.11: 11965851-11965851 on Build GRCh38
rs548486550 Chr.11: 11965722-11965722 on Build GRCh38
rs1021806601 Chr.11: 11965523-11965523 on Build GRCh38
rs776469436 Chr.11: 11965455-11965455 on Build GRCh38
rs879280284 Chr.11: 11965590-11965590 on Build GRCh38
rs79956160 Chr.11: 11965745-11965745 on Build GRCh38
rs963226959 Chr.11: 11965849-11965849 on Build GRCh38
rs770520501 Chr.11: 11965413-11965413 on Build GRCh38
rs770458041 Chr.11: 11965408-11965409 on Build GRCh38
rs142548699 Chr.11: 11965542-11965542 on Build GRCh38
rs987196795 Chr.11: 11965461-11965461 on Build GRCh38
rs772071952 Chr.11: 11965798-11965798 on Build GRCh38
rs1047028001 Chr.11: 11965566-11965566 on Build GRCh38
rs949669766 Chr.11: 11965776-11965776 on Build GRCh38
rs983361753 Chr.11: 11965772-11965772 on Build GRCh38
rs191282963 Chr.11: 11965636-11965636 on Build GRCh38
rs549189900 Chr.11: 11965754-11965754 on Build GRCh38
rs1015206324 Chr.11: 11965746-11965746 on Build GRCh38
rs558537974 Chr.11: 11965665-11965665 on Build GRCh38
rs1031135414 Chr.11: 11965497-11965497 on Build GRCh38
rs781300155 Chr.11: 11965773-11965773 on Build GRCh38
rs559484642 Chr.11: 11965679-11965679 on Build GRCh38
rs776306314 Chr.11: 11965641-11965641 on Build GRCh38
rs1021310644 Chr.11: 11965747-11965747 on Build GRCh38
rs745692803 Chr.11: 11965797-11965797 on Build GRCh38
rs775567196 Chr.11: 11965837-11965837 on Build GRCh38
rs931969330 Chr.11: 11965678-11965678 on Build GRCh38
rs959974960 Chr.11: 11965771-11965771 on Build GRCh38
rs776232698 Chr.11: 11965857-11965857 on Build GRCh38
rs890469545 Chr.11: 11965703-11965703 on Build GRCh38
rs187756923 Chr.11: 11965515-11965515 on Build GRCh38
rs759451056 Chr.11: 11965633-11965633 on Build GRCh38
rs893790425 Chr.11: 11965860-11965860 on Build GRCh38
rs949534647 Chr.11: 11965565-11965565 on Build GRCh38
rs142127034 Chr.11: 11965744-11965744 on Build GRCh38
rs1028798588 Chr.11: 11965422-11965422 on Build GRCh38
rs907599412 Chr.11: 11965474-11965474 on Build GRCh38
rs778524278 Chr.11: 11965794-11965794 on Build GRCh38
rs983779913 Chr.11: 11965631-11965631 on Build GRCh38
rs371266075 Chr.11: 11965748-11965748 on Build GRCh38
rs924109994 Chr.11: 11965830-11965830 on Build GRCh38
rs1053135630 Chr.11: 11965740-11965740 on Build GRCh38
rs182375599 Chr.11: 11965690-11965690 on Build GRCh38
rs973089612 Chr.11: 11965487-11965487 on Build GRCh38
rs768247684 Chr.11: 11965848-11965848 on Build GRCh38
rs890752105 Chr.11: 11965416-11965416 on Build GRCh38
rs1053756849 Chr.11: 11965671-11965671 on Build GRCh38
rs760755000 Chr.11: 11965843-11965843 on Build GRCh38
rs755035718 Chr.11: 11965768-11965768 on Build GRCh38
rs1001147373 Chr.11: 11965757-11965757 on Build GRCh38
rs572845392 Chr.11: 11965505-11965505 on Build GRCh38

View Full Product Details