Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14972079-14972585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCA
Entrez Gene ID
796
Gene Name
calcitonin related polypeptide alpha
Gene Aliases
CALC1, CGRP, CGRP-I, CGRP1, CT, KC, PCT
UniGene
Hs.37058
Gene Chromosome Location
Chr.11: 14966668-14972361 on Build GRCh38
Transcripts
NM_001033952.2 NM_001033953.2 NM_001741.2 NR_125898.1 XM_017018283.1 XM_017018284.1

SNP Detail

SNP ID Chromosome Location
rs957058884 Chr.11: 14972356-14972356 on Build GRCh38
rs201037927 Chr.11: 14972506-14972506 on Build GRCh38
rs13306225 Chr.11: 14972412-14972412 on Build GRCh38
rs782025106 Chr.11: 14972112-14972112 on Build GRCh38
rs1039132101 Chr.11: 14972215-14972215 on Build GRCh38
rs942003220 Chr.11: 14972214-14972214 on Build GRCh38
rs894045103 Chr.11: 14972562-14972562 on Build GRCh38
rs12361439 Chr.11: 14972345-14972345 on Build GRCh38
rs781894015 Chr.11: 14972457-14972457 on Build GRCh38
rs983341308 Chr.11: 14972429-14972429 on Build GRCh38
rs915655174 Chr.11: 14972116-14972116 on Build GRCh38
rs902656925 Chr.11: 14972307-14972307 on Build GRCh38
rs887783623 Chr.11: 14972204-14972204 on Build GRCh38
rs190849846 Chr.11: 14972555-14972555 on Build GRCh38
rs909127197 Chr.11: 14972431-14972431 on Build GRCh38
rs186815085 Chr.11: 14972492-14972492 on Build GRCh38
rs370033353 Chr.11: 14972278-14972279 on Build GRCh38
rs928479751 Chr.11: 14972409-14972409 on Build GRCh38
rs538005968 Chr.11: 14972198-14972198 on Build GRCh38
rs35943755 Chr.11: 14972149-14972149 on Build GRCh38
rs933416631 Chr.11: 14972469-14972469 on Build GRCh38
rs34692826 Chr.11: 14972358-14972358 on Build GRCh38
rs929230738 Chr.11: 14972185-14972185 on Build GRCh38
rs779347647 Chr.11: 14972194-14972194 on Build GRCh38
rs2644690 Chr.11: 14972230-14972230 on Build GRCh38
rs1030927124 Chr.11: 14972242-14972242 on Build GRCh38
rs781819731 Chr.11: 14972189-14972189 on Build GRCh38
rs558136997 Chr.11: 14972235-14972235 on Build GRCh38
rs372688687 Chr.11: 14972122-14972122 on Build GRCh38
rs999686304 Chr.11: 14972525-14972525 on Build GRCh38
rs902688383 Chr.11: 14972499-14972499 on Build GRCh38
rs1032560640 Chr.11: 14972336-14972336 on Build GRCh38
rs1022738311 Chr.11: 14972381-14972381 on Build GRCh38
rs35955129 Chr.11: 14972451-14972451 on Build GRCh38
rs34468174 Chr.11: 14972334-14972334 on Build GRCh38
rs551974973 Chr.11: 14972146-14972146 on Build GRCh38
rs782700663 Chr.11: 14972445-14972445 on Build GRCh38
rs948621410 Chr.11: 14972144-14972144 on Build GRCh38
rs981185089 Chr.11: 14972408-14972408 on Build GRCh38

View Full Product Details