Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19230340-19230878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F8
Entrez Gene ID
79733
Gene Name
E2F transcription factor 8
Gene Aliases
E2F-8
UniGene
Hs.523526
Gene Chromosome Location
Chr.11: 19224063-19241655 on Build GRCh38
Transcripts
NM_001256371.1 NM_001256372.1 NM_024680.3 XM_011520367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1317343 c.1087T>C p.F363L Chr.11: 19230814-19230814 on Build GRCh38
6651070 c.1233A>G p.I411M Chr.11: 19230668-19230668 on Build GRCh38
4679990 c.1157G>A p.R386H Chr.11: 19230744-19230744 on Build GRCh38
3808964 c.1254T>G p.P418P Chr.11: 19230647-19230647 on Build GRCh38
1353130 c.1177C>T p.R393* Chr.11: 19230724-19230724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778628199 Chr.11: 19230636-19230636 on Build GRCh38
rs774767215 Chr.11: 19230844-19230844 on Build GRCh38
rs1034631955 Chr.11: 19230778-19230778 on Build GRCh38
rs1034945298 Chr.11: 19230516-19230516 on Build GRCh38
rs758176781 Chr.11: 19230719-19230719 on Build GRCh38
rs767997311 Chr.11: 19230659-19230659 on Build GRCh38
rs977030113 Chr.11: 19230365-19230365 on Build GRCh38
rs756195991 Chr.11: 19230709-19230709 on Build GRCh38
rs752843481 Chr.11: 19230695-19230695 on Build GRCh38
rs914690289 Chr.11: 19230541-19230541 on Build GRCh38
rs568877282 Chr.11: 19230644-19230644 on Build GRCh38
rs748660242 Chr.11: 19230678-19230678 on Build GRCh38
rs779838635 Chr.11: 19230720-19230720 on Build GRCh38
rs147067430 Chr.11: 19230807-19230807 on Build GRCh38
rs763563360 Chr.11: 19230753-19230753 on Build GRCh38
rs150203629 Chr.11: 19230787-19230787 on Build GRCh38
rs375962806 Chr.11: 19230786-19230786 on Build GRCh38
rs745642528 Chr.11: 19230638-19230638 on Build GRCh38
rs561021375 Chr.11: 19230475-19230475 on Build GRCh38
rs578202327 Chr.11: 19230803-19230803 on Build GRCh38
rs949825734 Chr.11: 19230463-19230463 on Build GRCh38
rs768389466 Chr.11: 19230729-19230729 on Build GRCh38
rs991450595 Chr.11: 19230446-19230446 on Build GRCh38
rs142903161 Chr.11: 19230435-19230435 on Build GRCh38
rs760900942 Chr.11: 19230854-19230854 on Build GRCh38
rs772385728 Chr.11: 19230849-19230849 on Build GRCh38
rs767455024 Chr.11: 19230691-19230691 on Build GRCh38
rs754611426 Chr.11: 19230679-19230679 on Build GRCh38
rs528273261 Chr.11: 19230488-19230488 on Build GRCh38
rs187651949 Chr.11: 19230432-19230432 on Build GRCh38
rs749112429 Chr.11: 19230612-19230612 on Build GRCh38
rs3740966 Chr.11: 19230716-19230716 on Build GRCh38
rs369778521 Chr.11: 19230733-19230733 on Build GRCh38
rs967361665 Chr.11: 19230551-19230551 on Build GRCh38
rs145881683 Chr.11: 19230739-19230739 on Build GRCh38
rs771896495 Chr.11: 19230639-19230639 on Build GRCh38
rs898522012 Chr.11: 19230545-19230545 on Build GRCh38
rs761718323 Chr.11: 19230509-19230509 on Build GRCh38
rs1003008925 Chr.11: 19230489-19230489 on Build GRCh38
rs971316296 Chr.11: 19230789-19230789 on Build GRCh38
rs199676141 Chr.11: 19230744-19230744 on Build GRCh38
rs780774281 Chr.11: 19230731-19230731 on Build GRCh38
rs774809310 Chr.11: 19230766-19230766 on Build GRCh38
rs779789465 Chr.11: 19230808-19230808 on Build GRCh38
rs1050051918 Chr.11: 19230563-19230563 on Build GRCh38
rs902917996 Chr.11: 19230692-19230692 on Build GRCh38
rs372440321 Chr.11: 19230851-19230851 on Build GRCh38
rs749771997 Chr.11: 19230833-19230833 on Build GRCh38
rs77599073 Chr.11: 19230828-19230828 on Build GRCh38
rs542948535 Chr.11: 19230448-19230448 on Build GRCh38
rs564651808 Chr.11: 19230526-19230526 on Build GRCh38
rs138725272 Chr.11: 19230793-19230793 on Build GRCh38
rs763402043 Chr.11: 19230677-19230677 on Build GRCh38
rs535247090 Chr.11: 19230772-19230772 on Build GRCh38
rs773892838 Chr.11: 19230671-19230671 on Build GRCh38
rs762189735 Chr.11: 19230660-19230660 on Build GRCh38
rs1048687329 Chr.11: 19230414-19230414 on Build GRCh38
rs140933620 Chr.11: 19230746-19230746 on Build GRCh38
rs1020044456 Chr.11: 19230822-19230822 on Build GRCh38
rs751894195 Chr.11: 19230366-19230366 on Build GRCh38
rs59512436 Chr.11: 19230568-19230568 on Build GRCh38
rs1004466044 Chr.11: 19230742-19230742 on Build GRCh38
rs765797631 Chr.11: 19230670-19230670 on Build GRCh38
rs959116838 Chr.11: 19230785-19230785 on Build GRCh38
rs771587599 Chr.11: 19230837-19230837 on Build GRCh38
rs746578931 Chr.11: 19230722-19230722 on Build GRCh38
rs151170626 Chr.11: 19230522-19230522 on Build GRCh38
rs149364014 Chr.11: 19230806-19230806 on Build GRCh38
rs781342334 Chr.11: 19230601-19230601 on Build GRCh38
rs923840670 Chr.11: 19230562-19230562 on Build GRCh38
rs187243133 Chr.11: 19230831-19230831 on Build GRCh38
rs754995545 Chr.11: 19230380-19230380 on Build GRCh38
rs746845601 Chr.11: 19230812-19230812 on Build GRCh38
rs1040177542 Chr.11: 19230569-19230569 on Build GRCh38
rs754319955 Chr.11: 19230401-19230402 on Build GRCh38
rs1863382 Chr.11: 19230421-19230421 on Build GRCh38
rs138078778 Chr.11: 19230817-19230817 on Build GRCh38
rs1863381 Chr.11: 19230560-19230560 on Build GRCh38
rs370239272 Chr.11: 19230848-19230848 on Build GRCh38
rs146643516 Chr.11: 19230745-19230745 on Build GRCh38
rs1015127114 Chr.11: 19230468-19230468 on Build GRCh38
rs778793567 Chr.11: 19230713-19230713 on Build GRCh38
rs770931138 Chr.11: 19230628-19230628 on Build GRCh38
rs578119875 Chr.11: 19230363-19230363 on Build GRCh38
rs990217258 Chr.11: 19230539-19230539 on Build GRCh38
rs191569218 Chr.11: 19230757-19230757 on Build GRCh38
rs940104345 Chr.11: 19230524-19230524 on Build GRCh38

View Full Product Details