Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22624514-22625009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6302881 c.1001C>A p.A334E Chr.11: 22624810-22624810 on Build GRCh38
5020903 c.959C>T p.P320L Chr.11: 22624852-22624852 on Build GRCh38
6132214 c.1056A>T p.L352F Chr.11: 22624755-22624755 on Build GRCh38
5040879 c.946C>G p.Q316E Chr.11: 22624865-22624865 on Build GRCh38
272671 c.859T>C p.Y287H Chr.11: 22624952-22624952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201285915 Chr.11: 22624724-22624724 on Build GRCh38
rs371631248 Chr.11: 22624875-22624875 on Build GRCh38
rs764085506 Chr.11: 22624701-22624701 on Build GRCh38
rs745879868 Chr.11: 22624785-22624785 on Build GRCh38
rs915379462 Chr.11: 22624912-22624912 on Build GRCh38
rs587778343 Chr.11: 22624770-22624770 on Build GRCh38
rs753213642 Chr.11: 22624829-22624829 on Build GRCh38
rs544726894 Chr.11: 22624555-22624555 on Build GRCh38
rs771197883 Chr.11: 22624779-22624779 on Build GRCh38
rs781234202 Chr.11: 22624770-22624782 on Build GRCh38
rs768798175 Chr.11: 22624960-22624960 on Build GRCh38
rs199794268 Chr.11: 22624766-22624766 on Build GRCh38
rs375071905 Chr.11: 22624886-22624886 on Build GRCh38
rs970737747 Chr.11: 22624537-22624537 on Build GRCh38
rs772941983 Chr.11: 22624810-22624810 on Build GRCh38
rs750436220 Chr.11: 22624838-22624839 on Build GRCh38
rs959443639 Chr.11: 22624903-22624903 on Build GRCh38
rs1021038958 Chr.11: 22624972-22624972 on Build GRCh38
rs746120363 Chr.11: 22624859-22624859 on Build GRCh38
rs544124149 Chr.11: 22624682-22624682 on Build GRCh38
rs190980430 Chr.11: 22624948-22624948 on Build GRCh38
rs753723379 Chr.11: 22624705-22624705 on Build GRCh38
rs758608960 Chr.11: 22624952-22624952 on Build GRCh38
rs778035919 Chr.11: 22624771-22624771 on Build GRCh38
rs761621055 Chr.11: 22624707-22624707 on Build GRCh38
rs1029409487 Chr.11: 22624668-22624668 on Build GRCh38
rs149687560 Chr.11: 22624799-22624799 on Build GRCh38
rs748104707 Chr.11: 22624869-22624869 on Build GRCh38
rs756216888 Chr.11: 22624642-22624642 on Build GRCh38
rs755837313 Chr.11: 22624922-22624922 on Build GRCh38
rs886048157 Chr.11: 22624805-22624805 on Build GRCh38
rs748482579 Chr.11: 22624649-22624649 on Build GRCh38
rs767027638 Chr.11: 22624905-22624905 on Build GRCh38
rs781690040 Chr.11: 22624646-22624646 on Build GRCh38
rs762692664 Chr.11: 22624814-22624814 on Build GRCh38
rs759211548 Chr.11: 22624987-22624987 on Build GRCh38
rs199717587 Chr.11: 22624904-22624904 on Build GRCh38
rs757578045 Chr.11: 22624938-22624938 on Build GRCh38
rs146848948 Chr.11: 22624962-22624962 on Build GRCh38
rs368940253 Chr.11: 22624954-22624954 on Build GRCh38
rs763112358 Chr.11: 22624677-22624677 on Build GRCh38
rs1014086470 Chr.11: 22624556-22624556 on Build GRCh38
rs978253197 Chr.11: 22624929-22624929 on Build GRCh38
rs372140217 Chr.11: 22624900-22624900 on Build GRCh38
rs16909782 Chr.11: 22624580-22624580 on Build GRCh38
rs36045913 Chr.11: 22624986-22624986 on Build GRCh38
rs749487579 Chr.11: 22624773-22624773 on Build GRCh38
rs148768428 Chr.11: 22624864-22624864 on Build GRCh38
rs187993131 Chr.11: 22624786-22624786 on Build GRCh38
rs1029853294 Chr.11: 22624950-22624950 on Build GRCh38
rs770063791 Chr.11: 22624655-22624655 on Build GRCh38
rs200505409 Chr.11: 22624670-22624670 on Build GRCh38
rs756625596 Chr.11: 22624840-22624840 on Build GRCh38
rs749718315 Chr.11: 22624850-22624850 on Build GRCh38
rs45451294 Chr.11: 22624852-22624852 on Build GRCh38
rs758175326 Chr.11: 22624747-22624747 on Build GRCh38
rs762782397 Chr.11: 22624887-22624887 on Build GRCh38
rs61752917 Chr.11: 22624757-22624757 on Build GRCh38
rs887011892 Chr.11: 22624574-22624574 on Build GRCh38
rs766067137 Chr.11: 22624816-22624816 on Build GRCh38
rs1025590894 Chr.11: 22624560-22624560 on Build GRCh38
rs780747014 Chr.11: 22624934-22624934 on Build GRCh38
rs761698961 Chr.11: 22624809-22624809 on Build GRCh38
rs7103293 Chr.11: 22624928-22624928 on Build GRCh38
rs761238804 Chr.11: 22624926-22624926 on Build GRCh38
rs754017292 Chr.11: 22624784-22624784 on Build GRCh38
rs1016588203 Chr.11: 22624599-22624599 on Build GRCh38
rs774102674 Chr.11: 22624901-22624901 on Build GRCh38
rs750623273 Chr.11: 22624951-22624951 on Build GRCh38
rs549350470 Chr.11: 22624781-22624781 on Build GRCh38
rs183269887 Chr.11: 22624974-22624974 on Build GRCh38
rs368170054 Chr.11: 22624763-22624763 on Build GRCh38
rs766964364 Chr.11: 22624825-22624825 on Build GRCh38
rs754545069 Chr.11: 22624755-22624755 on Build GRCh38
rs571674814 Chr.11: 22624924-22624924 on Build GRCh38
rs372311535 Chr.11: 22624628-22624628 on Build GRCh38
rs200073415 Chr.11: 22624884-22624884 on Build GRCh38
rs200416138 Chr.11: 22624841-22624841 on Build GRCh38
rs770964562 Chr.11: 22624679-22624679 on Build GRCh38
rs748622528 Chr.11: 22624761-22624761 on Build GRCh38
rs376792172 Chr.11: 22624943-22624943 on Build GRCh38
rs751184770 Chr.11: 22624753-22624753 on Build GRCh38
rs757431699 Chr.11: 22624838-22624838 on Build GRCh38
rs912132298 Chr.11: 22624930-22624930 on Build GRCh38
rs982546703 Chr.11: 22624552-22624552 on Build GRCh38
rs587778342 Chr.11: 22624856-22624856 on Build GRCh38
rs1039138213 Chr.11: 22624768-22624768 on Build GRCh38
rs751274756 Chr.11: 22624818-22624818 on Build GRCh38
rs760590516 Chr.11: 22624697-22624697 on Build GRCh38

View Full Product Details