Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22821914-22822446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2
Entrez Gene ID
2620
Gene Name
growth arrest specific 2
Gene Aliases
-
UniGene
Hs.711297
Gene Chromosome Location
Chr.11: 22626113-22885930 on Build GRCh38
Transcripts
XM_017017532.1 NM_001320340.1 NM_001320341.1 NM_001320342.1 NM_148893.2 NR_135213.1

Gene Symbol
SVIP
Entrez Gene ID
258010
Gene Name
small VCP interacting protein
Gene Aliases
-
UniGene
Hs.349096
Gene Chromosome Location
Chr.11: 22820263-22829862 on Build GRCh38
Transcripts
XM_017017532.1 NM_001320340.1 NM_001320341.1 NM_001320342.1 NM_148893.2 NR_135213.1

SNP Detail

SNP ID Chromosome Location
rs1010787851 Chr.11: 22822260-22822260 on Build GRCh38
rs761313846 Chr.11: 22822259-22822259 on Build GRCh38
rs373546642 Chr.11: 22822227-22822227 on Build GRCh38
rs965445358 Chr.11: 22822379-22822379 on Build GRCh38
rs77679271 Chr.11: 22822420-22822420 on Build GRCh38
rs970242753 Chr.11: 22822157-22822157 on Build GRCh38
rs1054884069 Chr.11: 22822255-22822255 on Build GRCh38
rs939224076 Chr.11: 22822224-22822224 on Build GRCh38
rs889810464 Chr.11: 22822391-22822391 on Build GRCh38
rs372114510 Chr.11: 22822030-22822030 on Build GRCh38
rs764719193 Chr.11: 22822371-22822371 on Build GRCh38
rs928776162 Chr.11: 22822176-22822176 on Build GRCh38
rs550912756 Chr.11: 22822360-22822360 on Build GRCh38
rs1031399955 Chr.11: 22822066-22822066 on Build GRCh38
rs889003403 Chr.11: 22821959-22821959 on Build GRCh38
rs921547540 Chr.11: 22822010-22822010 on Build GRCh38
rs111518678 Chr.11: 22822216-22822216 on Build GRCh38
rs998836072 Chr.11: 22822333-22822333 on Build GRCh38
rs989824171 Chr.11: 22822111-22822111 on Build GRCh38
rs533432524 Chr.11: 22822399-22822399 on Build GRCh38
rs577813412 Chr.11: 22821949-22821949 on Build GRCh38
rs79232341 Chr.11: 22822421-22822421 on Build GRCh38
rs939784306 Chr.11: 22822130-22822130 on Build GRCh38
rs965881511 Chr.11: 22822015-22822015 on Build GRCh38
rs562509862 Chr.11: 22822027-22822027 on Build GRCh38
rs147353550 Chr.11: 22822148-22822148 on Build GRCh38
rs112993566 Chr.11: 22822147-22822147 on Build GRCh38
rs981675419 Chr.11: 22822159-22822159 on Build GRCh38
rs1022056645 Chr.11: 22822280-22822280 on Build GRCh38
rs541897891 Chr.11: 22822081-22822081 on Build GRCh38
rs191803012 Chr.11: 22821952-22821952 on Build GRCh38
rs976958177 Chr.11: 22822387-22822387 on Build GRCh38
rs550468413 Chr.11: 22822411-22822411 on Build GRCh38
rs904497884 Chr.11: 22822287-22822287 on Build GRCh38
rs571030346 Chr.11: 22822398-22822398 on Build GRCh38
rs879698191 Chr.11: 22822178-22822178 on Build GRCh38
rs563379292 Chr.11: 22822115-22822115 on Build GRCh38

View Full Product Details