Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27350072-27350570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC34
Entrez Gene ID
91057
Gene Name
coiled-coil domain containing 34
Gene Aliases
L15, NY-REN-41, RAMA3
UniGene
Hs.143733
Gene Chromosome Location
Chr.11: 27338514-27363248 on Build GRCh38
Transcripts
NM_030771.1 NM_080654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323439 c.524G>A p.R175K Chr.11: 27350414-27350414 on Build GRCh38
4669905 c.550_551insA p.R184fs*6 Chr.11: 27350387-27350388 on Build GRCh38
5014985 c.598A>T p.N200Y Chr.11: 27350340-27350340 on Build GRCh38
2113302 c.621A>G p.K207K Chr.11: 27350317-27350317 on Build GRCh38
6324092 c.529G>A p.E177K Chr.11: 27350409-27350409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776602769 Chr.11: 27350173-27350174 on Build GRCh38
rs754108582 Chr.11: 27350434-27350434 on Build GRCh38
rs776561161 Chr.11: 27350409-27350409 on Build GRCh38
rs1011457345 Chr.11: 27350188-27350188 on Build GRCh38
rs754725072 Chr.11: 27350341-27350341 on Build GRCh38
rs868243829 Chr.11: 27350354-27350354 on Build GRCh38
rs1053247935 Chr.11: 27350492-27350492 on Build GRCh38
rs747671085 Chr.11: 27350244-27350244 on Build GRCh38
rs780941577 Chr.11: 27350344-27350344 on Build GRCh38
rs200130322 Chr.11: 27350297-27350297 on Build GRCh38
rs755584192 Chr.11: 27350361-27350361 on Build GRCh38
rs372057780 Chr.11: 27350317-27350317 on Build GRCh38
rs777407261 Chr.11: 27350487-27350487 on Build GRCh38
rs746579353 Chr.11: 27350531-27350531 on Build GRCh38
rs779857663 Chr.11: 27350230-27350230 on Build GRCh38
rs144106436 Chr.11: 27350116-27350116 on Build GRCh38
rs1052957358 Chr.11: 27350119-27350119 on Build GRCh38
rs955895940 Chr.11: 27350342-27350342 on Build GRCh38
rs746359107 Chr.11: 27350412-27350414 on Build GRCh38
rs1022689662 Chr.11: 27350273-27350273 on Build GRCh38
rs760130773 Chr.11: 27350143-27350143 on Build GRCh38
rs893078234 Chr.11: 27350187-27350187 on Build GRCh38
rs763228021 Chr.11: 27350269-27350269 on Build GRCh38
rs753156605 Chr.11: 27350279-27350279 on Build GRCh38
rs368492832 Chr.11: 27350396-27350396 on Build GRCh38
rs747641807 Chr.11: 27350359-27350359 on Build GRCh38
rs539575089 Chr.11: 27350277-27350277 on Build GRCh38
rs752336618 Chr.11: 27350483-27350483 on Build GRCh38
rs759819505 Chr.11: 27350295-27350295 on Build GRCh38
rs780888054 Chr.11: 27350237-27350237 on Build GRCh38
rs752825920 Chr.11: 27350304-27350304 on Build GRCh38
rs887091591 Chr.11: 27350167-27350167 on Build GRCh38
rs753875243 Chr.11: 27350469-27350470 on Build GRCh38
rs750692514 Chr.11: 27350406-27350410 on Build GRCh38
rs550723980 Chr.11: 27350163-27350163 on Build GRCh38
rs186613235 Chr.11: 27350309-27350309 on Build GRCh38
rs764177079 Chr.11: 27350407-27350407 on Build GRCh38
rs766336152 Chr.11: 27350320-27350320 on Build GRCh38
rs749860561 Chr.11: 27350374-27350374 on Build GRCh38
rs373292888 Chr.11: 27350489-27350489 on Build GRCh38
rs1027160736 Chr.11: 27350365-27350365 on Build GRCh38
rs755708578 Chr.11: 27350484-27350484 on Build GRCh38
rs1004174133 Chr.11: 27350197-27350197 on Build GRCh38
rs780237282 Chr.11: 27350428-27350432 on Build GRCh38
rs771420586 Chr.11: 27350376-27350376 on Build GRCh38
rs374928596 Chr.11: 27350233-27350233 on Build GRCh38
rs758864832 Chr.11: 27350428-27350428 on Build GRCh38
rs774682128 Chr.11: 27350291-27350291 on Build GRCh38
rs757185044 Chr.11: 27350199-27350199 on Build GRCh38
rs921420674 Chr.11: 27350455-27350459 on Build GRCh38
rs758172289 Chr.11: 27350216-27350216 on Build GRCh38
rs750212317 Chr.11: 27350201-27350201 on Build GRCh38
rs754133054 Chr.11: 27350243-27350244 on Build GRCh38
rs981114554 Chr.11: 27350464-27350464 on Build GRCh38
rs184047802 Chr.11: 27350458-27350458 on Build GRCh38
rs758143001 Chr.11: 27350314-27350314 on Build GRCh38
rs767991626 Chr.11: 27350190-27350190 on Build GRCh38
rs751397560 Chr.11: 27350335-27350335 on Build GRCh38
rs193131353 Chr.11: 27350414-27350414 on Build GRCh38
rs765122060 Chr.11: 27350308-27350308 on Build GRCh38
rs889803480 Chr.11: 27350275-27350275 on Build GRCh38
rs773728260 Chr.11: 27350261-27350261 on Build GRCh38
rs772307472 Chr.11: 27350397-27350397 on Build GRCh38
rs1048509191 Chr.11: 27350480-27350480 on Build GRCh38
rs12364852 Chr.11: 27350364-27350364 on Build GRCh38
rs779710295 Chr.11: 27350513-27350513 on Build GRCh38
rs543328459 Chr.11: 27350265-27350270 on Build GRCh38
rs770225045 Chr.11: 27350248-27350248 on Build GRCh38
rs766395096 Chr.11: 27350438-27350438 on Build GRCh38
rs751419821 Chr.11: 27350449-27350449 on Build GRCh38
rs760763446 Chr.11: 27350306-27350306 on Build GRCh38
rs374290862 Chr.11: 27350382-27350382 on Build GRCh38
rs760675624 Chr.11: 27350401-27350401 on Build GRCh38
rs767278483 Chr.11: 27350475-27350475 on Build GRCh38
rs866906321 Chr.11: 27350524-27350524 on Build GRCh38
rs374518134 Chr.11: 27350281-27350281 on Build GRCh38

View Full Product Details