Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27357292-27357825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC34
Entrez Gene ID
91057
Gene Name
coiled-coil domain containing 34
Gene Aliases
L15, NY-REN-41, RAMA3
UniGene
Hs.143733
Gene Chromosome Location
Chr.11: 27338514-27363248 on Build GRCh38
Transcripts
NM_030771.1 NM_080654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243507 c.476G>A p.R159Q Chr.11: 27357425-27357425 on Build GRCh38
7243508 c.476G>A p.R159Q Chr.11: 27357425-27357425 on Build GRCh38
7222914 c.471T>A p.R157R Chr.11: 27357430-27357430 on Build GRCh38
7222915 c.471T>A p.R157R Chr.11: 27357430-27357430 on Build GRCh38
341649 c.399A>T p.K133N Chr.11: 27357502-27357502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752428338 Chr.11: 27357442-27357442 on Build GRCh38
rs201921821 Chr.11: 27357513-27357513 on Build GRCh38
rs778632448 Chr.11: 27357613-27357613 on Build GRCh38
rs574851389 Chr.11: 27357317-27357317 on Build GRCh38
rs758039493 Chr.11: 27357711-27357711 on Build GRCh38
rs759413616 Chr.11: 27357434-27357434 on Build GRCh38
rs746683112 Chr.11: 27357420-27357422 on Build GRCh38
rs772356601 Chr.11: 27357373-27357373 on Build GRCh38
rs1044558915 Chr.11: 27357489-27357489 on Build GRCh38
rs1009857390 Chr.11: 27357632-27357632 on Build GRCh38
rs757812869 Chr.11: 27357357-27357357 on Build GRCh38
rs759613748 Chr.11: 27357539-27357539 on Build GRCh38
rs550549617 Chr.11: 27357637-27357637 on Build GRCh38
rs1041119536 Chr.11: 27357624-27357624 on Build GRCh38
rs754876812 Chr.11: 27357354-27357355 on Build GRCh38
rs760340377 Chr.11: 27357564-27357564 on Build GRCh38
rs760591930 Chr.11: 27357333-27357333 on Build GRCh38
rs746747136 Chr.11: 27357494-27357495 on Build GRCh38
rs1012465640 Chr.11: 27357778-27357778 on Build GRCh38
rs969728329 Chr.11: 27357795-27357795 on Build GRCh38
rs994318483 Chr.11: 27357687-27357687 on Build GRCh38
rs768943068 Chr.11: 27357511-27357511 on Build GRCh38
rs564868877 Chr.11: 27357764-27357764 on Build GRCh38
rs772037374 Chr.11: 27357437-27357438 on Build GRCh38
rs888687236 Chr.11: 27357743-27357743 on Build GRCh38
rs901355497 Chr.11: 27357650-27357650 on Build GRCh38
rs770337805 Chr.11: 27357550-27357551 on Build GRCh38
rs1028895683 Chr.11: 27357716-27357716 on Build GRCh38
rs761932748 Chr.11: 27357411-27357411 on Build GRCh38
rs147569475 Chr.11: 27357331-27357331 on Build GRCh38
rs780537848 Chr.11: 27357498-27357498 on Build GRCh38
rs747317938 Chr.11: 27357388-27357388 on Build GRCh38
rs748239159 Chr.11: 27357514-27357514 on Build GRCh38
rs768759531 Chr.11: 27357402-27357402 on Build GRCh38
rs764789367 Chr.11: 27357591-27357591 on Build GRCh38
rs779344286 Chr.11: 27357360-27357360 on Build GRCh38
rs771870900 Chr.11: 27357559-27357559 on Build GRCh38
rs763634655 Chr.11: 27357445-27357445 on Build GRCh38
rs769933003 Chr.11: 27357517-27357517 on Build GRCh38
rs766448184 Chr.11: 27357425-27357425 on Build GRCh38
rs200766443 Chr.11: 27357362-27357362 on Build GRCh38
rs753422968 Chr.11: 27357569-27357569 on Build GRCh38
rs146985302 Chr.11: 27357478-27357478 on Build GRCh38
rs747308218 Chr.11: 27357501-27357501 on Build GRCh38
rs147823135 Chr.11: 27357426-27357426 on Build GRCh38
rs753411958 Chr.11: 27357465-27357465 on Build GRCh38
rs988717619 Chr.11: 27357488-27357488 on Build GRCh38
rs753712619 Chr.11: 27357567-27357567 on Build GRCh38
rs3832796 Chr.11: 27357764-27357764 on Build GRCh38
rs779022175 Chr.11: 27357379-27357383 on Build GRCh38
rs1023057194 Chr.11: 27357790-27357790 on Build GRCh38
rs996995384 Chr.11: 27357659-27357659 on Build GRCh38
rs3741407 Chr.11: 27357580-27357580 on Build GRCh38
rs200975862 Chr.11: 27357439-27357439 on Build GRCh38
rs760413266 Chr.11: 27357443-27357443 on Build GRCh38
rs747901021 Chr.11: 27357433-27357435 on Build GRCh38
rs374839645 Chr.11: 27357480-27357480 on Build GRCh38
rs3196894 Chr.11: 27357432-27357432 on Build GRCh38
rs911192420 Chr.11: 27357598-27357598 on Build GRCh38
rs773412390 Chr.11: 27357531-27357531 on Build GRCh38
rs775593174 Chr.11: 27357382-27357382 on Build GRCh38
rs138159804 Chr.11: 27357479-27357479 on Build GRCh38
rs141376786 Chr.11: 27357436-27357436 on Build GRCh38
rs777533036 Chr.11: 27357470-27357470 on Build GRCh38

View Full Product Details