Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27660069-27660572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDNF
Entrez Gene ID
627
Gene Name
brain derived neurotrophic factor
Gene Aliases
ANON2, BULN2
UniGene
Hs.502182
Gene Chromosome Location
Chr.11: 27654893-27722058 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

Gene Symbol
BDNF-AS
Entrez Gene ID
497258
Gene Name
BDNF antisense RNA
Gene Aliases
ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
UniGene
Hs.675323
Gene Chromosome Location
Chr.11: 27506852-27698171 on Build GRCh38
Transcripts
NM_001143805.1 NM_001143806.1 NM_001143807.1 NM_001143808.1 NM_001143809.1 NM_001143810.1 NM_001143811.1 NM_001143812.1 NM_001143813.1 NM_001143814.1 NM_001709.4 NM_170731.4 NM_170732.4 NM_170733.3 NM_170734.3 XM_011520280.2 NR_002832.2 NR_033312.1 NR_033313.1 NR_033314.1 NR_033315.1

SNP Detail

SNP ID Chromosome Location
rs985639084 Chr.11: 27660479-27660479 on Build GRCh38
rs970563140 Chr.11: 27660411-27660411 on Build GRCh38
rs572365353 Chr.11: 27660180-27660180 on Build GRCh38
rs1026919334 Chr.11: 27660349-27660349 on Build GRCh38
rs74717653 Chr.11: 27660199-27660199 on Build GRCh38
rs766280279 Chr.11: 27660313-27660313 on Build GRCh38
rs993387663 Chr.11: 27660419-27660419 on Build GRCh38
rs57432706 Chr.11: 27660493-27660493 on Build GRCh38
rs982787424 Chr.11: 27660396-27660396 on Build GRCh38
rs892199666 Chr.11: 27660476-27660476 on Build GRCh38
rs1002938282 Chr.11: 27660426-27660426 on Build GRCh38
rs903506268 Chr.11: 27660235-27660235 on Build GRCh38
rs77959657 Chr.11: 27660434-27660434 on Build GRCh38
rs901469630 Chr.11: 27660333-27660333 on Build GRCh38
rs1019793350 Chr.11: 27660540-27660540 on Build GRCh38
rs763373270 Chr.11: 27660357-27660357 on Build GRCh38
rs1013190578 Chr.11: 27660156-27660156 on Build GRCh38
rs193044018 Chr.11: 27660468-27660468 on Build GRCh38
rs1043716067 Chr.11: 27660227-27660227 on Build GRCh38
rs565374712 Chr.11: 27660367-27660367 on Build GRCh38
rs989024309 Chr.11: 27660284-27660284 on Build GRCh38
rs956510594 Chr.11: 27660282-27660282 on Build GRCh38
rs748194649 Chr.11: 27660465-27660465 on Build GRCh38
rs369883847 Chr.11: 27660462-27660463 on Build GRCh38
rs117314519 Chr.11: 27660368-27660368 on Build GRCh38
rs189558882 Chr.11: 27660400-27660400 on Build GRCh38
rs1006031010 Chr.11: 27660376-27660376 on Build GRCh38
rs535486417 Chr.11: 27660292-27660292 on Build GRCh38
rs947570462 Chr.11: 27660291-27660291 on Build GRCh38
rs765941153 Chr.11: 27660541-27660541 on Build GRCh38
rs57083135 Chr.11: 27660309-27660309 on Build GRCh38
rs943104563 Chr.11: 27660318-27660318 on Build GRCh38
rs139827433 Chr.11: 27660097-27660097 on Build GRCh38
rs529099414 Chr.11: 27660251-27660251 on Build GRCh38
rs914753727 Chr.11: 27660286-27660286 on Build GRCh38
rs764270718 Chr.11: 27660232-27660232 on Build GRCh38
rs892252073 Chr.11: 27660290-27660290 on Build GRCh38
rs181528398 Chr.11: 27660111-27660111 on Build GRCh38
rs968941370 Chr.11: 27660473-27660473 on Build GRCh38
rs993382170 Chr.11: 27660356-27660356 on Build GRCh38
rs1040461461 Chr.11: 27660321-27660321 on Build GRCh38
rs567103999 Chr.11: 27660319-27660319 on Build GRCh38
rs566667401 Chr.11: 27660418-27660418 on Build GRCh38
rs41274434 Chr.11: 27660096-27660096 on Build GRCh38
rs956033723 Chr.11: 27660447-27660447 on Build GRCh38
rs1022737384 Chr.11: 27660399-27660399 on Build GRCh38
rs975326278 Chr.11: 27660330-27660330 on Build GRCh38
rs1030675794 Chr.11: 27660279-27660279 on Build GRCh38
rs894717240 Chr.11: 27660124-27660124 on Build GRCh38
rs184293959 Chr.11: 27660287-27660287 on Build GRCh38
rs928626443 Chr.11: 27660258-27660258 on Build GRCh38
rs908629026 Chr.11: 27660094-27660094 on Build GRCh38
rs1000490706 Chr.11: 27660260-27660263 on Build GRCh38

View Full Product Details