Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64959186-64959725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
UniGene
Hs.567793
Gene Chromosome Location
Chr.11: 64938230-64972104 on Build GRCh38
Transcripts
NM_001037225.2 NM_001300803.1 NM_001318808.1 XM_005273918.2 XM_006718502.2 XM_006718503.3 XM_006718504.2 XM_006718505.2 XM_011544940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584960 c.46C>G p.L16V Chr.11: 64959360-64959360 on Build GRCh38
5839204 c.38C>T p.T13M Chr.11: 64959368-64959368 on Build GRCh38
6537403 c.41G>C p.R14T Chr.11: 64959365-64959365 on Build GRCh38
3953639 c.3G>T p.M1I Chr.11: 64959403-64959403 on Build GRCh38
6614136 c.21C>T p.T7T Chr.11: 64959385-64959385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034919597 Chr.11: 64959498-64959498 on Build GRCh38
rs778536126 Chr.11: 64959291-64959291 on Build GRCh38
rs202158936 Chr.11: 64959286-64959286 on Build GRCh38
rs900234732 Chr.11: 64959413-64959413 on Build GRCh38
rs602404 Chr.11: 64959229-64959229 on Build GRCh38
rs555081834 Chr.11: 64959692-64959692 on Build GRCh38
rs772822310 Chr.11: 64959417-64959417 on Build GRCh38
rs17146508 Chr.11: 64959615-64959615 on Build GRCh38
rs781719660 Chr.11: 64959380-64959380 on Build GRCh38
rs116085655 Chr.11: 64959554-64959554 on Build GRCh38
rs553613295 Chr.11: 64959557-64959557 on Build GRCh38
rs749060692 Chr.11: 64959365-64959371 on Build GRCh38
rs1011105442 Chr.11: 64959416-64959416 on Build GRCh38
rs368913675 Chr.11: 64959446-64959446 on Build GRCh38
rs370436062 Chr.11: 64959284-64959284 on Build GRCh38
rs367907837 Chr.11: 64959422-64959422 on Build GRCh38
rs754455939 Chr.11: 64959358-64959358 on Build GRCh38
rs571182441 Chr.11: 64959523-64959523 on Build GRCh38
rs371562641 Chr.11: 64959433-64959433 on Build GRCh38
rs888993351 Chr.11: 64959246-64959246 on Build GRCh38
rs754714213 Chr.11: 64959677-64959677 on Build GRCh38
rs746400819 Chr.11: 64959381-64959381 on Build GRCh38
rs747842850 Chr.11: 64959292-64959292 on Build GRCh38
rs753774551 Chr.11: 64959356-64959356 on Build GRCh38
rs748832010 Chr.11: 64959287-64959287 on Build GRCh38
rs1022124993 Chr.11: 64959459-64959459 on Build GRCh38
rs947268754 Chr.11: 64959254-64959254 on Build GRCh38
rs533015164 Chr.11: 64959218-64959218 on Build GRCh38
rs767657443 Chr.11: 64959313-64959313 on Build GRCh38
rs748223232 Chr.11: 64959426-64959426 on Build GRCh38
rs769971920 Chr.11: 64959384-64959384 on Build GRCh38
rs374717202 Chr.11: 64959665-64959665 on Build GRCh38
rs918727556 Chr.11: 64959605-64959605 on Build GRCh38
rs1051322035 Chr.11: 64959635-64959635 on Build GRCh38
rs200188712 Chr.11: 64959315-64959315 on Build GRCh38
rs200290554 Chr.11: 64959368-64959368 on Build GRCh38
rs768472657 Chr.11: 64959384-64959389 on Build GRCh38
rs779356342 Chr.11: 64959352-64959352 on Build GRCh38
rs148543749 Chr.11: 64959352-64959352 on Build GRCh38
rs759649080 Chr.11: 64959325-64959325 on Build GRCh38
rs773899246 Chr.11: 64959425-64959429 on Build GRCh38
rs947216333 Chr.11: 64959637-64959637 on Build GRCh38
rs1038529368 Chr.11: 64959224-64959224 on Build GRCh38
rs761488245 Chr.11: 64959434-64959437 on Build GRCh38
rs922629605 Chr.11: 64959589-64959589 on Build GRCh38
rs375056403 Chr.11: 64959265-64959265 on Build GRCh38
rs867650230 Chr.11: 64959354-64959354 on Build GRCh38
rs934286525 Chr.11: 64959666-64959666 on Build GRCh38
rs548422925 Chr.11: 64959360-64959360 on Build GRCh38
rs763223170 Chr.11: 64959411-64959411 on Build GRCh38
rs537367438 Chr.11: 64959488-64959488 on Build GRCh38
rs773591860 Chr.11: 64959309-64959309 on Build GRCh38
rs1042971401 Chr.11: 64959657-64959657 on Build GRCh38
rs755933994 Chr.11: 64959350-64959350 on Build GRCh38
rs766228834 Chr.11: 64959616-64959616 on Build GRCh38
rs550954072 Chr.11: 64959506-64959506 on Build GRCh38
rs760502813 Chr.11: 64959419-64959419 on Build GRCh38
rs747753223 Chr.11: 64959363-64959363 on Build GRCh38
rs900204332 Chr.11: 64959693-64959693 on Build GRCh38
rs142880411 Chr.11: 64959379-64959379 on Build GRCh38
rs750259177 Chr.11: 64959348-64959348 on Build GRCh38
rs774342709 Chr.11: 64959222-64959222 on Build GRCh38
rs1008583506 Chr.11: 64959367-64959367 on Build GRCh38
rs759496719 Chr.11: 64959444-64959444 on Build GRCh38
rs754526127 Chr.11: 64959290-64959290 on Build GRCh38
rs985973160 Chr.11: 64959612-64959612 on Build GRCh38
rs940051351 Chr.11: 64959212-64959212 on Build GRCh38
rs941472281 Chr.11: 64959220-64959220 on Build GRCh38
rs974567469 Chr.11: 64959604-64959604 on Build GRCh38
rs747435219 Chr.11: 64959311-64959311 on Build GRCh38
rs542599712 Chr.11: 64959581-64959581 on Build GRCh38
rs529276250 Chr.11: 64959569-64959569 on Build GRCh38
rs952904542 Chr.11: 64959449-64959449 on Build GRCh38
rs1056791349 Chr.11: 64959671-64959671 on Build GRCh38
rs747416147 Chr.11: 64959376-64959376 on Build GRCh38
rs775733031 Chr.11: 64959392-64959392 on Build GRCh38
rs765479119 Chr.11: 64959327-64959327 on Build GRCh38
rs767458213 Chr.11: 64959277-64959277 on Build GRCh38
rs941453926 Chr.11: 64959590-64959590 on Build GRCh38
rs977944379 Chr.11: 64959496-64959496 on Build GRCh38
rs772405396 Chr.11: 64959294-64959294 on Build GRCh38
rs961322724 Chr.11: 64959509-64959509 on Build GRCh38
rs375999745 Chr.11: 64959412-64959412 on Build GRCh38
rs990823898 Chr.11: 64959549-64959549 on Build GRCh38
rs529072438 Chr.11: 64959335-64959335 on Build GRCh38
rs775822980 Chr.11: 64959333-64959333 on Build GRCh38
rs942972599 Chr.11: 64959672-64959672 on Build GRCh38
rs1050264143 Chr.11: 64959241-64959241 on Build GRCh38
rs778045629 Chr.11: 64959371-64959371 on Build GRCh38
rs902822238 Chr.11: 64959282-64959282 on Build GRCh38
rs766131049 Chr.11: 64959420-64959420 on Build GRCh38
rs368431514 Chr.11: 64959336-64959336 on Build GRCh38
rs866513707 Chr.11: 64959398-64959398 on Build GRCh38

View Full Product Details