Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65083855-65084346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
UniGene
Hs.434886
Gene Chromosome Location
Chr.11: 65061009-65084143 on Build GRCh38
Transcripts
NM_080668.3 XM_005273733.3 XM_005273734.3 XM_011544743.2 XM_011544747.2 XM_011544748.2 XM_011544749.2 XM_017017158.1 NM_006782.3

Gene Symbol
ZFPL1
Entrez Gene ID
7542
Gene Name
zinc finger protein like 1
Gene Aliases
D11S750, MCG4
UniGene
Hs.712317
Gene Chromosome Location
Chr.11: 65084222-65088402 on Build GRCh38
Transcripts
NM_080668.3 XM_005273733.3 XM_005273734.3 XM_011544743.2 XM_011544747.2 XM_011544748.2 XM_011544749.2 XM_017017158.1 NM_006782.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6562923 c.17C>A p.T6K Chr.11: 65083962-65083962 on Build GRCh38
6624516 c.46G>A p.G16R Chr.11: 65083933-65083933 on Build GRCh38
6894797 c.23C>A p.S8Y Chr.11: 65083956-65083956 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772912965 Chr.11: 65083947-65083947 on Build GRCh38
rs916556103 Chr.11: 65084012-65084012 on Build GRCh38
rs969394466 Chr.11: 65084268-65084268 on Build GRCh38
rs557931801 Chr.11: 65084029-65084029 on Build GRCh38
rs370492535 Chr.11: 65084305-65084305 on Build GRCh38
rs891398499 Chr.11: 65084060-65084060 on Build GRCh38
rs545279705 Chr.11: 65084186-65084186 on Build GRCh38
rs1010613755 Chr.11: 65084111-65084111 on Build GRCh38
rs775947881 Chr.11: 65083991-65083991 on Build GRCh38
rs796321841 Chr.11: 65083983-65083983 on Build GRCh38
rs1014255414 Chr.11: 65084252-65084252 on Build GRCh38
rs912691021 Chr.11: 65084239-65084239 on Build GRCh38
rs1031292091 Chr.11: 65084037-65084037 on Build GRCh38
rs764111316 Chr.11: 65083990-65083990 on Build GRCh38
rs1017235115 Chr.11: 65084073-65084073 on Build GRCh38
rs749138054 Chr.11: 65083908-65083908 on Build GRCh38
rs990838946 Chr.11: 65083992-65083992 on Build GRCh38
rs990703150 Chr.11: 65084160-65084160 on Build GRCh38
rs750288574 Chr.11: 65084138-65084138 on Build GRCh38
rs776825000 Chr.11: 65083975-65083975 on Build GRCh38
rs984838414 Chr.11: 65084271-65084271 on Build GRCh38
rs1029814204 Chr.11: 65083933-65083933 on Build GRCh38
rs757558868 Chr.11: 65083898-65083898 on Build GRCh38
rs1051463652 Chr.11: 65084286-65084286 on Build GRCh38
rs748134667 Chr.11: 65083927-65083927 on Build GRCh38
rs758419261 Chr.11: 65084024-65084024 on Build GRCh38
rs1023600522 Chr.11: 65084260-65084260 on Build GRCh38
rs184454606 Chr.11: 65084013-65084013 on Build GRCh38
rs569223577 Chr.11: 65083996-65083996 on Build GRCh38
rs527487989 Chr.11: 65084261-65084261 on Build GRCh38
rs547109108 Chr.11: 65084280-65084280 on Build GRCh38
rs930195817 Chr.11: 65084045-65084045 on Build GRCh38
rs900142330 Chr.11: 65084075-65084075 on Build GRCh38
rs999720674 Chr.11: 65084212-65084212 on Build GRCh38
rs948097762 Chr.11: 65084062-65084062 on Build GRCh38
rs939247293 Chr.11: 65083951-65083951 on Build GRCh38
rs35819335 Chr.11: 65083982-65083982 on Build GRCh38
rs116468755 Chr.11: 65084309-65084309 on Build GRCh38
rs767802234 Chr.11: 65084004-65084004 on Build GRCh38
rs372338615 Chr.11: 65083918-65083918 on Build GRCh38
rs778916067 Chr.11: 65084015-65084015 on Build GRCh38
rs549327680 Chr.11: 65083972-65083972 on Build GRCh38
rs962505868 Chr.11: 65084274-65084274 on Build GRCh38
rs573610682 Chr.11: 65084069-65084069 on Build GRCh38
rs960627628 Chr.11: 65084136-65084136 on Build GRCh38
rs941383534 Chr.11: 65084170-65084170 on Build GRCh38
rs375772067 Chr.11: 65083888-65083888 on Build GRCh38
rs974474454 Chr.11: 65084175-65084175 on Build GRCh38
rs562435169 Chr.11: 65084101-65084101 on Build GRCh38
rs746580955 Chr.11: 65083962-65083962 on Build GRCh38
rs930030204 Chr.11: 65084205-65084205 on Build GRCh38
rs1035243041 Chr.11: 65084131-65084131 on Build GRCh38
rs1043480054 Chr.11: 65084319-65084319 on Build GRCh38
rs972564327 Chr.11: 65084275-65084275 on Build GRCh38
rs941237359 Chr.11: 65084124-65084124 on Build GRCh38
rs774441843 Chr.11: 65084019-65084019 on Build GRCh38
rs189412398 Chr.11: 65084036-65084036 on Build GRCh38
rs192755142 Chr.11: 65083895-65083895 on Build GRCh38
rs748081644 Chr.11: 65084016-65084016 on Build GRCh38
rs150470368 Chr.11: 65084130-65084130 on Build GRCh38
rs535457002 Chr.11: 65083968-65083968 on Build GRCh38
rs1051288462 Chr.11: 65084227-65084227 on Build GRCh38
rs779656242 Chr.11: 65083907-65083907 on Build GRCh38
rs942962184 Chr.11: 65084241-65084241 on Build GRCh38
rs916588749 Chr.11: 65084169-65084169 on Build GRCh38
rs763520094 Chr.11: 65083995-65083995 on Build GRCh38
rs1051686557 Chr.11: 65084047-65084047 on Build GRCh38
rs765642943 Chr.11: 65083989-65083989 on Build GRCh38
rs553569981 Chr.11: 65084061-65084061 on Build GRCh38
rs759971528 Chr.11: 65083978-65083978 on Build GRCh38
rs533927705 Chr.11: 65084059-65084059 on Build GRCh38
rs754788323 Chr.11: 65084014-65084014 on Build GRCh38
rs897232145 Chr.11: 65084041-65084041 on Build GRCh38
rs536589552 Chr.11: 65084281-65084281 on Build GRCh38
rs565126382 Chr.11: 65084222-65084222 on Build GRCh38
rs909324703 Chr.11: 65084272-65084272 on Build GRCh38
rs779992074 Chr.11: 65084080-65084080 on Build GRCh38
rs529351852 Chr.11: 65084291-65084291 on Build GRCh38
rs757355571 Chr.11: 65084003-65084003 on Build GRCh38
rs966850140 Chr.11: 65084126-65084126 on Build GRCh38
rs889458198 Chr.11: 65084154-65084154 on Build GRCh38
rs771701995 Chr.11: 65083942-65083942 on Build GRCh38
rs1022084029 Chr.11: 65084118-65084118 on Build GRCh38
rs542280336 Chr.11: 65084078-65084078 on Build GRCh38

View Full Product Details