Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65183021-65183529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN1
Entrez Gene ID
823
Gene Name
calpain 1
Gene Aliases
CANP, CANP1, CANPL1, SPG76, muCANP, muCL
UniGene
Hs.502842
Gene Chromosome Location
Chr.11: 65181215-65212006 on Build GRCh38
Transcripts
NM_001198868.1 NM_001198869.1 NM_005186.3 NR_040008.1 XM_006718698.1 XM_011545292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6347323 c.323G>T p.C108F Chr.11: 65183183-65183183 on Build GRCh38
7176884 c.351C>G p.L117L Chr.11: 65183487-65183487 on Build GRCh38
3998535 c.363C>A p.I121I Chr.11: 65183499-65183499 on Build GRCh38
5967020 c.380A>G p.N127S Chr.11: 65183516-65183516 on Build GRCh38
79598 c.336G>A p.L112L Chr.11: 65183196-65183196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756493235 Chr.11: 65183239-65183239 on Build GRCh38
rs927762604 Chr.11: 65183356-65183356 on Build GRCh38
rs764619106 Chr.11: 65183136-65183136 on Build GRCh38
rs770916952 Chr.11: 65183126-65183126 on Build GRCh38
rs780553964 Chr.11: 65183408-65183408 on Build GRCh38
rs749341081 Chr.11: 65183179-65183179 on Build GRCh38
rs777552187 Chr.11: 65183210-65183210 on Build GRCh38
rs372457262 Chr.11: 65183468-65183468 on Build GRCh38
rs889192754 Chr.11: 65183426-65183426 on Build GRCh38
rs780457341 Chr.11: 65183178-65183178 on Build GRCh38
rs765872188 Chr.11: 65183460-65183460 on Build GRCh38
rs762462478 Chr.11: 65183222-65183222 on Build GRCh38
rs749441476 Chr.11: 65183116-65183116 on Build GRCh38
rs761505981 Chr.11: 65183225-65183225 on Build GRCh38
rs1010233435 Chr.11: 65183253-65183253 on Build GRCh38
rs555178066 Chr.11: 65183425-65183425 on Build GRCh38
rs755477674 Chr.11: 65183459-65183459 on Build GRCh38
rs75873264 Chr.11: 65183322-65183322 on Build GRCh38
rs768755758 Chr.11: 65183370-65183370 on Build GRCh38
rs779190441 Chr.11: 65183205-65183205 on Build GRCh38
rs776694688 Chr.11: 65183127-65183127 on Build GRCh38
rs370999727 Chr.11: 65183506-65183506 on Build GRCh38
rs754347903 Chr.11: 65183454-65183454 on Build GRCh38
rs17583 Chr.11: 65183499-65183499 on Build GRCh38
rs928499971 Chr.11: 65183065-65183065 on Build GRCh38
rs867424657 Chr.11: 65183046-65183046 on Build GRCh38
rs767838389 Chr.11: 65183226-65183226 on Build GRCh38
rs1052248788 Chr.11: 65183203-65183203 on Build GRCh38
rs369597923 Chr.11: 65183216-65183216 on Build GRCh38
rs2277307 Chr.11: 65183372-65183372 on Build GRCh38
rs570324200 Chr.11: 65183425-65183426 on Build GRCh38
rs373268116 Chr.11: 65183424-65183424 on Build GRCh38
rs769482925 Chr.11: 65183458-65183459 on Build GRCh38
rs754369068 Chr.11: 65183413-65183413 on Build GRCh38
rs757635396 Chr.11: 65183099-65183099 on Build GRCh38
rs759433792 Chr.11: 65183213-65183213 on Build GRCh38
rs752974994 Chr.11: 65183073-65183073 on Build GRCh38
rs759593212 Chr.11: 65183132-65183132 on Build GRCh38
rs117079749 Chr.11: 65183309-65183309 on Build GRCh38
rs757124933 Chr.11: 65183420-65183420 on Build GRCh38
rs397844195 Chr.11: 65183427-65183428 on Build GRCh38
rs748437197 Chr.11: 65183207-65183207 on Build GRCh38
rs575853678 Chr.11: 65183427-65183427 on Build GRCh38
rs752771134 Chr.11: 65183464-65183464 on Build GRCh38
rs190612316 Chr.11: 65183299-65183299 on Build GRCh38
rs767110140 Chr.11: 65183428-65183428 on Build GRCh38
rs1011682563 Chr.11: 65183273-65183273 on Build GRCh38
rs778906752 Chr.11: 65183416-65183416 on Build GRCh38
rs991107792 Chr.11: 65183410-65183410 on Build GRCh38
rs768622526 Chr.11: 65183181-65183181 on Build GRCh38
rs958269565 Chr.11: 65183263-65183263 on Build GRCh38
rs17885718 Chr.11: 65183167-65183167 on Build GRCh38
rs1035400636 Chr.11: 65183318-65183318 on Build GRCh38
rs747346683 Chr.11: 65183086-65183086 on Build GRCh38
rs758562395 Chr.11: 65183492-65183492 on Build GRCh38
rs562294874 Chr.11: 65183211-65183212 on Build GRCh38
rs746443334 Chr.11: 65183100-65183100 on Build GRCh38
rs534135243 Chr.11: 65183162-65183162 on Build GRCh38
rs893954472 Chr.11: 65183469-65183469 on Build GRCh38
rs370866733 Chr.11: 65183450-65183450 on Build GRCh38
rs772377336 Chr.11: 65183420-65183420 on Build GRCh38
rs752207698 Chr.11: 65183140-65183140 on Build GRCh38
rs750908895 Chr.11: 65183170-65183170 on Build GRCh38
rs17884377 Chr.11: 65183419-65183420 on Build GRCh38
rs577679236 Chr.11: 65183503-65183503 on Build GRCh38
rs768883809 Chr.11: 65183124-65183124 on Build GRCh38
rs763747881 Chr.11: 65183157-65183157 on Build GRCh38
rs748347074 Chr.11: 65183417-65183417 on Build GRCh38
rs1019967195 Chr.11: 65183293-65183293 on Build GRCh38
rs769854700 Chr.11: 65183110-65183110 on Build GRCh38
rs539895915 Chr.11: 65183223-65183223 on Build GRCh38
rs377326288 Chr.11: 65183212-65183212 on Build GRCh38
rs376674277 Chr.11: 65183125-65183125 on Build GRCh38
rs775689033 Chr.11: 65183113-65183113 on Build GRCh38
rs750749257 Chr.11: 65183229-65183229 on Build GRCh38
rs757616692 Chr.11: 65183164-65183164 on Build GRCh38
rs182125926 Chr.11: 65183429-65183429 on Build GRCh38
rs201751778 Chr.11: 65183171-65183171 on Build GRCh38
rs745597294 Chr.11: 65183422-65183422 on Build GRCh38
rs1020420950 Chr.11: 65183254-65183254 on Build GRCh38
rs762388125 Chr.11: 65183147-65183147 on Build GRCh38
rs941907335 Chr.11: 65183401-65183401 on Build GRCh38
rs981980394 Chr.11: 65183295-65183295 on Build GRCh38
rs778159460 Chr.11: 65183421-65183421 on Build GRCh38
rs201805317 Chr.11: 65183218-65183218 on Build GRCh38
rs546557386 Chr.11: 65183415-65183415 on Build GRCh38
rs868774831 Chr.11: 65183341-65183341 on Build GRCh38
rs769718790 Chr.11: 65183215-65183215 on Build GRCh38

View Full Product Details