Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65386890-65387397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD8
Entrez Gene ID
83786
Gene Name
FERM domain containing 8
Gene Aliases
FKSG44
UniGene
Hs.578433
Gene Chromosome Location
Chr.11: 65386570-65413525 on Build GRCh38
Transcripts
NM_001300832.1 NM_001300833.1 NM_031904.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2165517 c.85+7C>T p.? Chr.11: 65387128-65387128 on Build GRCh38
6069879 c.20G>T p.S7I Chr.11: 65387056-65387056 on Build GRCh38
7159978 c.43G>A p.E15K Chr.11: 65387079-65387079 on Build GRCh38
6261698 c.59G>T p.S20I Chr.11: 65387095-65387095 on Build GRCh38
6246716 c.8G>T p.G3V Chr.11: 65387044-65387044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894724980 Chr.11: 65386931-65386931 on Build GRCh38
rs751112371 Chr.11: 65387265-65387266 on Build GRCh38
rs1002178395 Chr.11: 65387370-65387370 on Build GRCh38
rs1057183334 Chr.11: 65386928-65386928 on Build GRCh38
rs746217293 Chr.11: 65386988-65386988 on Build GRCh38
rs748144632 Chr.11: 65387104-65387104 on Build GRCh38
rs762807697 Chr.11: 65387067-65387067 on Build GRCh38
rs762825075 Chr.11: 65387148-65387148 on Build GRCh38
rs1025796156 Chr.11: 65387020-65387020 on Build GRCh38
rs755400226 Chr.11: 65387099-65387099 on Build GRCh38
rs772161111 Chr.11: 65387048-65387048 on Build GRCh38
rs774146628 Chr.11: 65387241-65387241 on Build GRCh38
rs761439052 Chr.11: 65387081-65387081 on Build GRCh38
rs142241454 Chr.11: 65386993-65386993 on Build GRCh38
rs938645738 Chr.11: 65386924-65386924 on Build GRCh38
rs779522359 Chr.11: 65387100-65387100 on Build GRCh38
rs760339773 Chr.11: 65387173-65387173 on Build GRCh38
rs1054081820 Chr.11: 65387283-65387283 on Build GRCh38
rs970705043 Chr.11: 65387021-65387021 on Build GRCh38
rs994057505 Chr.11: 65387373-65387373 on Build GRCh38
rs892512440 Chr.11: 65386974-65386976 on Build GRCh38
rs748292470 Chr.11: 65387047-65387047 on Build GRCh38
rs775275261 Chr.11: 65387060-65387060 on Build GRCh38
rs772259371 Chr.11: 65387115-65387115 on Build GRCh38
rs571676297 Chr.11: 65387239-65387258 on Build GRCh38
rs769221214 Chr.11: 65387011-65387011 on Build GRCh38
rs758840411 Chr.11: 65387203-65387203 on Build GRCh38
rs767050401 Chr.11: 65387082-65387082 on Build GRCh38
rs757419896 Chr.11: 65387218-65387218 on Build GRCh38
rs547115606 Chr.11: 65387278-65387278 on Build GRCh38
rs766161238 Chr.11: 65387174-65387174 on Build GRCh38
rs111632299 Chr.11: 65387034-65387034 on Build GRCh38
rs564327841 Chr.11: 65386917-65386917 on Build GRCh38
rs144532045 Chr.11: 65387150-65387150 on Build GRCh38
rs906636034 Chr.11: 65387358-65387358 on Build GRCh38
rs754207805 Chr.11: 65387187-65387187 on Build GRCh38
rs892620722 Chr.11: 65387286-65387286 on Build GRCh38
rs765378520 Chr.11: 65387266-65387266 on Build GRCh38
rs1038374589 Chr.11: 65387312-65387312 on Build GRCh38
rs752740201 Chr.11: 65387107-65387108 on Build GRCh38
rs945557258 Chr.11: 65387295-65387295 on Build GRCh38
rs188855238 Chr.11: 65387194-65387194 on Build GRCh38
rs778792189 Chr.11: 65387028-65387028 on Build GRCh38
rs771493477 Chr.11: 65387108-65387108 on Build GRCh38
rs1046499731 Chr.11: 65387341-65387341 on Build GRCh38
rs567890124 Chr.11: 65387117-65387117 on Build GRCh38
rs533580385 Chr.11: 65386951-65386951 on Build GRCh38
rs539324067 Chr.11: 65387264-65387265 on Build GRCh38
rs765171186 Chr.11: 65387105-65387106 on Build GRCh38
rs773463027 Chr.11: 65387053-65387053 on Build GRCh38
rs973901350 Chr.11: 65387114-65387114 on Build GRCh38
rs553748558 Chr.11: 65387368-65387368 on Build GRCh38
rs529596076 Chr.11: 65387070-65387070 on Build GRCh38
rs753303472 Chr.11: 65387196-65387196 on Build GRCh38
rs144310109 Chr.11: 65387090-65387090 on Build GRCh38
rs951160357 Chr.11: 65387032-65387032 on Build GRCh38
rs78928443 Chr.11: 65387232-65387232 on Build GRCh38
rs914382067 Chr.11: 65387178-65387178 on Build GRCh38
rs752784150 Chr.11: 65387360-65387360 on Build GRCh38
rs772141355 Chr.11: 65387274-65387274 on Build GRCh38
rs759320847 Chr.11: 65387056-65387056 on Build GRCh38
rs569323385 Chr.11: 65387092-65387092 on Build GRCh38
rs773893329 Chr.11: 65386983-65386983 on Build GRCh38
rs546928867 Chr.11: 65386955-65386955 on Build GRCh38
rs749008914 Chr.11: 65387225-65387225 on Build GRCh38
rs200896880 Chr.11: 65386997-65386997 on Build GRCh38
rs151230902 Chr.11: 65387220-65387220 on Build GRCh38
rs749329031 Chr.11: 65387119-65387119 on Build GRCh38
rs764119670 Chr.11: 65387069-65387069 on Build GRCh38
rs951278329 Chr.11: 65387192-65387192 on Build GRCh38
rs749869924 Chr.11: 65386998-65386998 on Build GRCh38
rs749968187 Chr.11: 65387171-65387171 on Build GRCh38
rs761227112 Chr.11: 65387156-65387156 on Build GRCh38
rs78688868 Chr.11: 65387089-65387089 on Build GRCh38
rs779283301 Chr.11: 65387195-65387195 on Build GRCh38
rs771144414 Chr.11: 65387132-65387132 on Build GRCh38
rs748063813 Chr.11: 65387254-65387254 on Build GRCh38
rs767196748 Chr.11: 65387165-65387165 on Build GRCh38
rs768459283 Chr.11: 65387149-65387149 on Build GRCh38
rs772808462 Chr.11: 65387280-65387280 on Build GRCh38

View Full Product Details