Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65894028-65894573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOSL1
Entrez Gene ID
8061
Gene Name
FOS like 1, AP-1 transcription factor subunit
Gene Aliases
FRA, FRA1, fra-1
UniGene
Hs.283565
Gene Chromosome Location
Chr.11: 65892136-65900526 on Build GRCh38
Transcripts
NM_001300844.1 NM_001300855.1 NM_001300856.1 NM_001300857.1 NM_005438.4 NR_125339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2166693 c.320G>A p.R107H Chr.11: 65894099-65894099 on Build GRCh38
7259183 c.319C>T p.R107C Chr.11: 65894100-65894100 on Build GRCh38
6666032 c.368G>T p.R123M Chr.11: 65894051-65894051 on Build GRCh38
1561658 c.309G>A p.E103E Chr.11: 65894110-65894110 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767082299 Chr.11: 65894113-65894113 on Build GRCh38
rs911534757 Chr.11: 65894415-65894415 on Build GRCh38
rs943027000 Chr.11: 65894427-65894427 on Build GRCh38
rs776577128 Chr.11: 65894147-65894147 on Build GRCh38
rs73494422 Chr.11: 65894531-65894531 on Build GRCh38
rs891747598 Chr.11: 65894253-65894253 on Build GRCh38
rs757350956 Chr.11: 65894496-65894496 on Build GRCh38
rs751184760 Chr.11: 65894103-65894103 on Build GRCh38
rs368497099 Chr.11: 65894093-65894093 on Build GRCh38
rs768932313 Chr.11: 65894134-65894134 on Build GRCh38
rs963495957 Chr.11: 65894342-65894342 on Build GRCh38
rs1029044942 Chr.11: 65894365-65894365 on Build GRCh38
rs1029992287 Chr.11: 65894426-65894426 on Build GRCh38
rs976195070 Chr.11: 65894397-65894397 on Build GRCh38
rs953276696 Chr.11: 65894452-65894452 on Build GRCh38
rs373111405 Chr.11: 65894330-65894330 on Build GRCh38
rs781777272 Chr.11: 65894114-65894114 on Build GRCh38
rs551212950 Chr.11: 65894056-65894056 on Build GRCh38
rs752022395 Chr.11: 65894467-65894467 on Build GRCh38
rs970723330 Chr.11: 65894316-65894316 on Build GRCh38
rs905314560 Chr.11: 65894317-65894317 on Build GRCh38
rs949658824 Chr.11: 65894543-65894543 on Build GRCh38
rs1038786773 Chr.11: 65894451-65894451 on Build GRCh38
rs770821487 Chr.11: 65894139-65894139 on Build GRCh38
rs370021821 Chr.11: 65894318-65894318 on Build GRCh38
rs961742165 Chr.11: 65894394-65894394 on Build GRCh38
rs769884705 Chr.11: 65894075-65894075 on Build GRCh38
rs546654738 Chr.11: 65894236-65894236 on Build GRCh38
rs973636323 Chr.11: 65894361-65894361 on Build GRCh38
rs951971277 Chr.11: 65894099-65894099 on Build GRCh38
rs752420600 Chr.11: 65894097-65894097 on Build GRCh38
rs950961463 Chr.11: 65894371-65894371 on Build GRCh38
rs531114435 Chr.11: 65894053-65894053 on Build GRCh38
rs747527827 Chr.11: 65894059-65894059 on Build GRCh38
rs1046860568 Chr.11: 65894547-65894547 on Build GRCh38
rs1045637857 Chr.11: 65894529-65894529 on Build GRCh38
rs748274482 Chr.11: 65894137-65894137 on Build GRCh38
rs922976744 Chr.11: 65894210-65894210 on Build GRCh38
rs554738036 Chr.11: 65894506-65894506 on Build GRCh38
rs181979051 Chr.11: 65894275-65894275 on Build GRCh38
rs986293685 Chr.11: 65894227-65894227 on Build GRCh38
rs570756562 Chr.11: 65894096-65894096 on Build GRCh38
rs570150007 Chr.11: 65894331-65894331 on Build GRCh38
rs892479402 Chr.11: 65894283-65894283 on Build GRCh38
rs961987960 Chr.11: 65894166-65894166 on Build GRCh38
rs931750565 Chr.11: 65894219-65894219 on Build GRCh38
rs149112693 Chr.11: 65894083-65894083 on Build GRCh38
rs904326177 Chr.11: 65894228-65894228 on Build GRCh38
rs1038819447 Chr.11: 65894227-65894227 on Build GRCh38
rs557997837 Chr.11: 65894364-65894364 on Build GRCh38
rs144980043 Chr.11: 65894078-65894078 on Build GRCh38
rs890132608 Chr.11: 65894181-65894181 on Build GRCh38
rs746291005 Chr.11: 65894072-65894072 on Build GRCh38
rs1014816559 Chr.11: 65894376-65894376 on Build GRCh38
rs371886513 Chr.11: 65894094-65894094 on Build GRCh38
rs781685367 Chr.11: 65894069-65894069 on Build GRCh38
rs1052925893 Chr.11: 65894511-65894511 on Build GRCh38
rs780080576 Chr.11: 65894125-65894125 on Build GRCh38
rs980794725 Chr.11: 65894321-65894321 on Build GRCh38
rs771352442 Chr.11: 65894061-65894061 on Build GRCh38
rs190628862 Chr.11: 65894238-65894238 on Build GRCh38
rs1046477021 Chr.11: 65894298-65894298 on Build GRCh38
rs936947286 Chr.11: 65894240-65894240 on Build GRCh38
rs374499277 Chr.11: 65894079-65894079 on Build GRCh38
rs781194934 Chr.11: 65894517-65894517 on Build GRCh38
rs749528902 Chr.11: 65894128-65894128 on Build GRCh38
rs753059614 Chr.11: 65894284-65894284 on Build GRCh38
rs891627667 Chr.11: 65894524-65894524 on Build GRCh38
rs996241628 Chr.11: 65894230-65894230 on Build GRCh38
rs184519609 Chr.11: 65894100-65894100 on Build GRCh38
rs1051472988 Chr.11: 65894177-65894177 on Build GRCh38
rs507062 Chr.11: 65894185-65894185 on Build GRCh38
rs759198105 Chr.11: 65894149-65894149 on Build GRCh38
rs988360318 Chr.11: 65894527-65894527 on Build GRCh38
rs373742155 Chr.11: 65894163-65894163 on Build GRCh38
rs73494420 Chr.11: 65894282-65894282 on Build GRCh38
rs758770068 Chr.11: 65894319-65894319 on Build GRCh38
rs1034208741 Chr.11: 65894324-65894324 on Build GRCh38
rs766004966 Chr.11: 65894156-65894156 on Build GRCh38
rs760600411 Chr.11: 65894087-65894087 on Build GRCh38
rs774398775 Chr.11: 65894136-65894136 on Build GRCh38
rs773068029 Chr.11: 65894085-65894085 on Build GRCh38

View Full Product Details