Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65896432-65896968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOSL1
Entrez Gene ID
8061
Gene Name
FOS like 1, AP-1 transcription factor subunit
Gene Aliases
FRA, FRA1, fra-1
UniGene
Hs.283565
Gene Chromosome Location
Chr.11: 65892136-65900526 on Build GRCh38
Transcripts
NM_001300844.1 NM_001300855.1 NM_001300856.1 NM_001300857.1 NM_005438.4 NR_125339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298437 c.251G>T p.R84L Chr.11: 65896855-65896855 on Build GRCh38
6337796 c.295C>A p.Q99K Chr.11: 65896811-65896811 on Build GRCh38
1285032 c.259G>T p.G87W Chr.11: 65896847-65896847 on Build GRCh38
930549 c.235C>T p.R79W Chr.11: 65896871-65896871 on Build GRCh38
3935582 c.162A>C p.V54V Chr.11: 65896944-65896944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142205529 Chr.11: 65896832-65896832 on Build GRCh38
rs756869534 Chr.11: 65896775-65896775 on Build GRCh38
rs927834482 Chr.11: 65896465-65896465 on Build GRCh38
rs369289692 Chr.11: 65896856-65896856 on Build GRCh38
rs918371041 Chr.11: 65896667-65896667 on Build GRCh38
rs145383050 Chr.11: 65896477-65896477 on Build GRCh38
rs748182697 Chr.11: 65896826-65896826 on Build GRCh38
rs1002948107 Chr.11: 65896757-65896757 on Build GRCh38
rs1041718331 Chr.11: 65896739-65896739 on Build GRCh38
rs991654454 Chr.11: 65896925-65896925 on Build GRCh38
rs201410689 Chr.11: 65896828-65896828 on Build GRCh38
rs750176184 Chr.11: 65896878-65896879 on Build GRCh38
rs1034853230 Chr.11: 65896626-65896626 on Build GRCh38
rs766841945 Chr.11: 65896858-65896858 on Build GRCh38
rs772755652 Chr.11: 65896947-65896947 on Build GRCh38
rs893144620 Chr.11: 65896717-65896717 on Build GRCh38
rs1039902836 Chr.11: 65896713-65896713 on Build GRCh38
rs77537829 Chr.11: 65896526-65896526 on Build GRCh38
rs1028272252 Chr.11: 65896621-65896621 on Build GRCh38
rs769751521 Chr.11: 65896906-65896906 on Build GRCh38
rs903303352 Chr.11: 65896754-65896754 on Build GRCh38
rs893113603 Chr.11: 65896557-65896557 on Build GRCh38
rs180875695 Chr.11: 65896679-65896679 on Build GRCh38
rs774090243 Chr.11: 65896930-65896930 on Build GRCh38
rs767141115 Chr.11: 65896848-65896848 on Build GRCh38
rs571532302 Chr.11: 65896746-65896746 on Build GRCh38
rs946202267 Chr.11: 65896485-65896485 on Build GRCh38
rs755168615 Chr.11: 65896820-65896820 on Build GRCh38
rs960642671 Chr.11: 65896629-65896629 on Build GRCh38
rs1004378719 Chr.11: 65896547-65896547 on Build GRCh38
rs199745694 Chr.11: 65896780-65896780 on Build GRCh38
rs763900378 Chr.11: 65896773-65896773 on Build GRCh38
rs368840880 Chr.11: 65896937-65896937 on Build GRCh38
rs775345295 Chr.11: 65896838-65896838 on Build GRCh38
rs149444153 Chr.11: 65896910-65896910 on Build GRCh38
rs771678720 Chr.11: 65896945-65896945 on Build GRCh38
rs151147129 Chr.11: 65896842-65896842 on Build GRCh38
rs967452990 Chr.11: 65896656-65896656 on Build GRCh38
rs538737006 Chr.11: 65896703-65896703 on Build GRCh38
rs1023324748 Chr.11: 65896642-65896642 on Build GRCh38
rs780461891 Chr.11: 65896802-65896802 on Build GRCh38
rs750962628 Chr.11: 65896779-65896779 on Build GRCh38
rs757505120 Chr.11: 65896784-65896784 on Build GRCh38
rs747120128 Chr.11: 65896880-65896880 on Build GRCh38
rs637571 Chr.11: 65896875-65896875 on Build GRCh38
rs191333882 Chr.11: 65896631-65896631 on Build GRCh38
rs962343033 Chr.11: 65896652-65896652 on Build GRCh38
rs754007101 Chr.11: 65896475-65896475 on Build GRCh38
rs749672069 Chr.11: 65896816-65896816 on Build GRCh38
rs541351441 Chr.11: 65896476-65896476 on Build GRCh38
rs990205564 Chr.11: 65896650-65896650 on Build GRCh38
rs867402581 Chr.11: 65896932-65896932 on Build GRCh38
rs987402283 Chr.11: 65896702-65896702 on Build GRCh38
rs947657834 Chr.11: 65896871-65896871 on Build GRCh38
rs745825650 Chr.11: 65896829-65896829 on Build GRCh38
rs374811980 Chr.11: 65896843-65896843 on Build GRCh38
rs973852270 Chr.11: 65896655-65896655 on Build GRCh38
rs746916939 Chr.11: 65896568-65896568 on Build GRCh38
rs114984248 Chr.11: 65896732-65896732 on Build GRCh38
rs190968425 Chr.11: 65896774-65896774 on Build GRCh38
rs745369622 Chr.11: 65896839-65896839 on Build GRCh38
rs142924409 Chr.11: 65896515-65896515 on Build GRCh38
rs570102574 Chr.11: 65896798-65896798 on Build GRCh38
rs900992963 Chr.11: 65896776-65896776 on Build GRCh38
rs960612956 Chr.11: 65896470-65896470 on Build GRCh38
rs614520 Chr.11: 65896535-65896535 on Build GRCh38
rs1038505179 Chr.11: 65896499-65896499 on Build GRCh38
rs942972015 Chr.11: 65896706-65896706 on Build GRCh38
rs902349667 Chr.11: 65896603-65896603 on Build GRCh38
rs754196763 Chr.11: 65896861-65896861 on Build GRCh38
rs779143676 Chr.11: 65896825-65896825 on Build GRCh38
rs999806986 Chr.11: 65896606-65896606 on Build GRCh38
rs186058454 Chr.11: 65896772-65896772 on Build GRCh38
rs138397091 Chr.11: 65896512-65896512 on Build GRCh38
rs577994281 Chr.11: 65896500-65896500 on Build GRCh38
rs977476933 Chr.11: 65896664-65896664 on Build GRCh38
rs547591812 Chr.11: 65896524-65896524 on Build GRCh38
rs774942711 Chr.11: 65896763-65896763 on Build GRCh38
rs1011627580 Chr.11: 65896567-65896567 on Build GRCh38
rs779055210 Chr.11: 65896868-65896868 on Build GRCh38
rs768594766 Chr.11: 65896927-65896927 on Build GRCh38
rs768098467 Chr.11: 65896777-65896777 on Build GRCh38
rs974178466 Chr.11: 65896817-65896817 on Build GRCh38
rs576321480 Chr.11: 65896866-65896866 on Build GRCh38
rs749967518 Chr.11: 65896855-65896855 on Build GRCh38
rs758410472 Chr.11: 65896879-65896879 on Build GRCh38

View Full Product Details