Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65997524-65998049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AD
Entrez Gene ID
84285
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
haponin
UniGene
Hs.425178
Gene Chromosome Location
Chr.11: 65996545-66002217 on Build GRCh38
Transcripts
NM_001242481.1 NM_001242482.1 NM_001242483.1 NM_001242484.1 NM_001242485.1 NM_001242486.1 NM_032325.3 XM_017018412.1

SNP Detail

SNP ID Chromosome Location
rs188324805 Chr.11: 65998008-65998008 on Build GRCh38
rs944596808 Chr.11: 65997994-65997994 on Build GRCh38
rs920157685 Chr.11: 65997682-65997682 on Build GRCh38
rs569407997 Chr.11: 65997884-65997884 on Build GRCh38
rs908547152 Chr.11: 65997929-65997929 on Build GRCh38
rs556027745 Chr.11: 65997867-65997867 on Build GRCh38
rs996915341 Chr.11: 65997695-65997695 on Build GRCh38
rs927645102 Chr.11: 65997883-65997883 on Build GRCh38
rs751481879 Chr.11: 65997734-65997735 on Build GRCh38
rs930478329 Chr.11: 65997951-65997951 on Build GRCh38
rs183944683 Chr.11: 65997581-65997581 on Build GRCh38
rs920347166 Chr.11: 65997944-65997948 on Build GRCh38
rs527767594 Chr.11: 65997712-65997712 on Build GRCh38
rs1000767201 Chr.11: 65997760-65997760 on Build GRCh38
rs981788404 Chr.11: 65997866-65997866 on Build GRCh38
rs969556902 Chr.11: 65997832-65997832 on Build GRCh38
rs991843588 Chr.11: 65997604-65997604 on Build GRCh38
rs1024670124 Chr.11: 65997801-65997801 on Build GRCh38
rs925113833 Chr.11: 65998021-65998021 on Build GRCh38
rs114146217 Chr.11: 65997755-65997755 on Build GRCh38
rs937644715 Chr.11: 65997925-65997925 on Build GRCh38
rs969523585 Chr.11: 65997650-65997650 on Build GRCh38
rs901205266 Chr.11: 65998017-65998018 on Build GRCh38
rs981078212 Chr.11: 65997651-65997651 on Build GRCh38
rs907800460 Chr.11: 65997984-65997984 on Build GRCh38
rs754923148 Chr.11: 65997720-65997720 on Build GRCh38
rs903553208 Chr.11: 65997549-65997549 on Build GRCh38
rs12795607 Chr.11: 65997574-65997574 on Build GRCh38
rs976868444 Chr.11: 65997958-65997958 on Build GRCh38
rs1048140584 Chr.11: 65997602-65997602 on Build GRCh38
rs771612780 Chr.11: 65997963-65997963 on Build GRCh38
rs778883708 Chr.11: 65997886-65997886 on Build GRCh38
rs983020159 Chr.11: 65997915-65997915 on Build GRCh38
rs903752931 Chr.11: 65997756-65997756 on Build GRCh38
rs571408803 Chr.11: 65997613-65997613 on Build GRCh38
rs12796959 Chr.11: 65997592-65997592 on Build GRCh38
rs931796158 Chr.11: 65997752-65997752 on Build GRCh38
rs1019549012 Chr.11: 65997647-65997647 on Build GRCh38
rs983205527 Chr.11: 65997702-65997702 on Build GRCh38
rs577743413 Chr.11: 65998016-65998016 on Build GRCh38
rs527357109 Chr.11: 65997714-65997714 on Build GRCh38
rs531342881 Chr.11: 65997595-65997595 on Build GRCh38
rs12795055 Chr.11: 65997580-65997580 on Build GRCh38
rs1027130957 Chr.11: 65997853-65997853 on Build GRCh38
rs149463234 Chr.11: 65997935-65997935 on Build GRCh38
rs892099813 Chr.11: 65997772-65997772 on Build GRCh38
rs931351724 Chr.11: 65997687-65997687 on Build GRCh38
rs1002763637 Chr.11: 65997839-65997839 on Build GRCh38
rs901716296 Chr.11: 65997745-65997745 on Build GRCh38
rs12795592 Chr.11: 65997554-65997554 on Build GRCh38
rs547246843 Chr.11: 65997728-65997728 on Build GRCh38
rs1004170999 Chr.11: 65997606-65997606 on Build GRCh38
rs931858745 Chr.11: 65997973-65997973 on Build GRCh38
rs376146940 Chr.11: 65997671-65997671 on Build GRCh38
rs1013326031 Chr.11: 65997786-65997786 on Build GRCh38
rs1041112213 Chr.11: 65997725-65997725 on Build GRCh38
rs552416641 Chr.11: 65997835-65997835 on Build GRCh38
rs12796976 Chr.11: 65997609-65997609 on Build GRCh38
rs72930942 Chr.11: 65997603-65997603 on Build GRCh38
rs535644647 Chr.11: 65997770-65997770 on Build GRCh38
rs1052546084 Chr.11: 65997764-65997764 on Build GRCh38

View Full Product Details