Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65999267-65999769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AD
Entrez Gene ID
84285
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
haponin
UniGene
Hs.425178
Gene Chromosome Location
Chr.11: 65996545-66002217 on Build GRCh38
Transcripts
NM_001242481.1 NM_001242482.1 NM_001242483.1 NM_001242484.1 NM_001242485.1 NM_001242486.1 NM_032325.3 XM_017018412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429249 c.328G>T p.A110S Chr.11: 65999375-65999375 on Build GRCh38
7429248 c.329C>T p.A110V Chr.11: 65999374-65999374 on Build GRCh38
5625081 c.247G>A p.E83K Chr.11: 65999625-65999625 on Build GRCh38
4035604 c.221T>C p.I74T Chr.11: 65999651-65999651 on Build GRCh38
4135699 c.225A>T p.E75D Chr.11: 65999647-65999647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897094654 Chr.11: 65999557-65999557 on Build GRCh38
rs774204392 Chr.11: 65999572-65999572 on Build GRCh38
rs781160197 Chr.11: 65999417-65999418 on Build GRCh38
rs1018423769 Chr.11: 65999635-65999635 on Build GRCh38
rs773530732 Chr.11: 65999335-65999335 on Build GRCh38
rs759514781 Chr.11: 65999563-65999563 on Build GRCh38
rs778490889 Chr.11: 65999623-65999623 on Build GRCh38
rs745548551 Chr.11: 65999303-65999303 on Build GRCh38
rs766368387 Chr.11: 65999307-65999307 on Build GRCh38
rs200017157 Chr.11: 65999322-65999322 on Build GRCh38
rs957590080 Chr.11: 65999655-65999655 on Build GRCh38
rs780803779 Chr.11: 65999300-65999300 on Build GRCh38
rs200433192 Chr.11: 65999312-65999312 on Build GRCh38
rs506873 Chr.11: 65999744-65999744 on Build GRCh38
rs868517190 Chr.11: 65999355-65999355 on Build GRCh38
rs778406838 Chr.11: 65999721-65999721 on Build GRCh38
rs772585620 Chr.11: 65999329-65999329 on Build GRCh38
rs748698610 Chr.11: 65999428-65999428 on Build GRCh38
rs377762380 Chr.11: 65999592-65999592 on Build GRCh38
rs763083958 Chr.11: 65999668-65999668 on Build GRCh38
rs932988570 Chr.11: 65999749-65999749 on Build GRCh38
rs946303808 Chr.11: 65999713-65999713 on Build GRCh38
rs960732747 Chr.11: 65999574-65999574 on Build GRCh38
rs201148001 Chr.11: 65999591-65999591 on Build GRCh38
rs895016087 Chr.11: 65999516-65999516 on Build GRCh38
rs748716615 Chr.11: 65999409-65999409 on Build GRCh38
rs779823158 Chr.11: 65999402-65999402 on Build GRCh38
rs771380477 Chr.11: 65999634-65999634 on Build GRCh38
rs909393218 Chr.11: 65999673-65999673 on Build GRCh38
rs773725257 Chr.11: 65999432-65999432 on Build GRCh38
rs994048294 Chr.11: 65999590-65999590 on Build GRCh38
rs778490397 Chr.11: 65999617-65999617 on Build GRCh38
rs1035605142 Chr.11: 65999505-65999505 on Build GRCh38
rs757432033 Chr.11: 65999357-65999359 on Build GRCh38
rs565280494 Chr.11: 65999506-65999506 on Build GRCh38
rs775530793 Chr.11: 65999656-65999656 on Build GRCh38
rs985580870 Chr.11: 65999315-65999315 on Build GRCh38
rs751567674 Chr.11: 65999367-65999367 on Build GRCh38
rs373731888 Chr.11: 65999696-65999696 on Build GRCh38
rs754405462 Chr.11: 65999609-65999609 on Build GRCh38
rs531792509 Chr.11: 65999463-65999463 on Build GRCh38
rs761749453 Chr.11: 65999580-65999580 on Build GRCh38
rs368224000 Chr.11: 65999697-65999697 on Build GRCh38
rs201623056 Chr.11: 65999618-65999618 on Build GRCh38
rs1016750454 Chr.11: 65999554-65999554 on Build GRCh38
rs772499909 Chr.11: 65999430-65999430 on Build GRCh38
rs747437537 Chr.11: 65999448-65999448 on Build GRCh38
rs527977105 Chr.11: 65999545-65999545 on Build GRCh38
rs771057293 Chr.11: 65999523-65999523 on Build GRCh38
rs752251894 Chr.11: 65999717-65999717 on Build GRCh38
rs745893608 Chr.11: 65999581-65999582 on Build GRCh38
rs1005776874 Chr.11: 65999548-65999548 on Build GRCh38
rs755943069 Chr.11: 65999593-65999593 on Build GRCh38
rs937448764 Chr.11: 65999494-65999494 on Build GRCh38
rs377315522 Chr.11: 65999308-65999308 on Build GRCh38
rs779238205 Chr.11: 65999426-65999426 on Build GRCh38
rs760984235 Chr.11: 65999337-65999337 on Build GRCh38
rs774962491 Chr.11: 65999321-65999321 on Build GRCh38
rs766510565 Chr.11: 65999341-65999341 on Build GRCh38
rs753548811 Chr.11: 65999607-65999607 on Build GRCh38
rs757003583 Chr.11: 65999387-65999387 on Build GRCh38
rs760336439 Chr.11: 65999674-65999674 on Build GRCh38
rs199855372 Chr.11: 65999392-65999392 on Build GRCh38
rs776671323 Chr.11: 65999645-65999645 on Build GRCh38
rs551520901 Chr.11: 65999478-65999478 on Build GRCh38
rs148924410 Chr.11: 65999358-65999358 on Build GRCh38
rs145743811 Chr.11: 65999595-65999595 on Build GRCh38
rs909479102 Chr.11: 65999292-65999292 on Build GRCh38
rs755746711 Chr.11: 65999400-65999400 on Build GRCh38
rs747349944 Chr.11: 65999624-65999624 on Build GRCh38
rs759735712 Chr.11: 65999412-65999412 on Build GRCh38
rs759732408 Chr.11: 65999353-65999353 on Build GRCh38
rs776956968 Chr.11: 65999539-65999539 on Build GRCh38
rs370139198 Chr.11: 65999566-65999566 on Build GRCh38
rs567565897 Chr.11: 65999646-65999646 on Build GRCh38
rs767410671 Chr.11: 65999672-65999672 on Build GRCh38
rs507672 Chr.11: 65999682-65999682 on Build GRCh38
rs769702496 Chr.11: 65999654-65999654 on Build GRCh38
rs543169917 Chr.11: 65999328-65999328 on Build GRCh38
rs967225521 Chr.11: 65999467-65999467 on Build GRCh38
rs985267902 Chr.11: 65999596-65999596 on Build GRCh38
rs768260350 Chr.11: 65999410-65999410 on Build GRCh38
rs750016208 Chr.11: 65999394-65999394 on Build GRCh38
rs758026148 Chr.11: 65999718-65999718 on Build GRCh38
rs756168708 Chr.11: 65999684-65999686 on Build GRCh38
rs550007641 Chr.11: 65999740-65999740 on Build GRCh38
rs748883223 Chr.11: 65999309-65999309 on Build GRCh38
rs765041085 Chr.11: 65999704-65999704 on Build GRCh38

View Full Product Details