Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66003535-66004028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANF1
Entrez Gene ID
8815
Gene Name
barrier to autointegration factor 1
Gene Aliases
BAF, BCRP1, D14S1460, NGPS
UniGene
Hs.433759
Gene Chromosome Location
Chr.11: 66002079-66004149 on Build GRCh38
Transcripts
NM_001143985.1 NM_003860.3 XM_017018514.1 XM_017018515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5045598 c.211G>A p.A71T Chr.11: 66003713-66003713 on Build GRCh38
4035606 c.124G>A p.A42T Chr.11: 66003626-66003626 on Build GRCh38
259596 c.244C>T p.R82* Chr.11: 66003746-66003746 on Build GRCh38
238940 c.143A>T p.Q48L Chr.11: 66003645-66003645 on Build GRCh38
930599 c.256G>A p.D86N Chr.11: 66003758-66003758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377026745 Chr.11: 66003616-66003616 on Build GRCh38
rs957958312 Chr.11: 66003851-66003851 on Build GRCh38
rs756893410 Chr.11: 66003682-66003682 on Build GRCh38
rs1048566686 Chr.11: 66003618-66003618 on Build GRCh38
rs535755968 Chr.11: 66003884-66003884 on Build GRCh38
rs751354982 Chr.11: 66003798-66003798 on Build GRCh38
rs901194846 Chr.11: 66004003-66004003 on Build GRCh38
rs535232397 Chr.11: 66003918-66003918 on Build GRCh38
rs983367159 Chr.11: 66003866-66003866 on Build GRCh38
rs181035066 Chr.11: 66003807-66003807 on Build GRCh38
rs374456907 Chr.11: 66003712-66003712 on Build GRCh38
rs765062850 Chr.11: 66003665-66003665 on Build GRCh38
rs970781709 Chr.11: 66003885-66003885 on Build GRCh38
rs140412018 Chr.11: 66003717-66003717 on Build GRCh38
rs940318848 Chr.11: 66003984-66003984 on Build GRCh38
rs888718501 Chr.11: 66003637-66003637 on Build GRCh38
rs763175257 Chr.11: 66003609-66003609 on Build GRCh38
rs1032098068 Chr.11: 66003850-66003850 on Build GRCh38
rs908582802 Chr.11: 66003575-66003576 on Build GRCh38
rs757441761 Chr.11: 66003591-66003594 on Build GRCh38
rs376639113 Chr.11: 66003589-66003589 on Build GRCh38
rs750526063 Chr.11: 66003621-66003621 on Build GRCh38
rs780893705 Chr.11: 66003685-66003685 on Build GRCh38
rs902739068 Chr.11: 66003983-66003983 on Build GRCh38
rs745445839 Chr.11: 66003601-66003601 on Build GRCh38
rs370432107 Chr.11: 66003667-66003667 on Build GRCh38
rs56984820 Chr.11: 66003589-66003595 on Build GRCh38
rs144367403 Chr.11: 66003900-66003900 on Build GRCh38
rs766514105 Chr.11: 66003776-66003776 on Build GRCh38
rs757087596 Chr.11: 66003802-66003802 on Build GRCh38
rs564929774 Chr.11: 66003711-66003711 on Build GRCh38
rs767756912 Chr.11: 66003674-66003675 on Build GRCh38
rs762964076 Chr.11: 66003779-66003779 on Build GRCh38
rs760849477 Chr.11: 66003630-66003630 on Build GRCh38
rs763434920 Chr.11: 66003585-66003588 on Build GRCh38
rs140057395 Chr.11: 66003706-66003706 on Build GRCh38
rs983316418 Chr.11: 66003574-66003574 on Build GRCh38
rs988457519 Chr.11: 66003860-66003860 on Build GRCh38
rs984289822 Chr.11: 66003957-66003957 on Build GRCh38
rs772391821 Chr.11: 66003737-66003737 on Build GRCh38
rs748804760 Chr.11: 66003818-66003818 on Build GRCh38
rs745344998 Chr.11: 66003690-66003690 on Build GRCh38
rs546873462 Chr.11: 66003864-66003864 on Build GRCh38
rs191905461 Chr.11: 66003560-66003560 on Build GRCh38
rs542636194 Chr.11: 66003581-66003581 on Build GRCh38
rs748590381 Chr.11: 66003726-66003726 on Build GRCh38
rs751909975 Chr.11: 66003587-66003589 on Build GRCh38
rs143413856 Chr.11: 66003673-66003673 on Build GRCh38
rs764079436 Chr.11: 66003786-66003786 on Build GRCh38
rs886048517 Chr.11: 66003836-66003836 on Build GRCh38
rs779626661 Chr.11: 66003817-66003817 on Build GRCh38
rs910016747 Chr.11: 66003965-66003965 on Build GRCh38
rs758089950 Chr.11: 66003670-66003670 on Build GRCh38
rs774841174 Chr.11: 66003725-66003725 on Build GRCh38
rs779603619 Chr.11: 66003822-66003822 on Build GRCh38
rs1037686396 Chr.11: 66003991-66003991 on Build GRCh38
rs775225097 Chr.11: 66003777-66003777 on Build GRCh38
rs537800431 Chr.11: 66003956-66003956 on Build GRCh38
rs148797275 Chr.11: 66003949-66003949 on Build GRCh38
rs926374176 Chr.11: 66003948-66003948 on Build GRCh38
rs147010511 Chr.11: 66003580-66003586 on Build GRCh38
rs762137248 Chr.11: 66003615-66003615 on Build GRCh38
rs998317304 Chr.11: 66003700-66003700 on Build GRCh38
rs760622680 Chr.11: 66003763-66003763 on Build GRCh38
rs773533777 Chr.11: 66003757-66003757 on Build GRCh38
rs201016838 Chr.11: 66003607-66003607 on Build GRCh38
rs750117956 Chr.11: 66003808-66003808 on Build GRCh38
rs913525858 Chr.11: 66003561-66003561 on Build GRCh38
rs1037043092 Chr.11: 66003676-66003676 on Build GRCh38
rs145574018 Chr.11: 66003872-66003872 on Build GRCh38
rs764369557 Chr.11: 66003586-66003593 on Build GRCh38
rs758262437 Chr.11: 66003575-66003575 on Build GRCh38
rs901160979 Chr.11: 66003681-66003681 on Build GRCh38
rs886048518 Chr.11: 66003892-66003892 on Build GRCh38
rs932855686 Chr.11: 66003588-66003588 on Build GRCh38
rs755536616 Chr.11: 66003821-66003821 on Build GRCh38
rs770200889 Chr.11: 66003603-66003603 on Build GRCh38
rs961404591 Chr.11: 66003883-66003883 on Build GRCh38
rs755509046 Chr.11: 66003816-66003816 on Build GRCh38
rs746696782 Chr.11: 66003577-66003577 on Build GRCh38
rs769171254 Chr.11: 66003610-66003610 on Build GRCh38
rs372591003 Chr.11: 66003613-66003613 on Build GRCh38
rs1049836920 Chr.11: 66003942-66003942 on Build GRCh38
rs953240419 Chr.11: 66003724-66003724 on Build GRCh38
rs780695833 Chr.11: 66003599-66003599 on Build GRCh38
rs763883395 Chr.11: 66003671-66003671 on Build GRCh38
rs775972466 Chr.11: 66003581-66003583 on Build GRCh38
rs886048519 Chr.11: 66003981-66003981 on Build GRCh38

View Full Product Details