Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66016887-66017398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER1
Entrez Gene ID
117144
Gene Name
cation channel sperm associated 1
Gene Aliases
CATSPER, SPGF7
UniGene
Hs.189105
Gene Chromosome Location
Chr.11: 66016752-66026518 on Build GRCh38
Transcripts
NM_053054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930601 c.2302G>A p.D768N Chr.11: 66017074-66017074 on Build GRCh38
1638925 c.2268C>A p.F756L Chr.11: 66017108-66017108 on Build GRCh38
6194340 c.2214C>T p.L738L Chr.11: 66017162-66017162 on Build GRCh38
159764 c.2317-1G>A p.? Chr.11: 66016917-66016917 on Build GRCh38
4445555 c.2316+3G>A p.? Chr.11: 66017057-66017057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772232447 Chr.11: 66017034-66017034 on Build GRCh38
rs775418332 Chr.11: 66017064-66017064 on Build GRCh38
rs763398904 Chr.11: 66017178-66017179 on Build GRCh38
rs774173209 Chr.11: 66017195-66017195 on Build GRCh38
rs763676594 Chr.11: 66016937-66016937 on Build GRCh38
rs529217754 Chr.11: 66016957-66016957 on Build GRCh38
rs756678169 Chr.11: 66016946-66016946 on Build GRCh38
rs777914787 Chr.11: 66017110-66017110 on Build GRCh38
rs748502205 Chr.11: 66017106-66017106 on Build GRCh38
rs765910925 Chr.11: 66017179-66017179 on Build GRCh38
rs577788922 Chr.11: 66017309-66017309 on Build GRCh38
rs748425480 Chr.11: 66017033-66017033 on Build GRCh38
rs375479637 Chr.11: 66017187-66017187 on Build GRCh38
rs147304194 Chr.11: 66017107-66017107 on Build GRCh38
rs930223523 Chr.11: 66017321-66017321 on Build GRCh38
rs1039812641 Chr.11: 66017352-66017352 on Build GRCh38
rs148760734 Chr.11: 66017118-66017118 on Build GRCh38
rs368765589 Chr.11: 66017210-66017210 on Build GRCh38
rs749945244 Chr.11: 66017012-66017012 on Build GRCh38
rs934077214 Chr.11: 66017231-66017231 on Build GRCh38
rs35358803 Chr.11: 66017213-66017214 on Build GRCh38
rs915138993 Chr.11: 66016943-66016943 on Build GRCh38
rs35057440 Chr.11: 66017148-66017149 on Build GRCh38
rs780132812 Chr.11: 66017337-66017337 on Build GRCh38
rs1043861460 Chr.11: 66017267-66017267 on Build GRCh38
rs889972688 Chr.11: 66017242-66017242 on Build GRCh38
rs369677297 Chr.11: 66017191-66017191 on Build GRCh38
rs753092816 Chr.11: 66017180-66017180 on Build GRCh38
rs377188056 Chr.11: 66017042-66017042 on Build GRCh38
rs370395617 Chr.11: 66017131-66017131 on Build GRCh38
rs770416620 Chr.11: 66017114-66017116 on Build GRCh38
rs546845397 Chr.11: 66016925-66016925 on Build GRCh38
rs200328285 Chr.11: 66017102-66017102 on Build GRCh38
rs373893761 Chr.11: 66017024-66017024 on Build GRCh38
rs201027479 Chr.11: 66017196-66017196 on Build GRCh38
rs564708538 Chr.11: 66017232-66017232 on Build GRCh38
rs750051513 Chr.11: 66017135-66017135 on Build GRCh38
rs758701254 Chr.11: 66017186-66017186 on Build GRCh38
rs762896072 Chr.11: 66017122-66017122 on Build GRCh38
rs540544244 Chr.11: 66017345-66017345 on Build GRCh38
rs945039028 Chr.11: 66016970-66016970 on Build GRCh38
rs534081949 Chr.11: 66017093-66017093 on Build GRCh38
rs200011892 Chr.11: 66017091-66017091 on Build GRCh38
rs766174677 Chr.11: 66017212-66017212 on Build GRCh38
rs1006872369 Chr.11: 66017344-66017344 on Build GRCh38
rs566551492 Chr.11: 66016938-66016938 on Build GRCh38
rs549245302 Chr.11: 66017029-66017029 on Build GRCh38
rs368797173 Chr.11: 66017043-66017043 on Build GRCh38
rs750909314 Chr.11: 66017087-66017087 on Build GRCh38
rs1051704661 Chr.11: 66016982-66016982 on Build GRCh38
rs751160595 Chr.11: 66017078-66017078 on Build GRCh38
rs891238251 Chr.11: 66017125-66017125 on Build GRCh38
rs763905604 Chr.11: 66017075-66017075 on Build GRCh38
rs200453781 Chr.11: 66017208-66017208 on Build GRCh38
rs774626217 Chr.11: 66017186-66017187 on Build GRCh38
rs776064257 Chr.11: 66017151-66017152 on Build GRCh38
rs904135181 Chr.11: 66017279-66017279 on Build GRCh38
rs768625880 Chr.11: 66017123-66017123 on Build GRCh38
rs774195313 Chr.11: 66017130-66017130 on Build GRCh38
rs370297715 Chr.11: 66017031-66017031 on Build GRCh38
rs867832719 Chr.11: 66016968-66016968 on Build GRCh38
rs941536477 Chr.11: 66017240-66017240 on Build GRCh38
rs969108212 Chr.11: 66017368-66017368 on Build GRCh38
rs375937366 Chr.11: 66017028-66017028 on Build GRCh38
rs753448935 Chr.11: 66017177-66017177 on Build GRCh38
rs895306245 Chr.11: 66017073-66017073 on Build GRCh38
rs141424306 Chr.11: 66017032-66017032 on Build GRCh38
rs767966250 Chr.11: 66017365-66017365 on Build GRCh38
rs760225234 Chr.11: 66017159-66017159 on Build GRCh38
rs762666546 Chr.11: 66017070-66017070 on Build GRCh38
rs150841034 Chr.11: 66017192-66017192 on Build GRCh38
rs567374011 Chr.11: 66016965-66016965 on Build GRCh38
rs776277157 Chr.11: 66017222-66017222 on Build GRCh38
rs780772575 Chr.11: 66016964-66016964 on Build GRCh38
rs749940054 Chr.11: 66017086-66017086 on Build GRCh38
rs1038503832 Chr.11: 66017243-66017243 on Build GRCh38
rs918872786 Chr.11: 66017257-66017257 on Build GRCh38
rs761500535 Chr.11: 66017083-66017083 on Build GRCh38
rs142726200 Chr.11: 66017092-66017092 on Build GRCh38
rs918556814 Chr.11: 66017230-66017231 on Build GRCh38
rs761673284 Chr.11: 66017187-66017187 on Build GRCh38
rs999776959 Chr.11: 66017304-66017304 on Build GRCh38
rs142474326 Chr.11: 66017132-66017132 on Build GRCh38
rs866176124 Chr.11: 66017040-66017040 on Build GRCh38
rs200533904 Chr.11: 66017164-66017164 on Build GRCh38
rs112873572 Chr.11: 66017353-66017353 on Build GRCh38
rs1030657463 Chr.11: 66017067-66017067 on Build GRCh38
rs1051560626 Chr.11: 66017333-66017333 on Build GRCh38
rs757275617 Chr.11: 66017189-66017189 on Build GRCh38
rs755511932 Chr.11: 66017178-66017178 on Build GRCh38
rs748414808 Chr.11: 66017230-66017230 on Build GRCh38
rs889918442 Chr.11: 66017004-66017004 on Build GRCh38
rs201854224 Chr.11: 66017193-66017193 on Build GRCh38

View Full Product Details