Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66020972-66021466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER1
Entrez Gene ID
117144
Gene Name
cation channel sperm associated 1
Gene Aliases
CATSPER, SPGF7
UniGene
Hs.189105
Gene Chromosome Location
Chr.11: 66016752-66026518 on Build GRCh38
Transcripts
NM_053054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2166990 c.1780C>A p.L594I Chr.11: 66021097-66021097 on Build GRCh38
5958906 c.1772_1773delTT p.F591fs*18 Chr.11: 66021104-66021105 on Build GRCh38
6996377 c.1750G>A p.A584T Chr.11: 66021127-66021127 on Build GRCh38
1317275 c.1709A>G p.Q570R Chr.11: 66021168-66021168 on Build GRCh38
117579 c.1774A>C p.T592P Chr.11: 66021103-66021103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537436804 Chr.11: 66021062-66021062 on Build GRCh38
rs759927634 Chr.11: 66021135-66021135 on Build GRCh38
rs764420327 Chr.11: 66021421-66021421 on Build GRCh38
rs114665581 Chr.11: 66021408-66021408 on Build GRCh38
rs768681559 Chr.11: 66021072-66021072 on Build GRCh38
rs1041961631 Chr.11: 66021392-66021392 on Build GRCh38
rs757275539 Chr.11: 66021178-66021178 on Build GRCh38
rs201501413 Chr.11: 66021210-66021210 on Build GRCh38
rs962175260 Chr.11: 66020998-66020998 on Build GRCh38
rs763320317 Chr.11: 66021171-66021171 on Build GRCh38
rs946287037 Chr.11: 66021071-66021071 on Build GRCh38
rs766568220 Chr.11: 66021158-66021158 on Build GRCh38
rs765409156 Chr.11: 66021343-66021343 on Build GRCh38
rs771796187 Chr.11: 66021121-66021121 on Build GRCh38
rs528800312 Chr.11: 66021034-66021034 on Build GRCh38
rs914860870 Chr.11: 66021061-66021062 on Build GRCh38
rs953755385 Chr.11: 66021405-66021405 on Build GRCh38
rs371032491 Chr.11: 66021230-66021230 on Build GRCh38
rs533069171 Chr.11: 66021439-66021439 on Build GRCh38
rs199596488 Chr.11: 66021195-66021195 on Build GRCh38
rs373925688 Chr.11: 66021218-66021218 on Build GRCh38
rs199950402 Chr.11: 66021103-66021103 on Build GRCh38
rs202136090 Chr.11: 66021151-66021151 on Build GRCh38
rs990370665 Chr.11: 66021379-66021379 on Build GRCh38
rs200751031 Chr.11: 66021227-66021227 on Build GRCh38
rs868350071 Chr.11: 66021120-66021120 on Build GRCh38
rs760918593 Chr.11: 66021005-66021005 on Build GRCh38
rs1001245813 Chr.11: 66021263-66021263 on Build GRCh38
rs564210848 Chr.11: 66021431-66021431 on Build GRCh38
rs758544292 Chr.11: 66021004-66021004 on Build GRCh38
rs765838269 Chr.11: 66021148-66021148 on Build GRCh38
rs773003205 Chr.11: 66021128-66021128 on Build GRCh38
rs778027252 Chr.11: 66021005-66021005 on Build GRCh38
rs907098087 Chr.11: 66021258-66021258 on Build GRCh38
rs570606904 Chr.11: 66021173-66021173 on Build GRCh38
rs986530834 Chr.11: 66021087-66021087 on Build GRCh38
rs200565668 Chr.11: 66021051-66021051 on Build GRCh38
rs778571427 Chr.11: 66021095-66021095 on Build GRCh38
rs764230958 Chr.11: 66021172-66021172 on Build GRCh38
rs566239965 Chr.11: 66021012-66021012 on Build GRCh38
rs953577725 Chr.11: 66021362-66021362 on Build GRCh38
rs965995806 Chr.11: 66021247-66021247 on Build GRCh38
rs754896423 Chr.11: 66021222-66021223 on Build GRCh38
rs976039286 Chr.11: 66021256-66021256 on Build GRCh38
rs148057542 Chr.11: 66021134-66021134 on Build GRCh38
rs201420941 Chr.11: 66020994-66020994 on Build GRCh38
rs569088676 Chr.11: 66021228-66021228 on Build GRCh38
rs781223724 Chr.11: 66021184-66021184 on Build GRCh38
rs928303195 Chr.11: 66021395-66021395 on Build GRCh38
rs370610330 Chr.11: 66021216-66021216 on Build GRCh38
rs887562713 Chr.11: 66021286-66021286 on Build GRCh38
rs763107436 Chr.11: 66021401-66021401 on Build GRCh38
rs976349185 Chr.11: 66021011-66021011 on Build GRCh38
rs368518822 Chr.11: 66021127-66021127 on Build GRCh38
rs111960185 Chr.11: 66021380-66021380 on Build GRCh38
rs976110886 Chr.11: 66021032-66021032 on Build GRCh38
rs548838338 Chr.11: 66021048-66021048 on Build GRCh38
rs760223439 Chr.11: 66021129-66021129 on Build GRCh38
rs770588188 Chr.11: 66021130-66021130 on Build GRCh38
rs958036762 Chr.11: 66021432-66021432 on Build GRCh38
rs753917503 Chr.11: 66021194-66021196 on Build GRCh38
rs1006395343 Chr.11: 66021304-66021304 on Build GRCh38
rs772024892 Chr.11: 66021220-66021220 on Build GRCh38
rs962206468 Chr.11: 66021370-66021370 on Build GRCh38
rs371460795 Chr.11: 66021049-66021049 on Build GRCh38
rs1055849301 Chr.11: 66021264-66021264 on Build GRCh38
rs946326709 Chr.11: 66021391-66021391 on Build GRCh38
rs759143489 Chr.11: 66021445-66021445 on Build GRCh38
rs750585156 Chr.11: 66021188-66021188 on Build GRCh38

View Full Product Details