Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66041358-66041855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST3
Entrez Gene ID
89792
Gene Name
galactose-3-O-sulfotransferase 3
Gene Aliases
GAL3ST-3, GAL3ST2
UniGene
Hs.208343
Gene Chromosome Location
Chr.11: 66040765-66049180 on Build GRCh38
Transcripts
NM_033036.2 XM_017018519.1

SNP Detail

SNP ID Chromosome Location
rs369683301 Chr.11: 66041596-66041596 on Build GRCh38
rs976309575 Chr.11: 66041445-66041445 on Build GRCh38
rs912373364 Chr.11: 66041456-66041456 on Build GRCh38
rs1057016046 Chr.11: 66041558-66041558 on Build GRCh38
rs942510646 Chr.11: 66041804-66041804 on Build GRCh38
rs750861344 Chr.11: 66041672-66041672 on Build GRCh38
rs1041743113 Chr.11: 66041526-66041526 on Build GRCh38
rs1012515338 Chr.11: 66041585-66041585 on Build GRCh38
rs181693607 Chr.11: 66041813-66041813 on Build GRCh38
rs902587016 Chr.11: 66041494-66041494 on Build GRCh38
rs958845108 Chr.11: 66041618-66041618 on Build GRCh38
rs1013161167 Chr.11: 66041631-66041631 on Build GRCh38
rs1027871598 Chr.11: 66041590-66041590 on Build GRCh38
rs965941566 Chr.11: 66041433-66041433 on Build GRCh38
rs942624777 Chr.11: 66041479-66041479 on Build GRCh38
rs776625139 Chr.11: 66041418-66041418 on Build GRCh38
rs907575691 Chr.11: 66041431-66041431 on Build GRCh38
rs758081446 Chr.11: 66041532-66041532 on Build GRCh38
rs914489098 Chr.11: 66041795-66041795 on Build GRCh38
rs79912656 Chr.11: 66041722-66041722 on Build GRCh38
rs919354842 Chr.11: 66041421-66041421 on Build GRCh38
rs143434275 Chr.11: 66041611-66041611 on Build GRCh38
rs919226968 Chr.11: 66041702-66041702 on Build GRCh38
rs145828054 Chr.11: 66041775-66041775 on Build GRCh38
rs751263054 Chr.11: 66041393-66041396 on Build GRCh38
rs1042767719 Chr.11: 66041812-66041812 on Build GRCh38
rs946681607 Chr.11: 66041461-66041461 on Build GRCh38
rs1039626716 Chr.11: 66041488-66041488 on Build GRCh38
rs566504065 Chr.11: 66041428-66041428 on Build GRCh38
rs764742265 Chr.11: 66041454-66041454 on Build GRCh38
rs951694195 Chr.11: 66041608-66041608 on Build GRCh38
rs914502973 Chr.11: 66041816-66041816 on Build GRCh38
rs568861498 Chr.11: 66041514-66041514 on Build GRCh38
rs987542143 Chr.11: 66041721-66041721 on Build GRCh38
rs553246084 Chr.11: 66041729-66041729 on Build GRCh38
rs56325022 Chr.11: 66041451-66041451 on Build GRCh38
rs902936335 Chr.11: 66041555-66041555 on Build GRCh38
rs756500363 Chr.11: 66041680-66041680 on Build GRCh38
rs892833133 Chr.11: 66041495-66041495 on Build GRCh38
rs963928703 Chr.11: 66041662-66041662 on Build GRCh38
rs975835995 Chr.11: 66041651-66041651 on Build GRCh38
rs566205781 Chr.11: 66041505-66041505 on Build GRCh38
rs561298028 Chr.11: 66041565-66041566 on Build GRCh38
rs190585401 Chr.11: 66041704-66041704 on Build GRCh38
rs763818847 Chr.11: 66041669-66041669 on Build GRCh38
rs1003006369 Chr.11: 66041560-66041560 on Build GRCh38
rs1016459723 Chr.11: 66041638-66041638 on Build GRCh38
rs1014707252 Chr.11: 66041622-66041622 on Build GRCh38
rs966170049 Chr.11: 66041625-66041625 on Build GRCh38

View Full Product Details